Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3795662-3796155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25B
Entrez Gene ID
994
Gene Name
cell division cycle 25B
Gene Aliases
-
UniGene
Hs.153752
Gene Chromosome Location
Chr.20: 3786772-3806121 on Build GRCh38
Transcripts
NM_001287516.1 NM_001287517.1 NM_001287518.1 NM_001287519.1 NM_001287520.1 NM_001287522.1 NM_004358.4 NM_021872.3 NM_021873.3 NR_136335.1 NR_136336.1

SNP Detail

SNP ID Chromosome Location
rs1005683821 Chr.20: 3796032-3796032 on Build GRCh38
rs960899690 Chr.20: 3796098-3796098 on Build GRCh38
rs769565371 Chr.20: 3795935-3795935 on Build GRCh38
rs956713910 Chr.20: 3795845-3795845 on Build GRCh38
rs774599012 Chr.20: 3795994-3795994 on Build GRCh38
rs931382006 Chr.20: 3795971-3795971 on Build GRCh38
rs934626503 Chr.20: 3795866-3795866 on Build GRCh38
rs1003045422 Chr.20: 3795684-3795684 on Build GRCh38
rs896921707 Chr.20: 3796060-3796060 on Build GRCh38
rs903606193 Chr.20: 3795798-3795798 on Build GRCh38
rs577933184 Chr.20: 3795793-3795793 on Build GRCh38
rs901411049 Chr.20: 3795942-3795942 on Build GRCh38
rs1049884490 Chr.20: 3795992-3795992 on Build GRCh38
rs537551827 Chr.20: 3795737-3795737 on Build GRCh38
rs908631980 Chr.20: 3796021-3796021 on Build GRCh38
rs1057318880 Chr.20: 3796095-3796095 on Build GRCh38
rs986666073 Chr.20: 3795856-3795856 on Build GRCh38
rs542271835 Chr.20: 3795969-3795969 on Build GRCh38
rs960679217 Chr.20: 3795714-3795714 on Build GRCh38
rs1057496151 Chr.20: 3795685-3795685 on Build GRCh38
rs775434656 Chr.20: 3795940-3795940 on Build GRCh38
rs183999341 Chr.20: 3795822-3795822 on Build GRCh38
rs560474488 Chr.20: 3795874-3795874 on Build GRCh38
rs1001062786 Chr.20: 3795812-3795812 on Build GRCh38
rs938819844 Chr.20: 3796065-3796065 on Build GRCh38
rs994222036 Chr.20: 3796077-3796077 on Build GRCh38
rs1034720015 Chr.20: 3796084-3796084 on Build GRCh38
rs1035325965 Chr.20: 3795686-3795686 on Build GRCh38
rs3761219 Chr.20: 3795721-3795721 on Build GRCh38
rs948780252 Chr.20: 3795757-3795757 on Build GRCh38
rs11569978 Chr.20: 3795911-3795911 on Build GRCh38
rs926032588 Chr.20: 3795859-3795859 on Build GRCh38
rs576500910 Chr.20: 3795728-3795728 on Build GRCh38
rs374054731 Chr.20: 3796004-3796004 on Build GRCh38
rs975395652 Chr.20: 3795949-3795949 on Build GRCh38
rs1016610167 Chr.20: 3796034-3796034 on Build GRCh38
rs3761220 Chr.20: 3795753-3795753 on Build GRCh38
rs894717779 Chr.20: 3796104-3796104 on Build GRCh38

View Full Product Details