Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33081280-33081779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
UniGene
Hs.38961
Gene Chromosome Location
Chr.20: 33081512-33111751 on Build GRCh38
Transcripts
NM_182519.2

SNP Detail

SNP ID Chromosome Location
rs972337898 Chr.20: 33081672-33081672 on Build GRCh38
rs949821305 Chr.20: 33081430-33081430 on Build GRCh38
rs150164177 Chr.20: 33081612-33081612 on Build GRCh38
rs563359107 Chr.20: 33081450-33081450 on Build GRCh38
rs755847159 Chr.20: 33081609-33081609 on Build GRCh38
rs764452707 Chr.20: 33081586-33081586 on Build GRCh38
rs200039664 Chr.20: 33081462-33081462 on Build GRCh38
rs771454392 Chr.20: 33081537-33081537 on Build GRCh38
rs935681030 Chr.20: 33081474-33081474 on Build GRCh38
rs974173510 Chr.20: 33081309-33081309 on Build GRCh38
rs927042183 Chr.20: 33081717-33081717 on Build GRCh38
rs971242283 Chr.20: 33081688-33081688 on Build GRCh38
rs539611757 Chr.20: 33081428-33081428 on Build GRCh38
rs918248315 Chr.20: 33081681-33081681 on Build GRCh38
rs221984 Chr.20: 33081711-33081711 on Build GRCh38
rs142229443 Chr.20: 33081473-33081473 on Build GRCh38
rs759873534 Chr.20: 33081556-33081556 on Build GRCh38
rs386813765 Chr.20: 33081461-33081462 on Build GRCh38
rs749119935 Chr.20: 33081635-33081635 on Build GRCh38
rs376219070 Chr.20: 33081466-33081466 on Build GRCh38
rs548034063 Chr.20: 33081445-33081445 on Build GRCh38
rs533582738 Chr.20: 33081497-33081497 on Build GRCh38
rs777582904 Chr.20: 33081499-33081500 on Build GRCh38
rs221985 Chr.20: 33081738-33081738 on Build GRCh38
rs750333125 Chr.20: 33081604-33081604 on Build GRCh38
rs896329872 Chr.20: 33081424-33081424 on Build GRCh38
rs953563689 Chr.20: 33081648-33081648 on Build GRCh38
rs1000778360 Chr.20: 33081522-33081522 on Build GRCh38
rs137996770 Chr.20: 33081461-33081461 on Build GRCh38
rs918249110 Chr.20: 33081339-33081339 on Build GRCh38
rs902050810 Chr.20: 33081600-33081600 on Build GRCh38
rs935614228 Chr.20: 33081737-33081737 on Build GRCh38
rs916949641 Chr.20: 33081376-33081376 on Build GRCh38
rs751640010 Chr.20: 33081587-33081587 on Build GRCh38
rs778593596 Chr.20: 33081669-33081669 on Build GRCh38
rs767145060 Chr.20: 33081602-33081602 on Build GRCh38
rs779848889 Chr.20: 33081613-33081613 on Build GRCh38
rs765706851 Chr.20: 33081569-33081569 on Build GRCh38
rs221983 Chr.20: 33081661-33081661 on Build GRCh38
rs929752370 Chr.20: 33081355-33081355 on Build GRCh38
rs201651044 Chr.20: 33081578-33081578 on Build GRCh38
rs1056189732 Chr.20: 33081410-33081410 on Build GRCh38
rs892050508 Chr.20: 33081504-33081504 on Build GRCh38
rs757343492 Chr.20: 33081592-33081592 on Build GRCh38
rs1006194482 Chr.20: 33081606-33081606 on Build GRCh38
rs991923983 Chr.20: 33081720-33081720 on Build GRCh38
rs986284498 Chr.20: 33081659-33081659 on Build GRCh38
rs775802423 Chr.20: 33081574-33081574 on Build GRCh38
rs767485471 Chr.20: 33081601-33081601 on Build GRCh38
rs752760419 Chr.20: 33081552-33081552 on Build GRCh38

View Full Product Details