Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36175462-36175973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L1
Entrez Gene ID
2036
Gene Name
erythrocyte membrane protein band 4.1 like 1
Gene Aliases
4.1N, MRD11
UniGene
Hs.437422
Gene Chromosome Location
Chr.20: 36064891-36232799 on Build GRCh38
Transcripts
NM_001258329.1 NM_001258330.1 NM_001258331.1 NM_012156.2 NM_177996.2 XM_011528667.1 XM_011528671.1 XM_011528673.1 XM_011528677.1 XM_011528681.1 XM_011528682.1 XM_011528683.1 XM_011528685.1 XM_017027711.1 XM_017027712.1 XM_017027713.1 XM_017027714.1 XM_017027715.1 XM_017027716.1 XM_017027717.1 XM_017027718.1 XM_017027719.1 XM_017027720.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5007681 c.258G>A p.S86S Chr.20: 36175631-36175631 on Build GRCh38
5468031 c.244G>A p.V82M Chr.20: 36175710-36175710 on Build GRCh38
5857770 c.164C>T p.S55L Chr.20: 36175630-36175630 on Build GRCh38
4097979 c.210G>A p.E70E Chr.20: 36175583-36175583 on Build GRCh38
4485222 c.290C>T p.A97V Chr.20: 36175663-36175663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777985190 Chr.20: 36175532-36175532 on Build GRCh38
rs769300932 Chr.20: 36175694-36175694 on Build GRCh38
rs754822027 Chr.20: 36175630-36175630 on Build GRCh38
rs200228097 Chr.20: 36175745-36175745 on Build GRCh38
rs771586848 Chr.20: 36175712-36175712 on Build GRCh38
rs569528757 Chr.20: 36175855-36175855 on Build GRCh38
rs756160954 Chr.20: 36175644-36175644 on Build GRCh38
rs758262452 Chr.20: 36175519-36175519 on Build GRCh38
rs931414124 Chr.20: 36175856-36175856 on Build GRCh38
rs764130223 Chr.20: 36175758-36175758 on Build GRCh38
rs777736229 Chr.20: 36175647-36175647 on Build GRCh38
rs1041780631 Chr.20: 36175825-36175825 on Build GRCh38
rs762619496 Chr.20: 36175609-36175609 on Build GRCh38
rs547950907 Chr.20: 36175751-36175751 on Build GRCh38
rs746221638 Chr.20: 36175673-36175673 on Build GRCh38
rs543226919 Chr.20: 36175491-36175491 on Build GRCh38
rs374593631 Chr.20: 36175504-36175504 on Build GRCh38
rs927138687 Chr.20: 36175703-36175703 on Build GRCh38
rs372360979 Chr.20: 36175744-36175744 on Build GRCh38
rs764956865 Chr.20: 36175823-36175823 on Build GRCh38
rs918718841 Chr.20: 36175519-36175519 on Build GRCh38
rs772631937 Chr.20: 36175722-36175722 on Build GRCh38
rs759257401 Chr.20: 36175750-36175750 on Build GRCh38
rs369804239 Chr.20: 36175693-36175693 on Build GRCh38
rs757523307 Chr.20: 36175668-36175668 on Build GRCh38
rs550250335 Chr.20: 36175944-36175944 on Build GRCh38
rs912581406 Chr.20: 36175487-36175487 on Build GRCh38
rs912528797 Chr.20: 36175783-36175783 on Build GRCh38
rs148961505 Chr.20: 36175665-36175665 on Build GRCh38
rs368127593 Chr.20: 36175560-36175560 on Build GRCh38
rs531650721 Chr.20: 36175914-36175914 on Build GRCh38
rs771372739 Chr.20: 36175538-36175538 on Build GRCh38
rs769827625 Chr.20: 36175517-36175517 on Build GRCh38
rs748241759 Chr.20: 36175506-36175506 on Build GRCh38
rs1019121649 Chr.20: 36175943-36175943 on Build GRCh38
rs985393637 Chr.20: 36175764-36175764 on Build GRCh38
rs760331342 Chr.20: 36175841-36175841 on Build GRCh38
rs761382038 Chr.20: 36175587-36175587 on Build GRCh38
rs772632635 Chr.20: 36175674-36175674 on Build GRCh38
rs543552366 Chr.20: 36175597-36175597 on Build GRCh38
rs375649709 Chr.20: 36175692-36175692 on Build GRCh38
rs1009873156 Chr.20: 36175785-36175785 on Build GRCh38
rs772866145 Chr.20: 36175598-36175598 on Build GRCh38
rs774053054 Chr.20: 36175747-36175747 on Build GRCh38
rs756034449 Chr.20: 36175757-36175757 on Build GRCh38
rs1031427659 Chr.20: 36175660-36175660 on Build GRCh38
rs779328780 Chr.20: 36175671-36175671 on Build GRCh38
rs752631065 Chr.20: 36175754-36175754 on Build GRCh38
rs772685833 Chr.20: 36175579-36175579 on Build GRCh38
rs774885999 Chr.20: 36175555-36175555 on Build GRCh38
rs1002994858 Chr.20: 36175651-36175651 on Build GRCh38
rs540713849 Chr.20: 36175670-36175670 on Build GRCh38
rs191128724 Chr.20: 36175586-36175586 on Build GRCh38
rs749484412 Chr.20: 36175534-36175534 on Build GRCh38
rs953602447 Chr.20: 36175561-36175561 on Build GRCh38
rs372366944 Chr.20: 36175610-36175610 on Build GRCh38
rs1009101284 Chr.20: 36175926-36175926 on Build GRCh38
rs145676614 Chr.20: 36175502-36175502 on Build GRCh38
rs1000366340 Chr.20: 36175899-36175899 on Build GRCh38
rs1029105355 Chr.20: 36175909-36175909 on Build GRCh38
rs937220998 Chr.20: 36175743-36175743 on Build GRCh38
rs148330736 Chr.20: 36175642-36175642 on Build GRCh38
rs973181813 Chr.20: 36175830-36175830 on Build GRCh38
rs767481392 Chr.20: 36175631-36175631 on Build GRCh38
rs143214838 Chr.20: 36175641-36175641 on Build GRCh38
rs916759559 Chr.20: 36175879-36175879 on Build GRCh38
rs762263477 Chr.20: 36175672-36175672 on Build GRCh38
rs1048544314 Chr.20: 36175863-36175863 on Build GRCh38

View Full Product Details