Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46121736-46122245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD40
Entrez Gene ID
958
Gene Name
CD40 molecule
Gene Aliases
Bp50, CDW40, TNFRSF5, p50
UniGene
Hs.472860
Gene Chromosome Location
Chr.20: 46118242-46129745 on Build GRCh38
Transcripts
NM_001250.5 NM_001302753.1 NM_001322421.1 NM_001322422.1 NM_152854.3 NR_126502.1 NR_136327.1 XM_005260619.3 XM_011529109.2 XM_017028135.1 XM_017028136.1 XM_017028137.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3547202 c.130+1G>A p.? Chr.20: 46121899-46121899 on Build GRCh38
3294358 c.128C>T p.P43L Chr.20: 46121896-46121896 on Build GRCh38
1165834 c.69C>T p.P23P Chr.20: 46121837-46121837 on Build GRCh38
443931 c.106C>T p.Q36* Chr.20: 46121874-46121874 on Build GRCh38
1412156 c.131G>T p.G44V Chr.20: 46122233-46122233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1013263761 Chr.20: 46122149-46122149 on Build GRCh38
rs775472655 Chr.20: 46121843-46121843 on Build GRCh38
rs748859483 Chr.20: 46122040-46122040 on Build GRCh38
rs923841997 Chr.20: 46121994-46121994 on Build GRCh38
rs61508870 Chr.20: 46121801-46121801 on Build GRCh38
rs771363187 Chr.20: 46121822-46121822 on Build GRCh38
rs112055473 Chr.20: 46122096-46122096 on Build GRCh38
rs755760762 Chr.20: 46121926-46121926 on Build GRCh38
rs371933529 Chr.20: 46122190-46122190 on Build GRCh38
rs969654773 Chr.20: 46122182-46122182 on Build GRCh38
rs979346762 Chr.20: 46121981-46121981 on Build GRCh38
rs547183856 Chr.20: 46121950-46121950 on Build GRCh38
rs754914712 Chr.20: 46121912-46121912 on Build GRCh38
rs892095824 Chr.20: 46122139-46122139 on Build GRCh38
rs187683423 Chr.20: 46121805-46121805 on Build GRCh38
rs192657612 Chr.20: 46122141-46122141 on Build GRCh38
rs1017859412 Chr.20: 46121923-46121923 on Build GRCh38
rs774936391 Chr.20: 46121824-46121824 on Build GRCh38
rs768929858 Chr.20: 46122210-46122210 on Build GRCh38
rs764464873 Chr.20: 46121863-46121863 on Build GRCh38
rs903587677 Chr.20: 46122126-46122126 on Build GRCh38
rs567636402 Chr.20: 46122130-46122130 on Build GRCh38
rs760957101 Chr.20: 46121852-46121852 on Build GRCh38
rs759072777 Chr.20: 46122101-46122101 on Build GRCh38
rs1025230336 Chr.20: 46122165-46122165 on Build GRCh38
rs147677886 Chr.20: 46121841-46121841 on Build GRCh38
rs749599565 Chr.20: 46121942-46121942 on Build GRCh38
rs908516604 Chr.20: 46121995-46121995 on Build GRCh38
rs569971779 Chr.20: 46122140-46122140 on Build GRCh38
rs754541736 Chr.20: 46121783-46121783 on Build GRCh38
rs770408810 Chr.20: 46122082-46122082 on Build GRCh38
rs777268106 Chr.20: 46121794-46121794 on Build GRCh38
rs766169434 Chr.20: 46122221-46122221 on Build GRCh38
rs566004958 Chr.20: 46122030-46122030 on Build GRCh38
rs749720262 Chr.20: 46121807-46121807 on Build GRCh38
rs762843067 Chr.20: 46121872-46121872 on Build GRCh38
rs11569317 Chr.20: 46122211-46122211 on Build GRCh38
rs113777961 Chr.20: 46122164-46122164 on Build GRCh38
rs759483970 Chr.20: 46121833-46121833 on Build GRCh38
rs555441743 Chr.20: 46121967-46121967 on Build GRCh38
rs919989132 Chr.20: 46121956-46121956 on Build GRCh38
rs374336021 Chr.20: 46122192-46122192 on Build GRCh38
rs751549823 Chr.20: 46121891-46121891 on Build GRCh38
rs944045387 Chr.20: 46122010-46122010 on Build GRCh38
rs752265727 Chr.20: 46121922-46121922 on Build GRCh38
rs11569316 Chr.20: 46122128-46122128 on Build GRCh38
rs750963838 Chr.20: 46121776-46121776 on Build GRCh38
rs774097104 Chr.20: 46122085-46122085 on Build GRCh38
rs780735906 Chr.20: 46121917-46121917 on Build GRCh38
rs765014005 Chr.20: 46122203-46122203 on Build GRCh38
rs952858244 Chr.20: 46121957-46121957 on Build GRCh38
rs766167612 Chr.20: 46121883-46121883 on Build GRCh38
rs750234130 Chr.20: 46121871-46121871 on Build GRCh38
rs1050296254 Chr.20: 46122120-46122120 on Build GRCh38
rs757665188 Chr.20: 46121948-46121948 on Build GRCh38
rs11569315 Chr.20: 46121785-46121785 on Build GRCh38
rs772505094 Chr.20: 46122194-46122194 on Build GRCh38
rs777407852 Chr.20: 46121937-46121937 on Build GRCh38
rs374320594 Chr.20: 46121955-46121955 on Build GRCh38
rs539361776 Chr.20: 46122058-46122058 on Build GRCh38
rs184310063 Chr.20: 46122147-46122147 on Build GRCh38
rs747149466 Chr.20: 46122202-46122202 on Build GRCh38

View Full Product Details