Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51600571-51601074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP9A
Entrez Gene ID
10079
Gene Name
ATPase phospholipid transporting 9A (putative)
Gene Aliases
ATPIIA
UniGene
Hs.649234
Gene Chromosome Location
Chr.20: 51596775-51768411 on Build GRCh38
Transcripts
NM_006045.2 XM_011528481.2

SNP Detail

SNP ID Chromosome Location
rs780067736 Chr.20: 51600806-51600807 on Build GRCh38
rs911171221 Chr.20: 51601054-51601054 on Build GRCh38
rs577861590 Chr.20: 51600997-51600997 on Build GRCh38
rs770710043 Chr.20: 51600804-51600807 on Build GRCh38
rs1041431215 Chr.20: 51600800-51600800 on Build GRCh38
rs952617560 Chr.20: 51600756-51600756 on Build GRCh38
rs988348254 Chr.20: 51600903-51600903 on Build GRCh38
rs1027339527 Chr.20: 51600803-51600804 on Build GRCh38
rs35997419 Chr.20: 51600845-51600846 on Build GRCh38
rs139187110 Chr.20: 51600611-51600611 on Build GRCh38
rs553158819 Chr.20: 51600993-51600993 on Build GRCh38
rs527939730 Chr.20: 51600808-51600809 on Build GRCh38
rs1034981127 Chr.20: 51600643-51600643 on Build GRCh38
rs911105202 Chr.20: 51600779-51600779 on Build GRCh38
rs985749826 Chr.20: 51600817-51600817 on Build GRCh38
rs961102751 Chr.20: 51600672-51600672 on Build GRCh38
rs149924347 Chr.20: 51600807-51600808 on Build GRCh38
rs896042526 Chr.20: 51600894-51600894 on Build GRCh38
rs547637805 Chr.20: 51600660-51600660 on Build GRCh38
rs115114197 Chr.20: 51600887-51600887 on Build GRCh38
rs200571035 Chr.20: 51600804-51600805 on Build GRCh38
rs752935960 Chr.20: 51601044-51601044 on Build GRCh38
rs1034962613 Chr.20: 51600748-51600748 on Build GRCh38
rs567048215 Chr.20: 51600651-51600651 on Build GRCh38
rs557028535 Chr.20: 51601010-51601010 on Build GRCh38
rs142039865 Chr.20: 51600806-51600813 on Build GRCh38
rs745612657 Chr.20: 51600981-51600981 on Build GRCh38
rs1043124535 Chr.20: 51600847-51600847 on Build GRCh38
rs763366256 Chr.20: 51600620-51600620 on Build GRCh38
rs1013286120 Chr.20: 51600919-51600919 on Build GRCh38
rs972580324 Chr.20: 51600720-51600720 on Build GRCh38
rs755166706 Chr.20: 51600807-51600810 on Build GRCh38
rs748284530 Chr.20: 51600847-51600848 on Build GRCh38
rs904817716 Chr.20: 51600598-51600598 on Build GRCh38
rs946113169 Chr.20: 51600836-51600836 on Build GRCh38
rs575154915 Chr.20: 51601023-51601023 on Build GRCh38
rs910181856 Chr.20: 51600841-51600845 on Build GRCh38
rs186664211 Chr.20: 51600987-51600987 on Build GRCh38
rs140474270 Chr.20: 51601009-51601009 on Build GRCh38
rs186754066 Chr.20: 51600663-51600663 on Build GRCh38
rs770261113 Chr.20: 51600599-51600599 on Build GRCh38
rs149939354 Chr.20: 51600771-51600771 on Build GRCh38
rs919762576 Chr.20: 51600750-51600750 on Build GRCh38
rs182335221 Chr.20: 51600928-51600928 on Build GRCh38
rs79346952 Chr.20: 51600807-51600807 on Build GRCh38
rs1002081575 Chr.20: 51600886-51600886 on Build GRCh38
rs879031295 Chr.20: 51600806-51600809 on Build GRCh38
rs1052509792 Chr.20: 51600630-51600630 on Build GRCh38
rs567415124 Chr.20: 51600975-51600975 on Build GRCh38
rs944078328 Chr.20: 51600796-51600796 on Build GRCh38
rs951836957 Chr.20: 51600806-51600821 on Build GRCh38
rs1003831115 Chr.20: 51600734-51600734 on Build GRCh38
rs546815086 Chr.20: 51600644-51600644 on Build GRCh38
rs946749594 Chr.20: 51600948-51600948 on Build GRCh38
rs190994320 Chr.20: 51600766-51600766 on Build GRCh38
rs965761052 Chr.20: 51600843-51600845 on Build GRCh38
rs145008765 Chr.20: 51600871-51600871 on Build GRCh38
rs542231035 Chr.20: 51601049-51601049 on Build GRCh38
rs57250019 Chr.20: 51600846-51600847 on Build GRCh38
rs540147579 Chr.20: 51600763-51600763 on Build GRCh38
rs759786103 Chr.20: 51600805-51600806 on Build GRCh38
rs1027477866 Chr.20: 51601043-51601043 on Build GRCh38
rs1013647032 Chr.20: 51600659-51600659 on Build GRCh38

View Full Product Details