Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33624584-33625102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306119 c.115G>A p.A39T Chr.21: 33624712-33624712 on Build GRCh38
3701683 c.144+3A>G p.? Chr.21: 33624680-33624680 on Build GRCh38
4101390 c.87G>A p.E29E Chr.21: 33624740-33624740 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008932280 Chr.21: 33624679-33624679 on Build GRCh38
rs750407402 Chr.21: 33624908-33624908 on Build GRCh38
rs991104627 Chr.21: 33624785-33624786 on Build GRCh38
rs972712108 Chr.21: 33624781-33624781 on Build GRCh38
rs775091857 Chr.21: 33624628-33624628 on Build GRCh38
rs201409581 Chr.21: 33624734-33624734 on Build GRCh38
rs747707916 Chr.21: 33624784-33624784 on Build GRCh38
rs983836699 Chr.21: 33624798-33624798 on Build GRCh38
rs781287785 Chr.21: 33624680-33624681 on Build GRCh38
rs201761848 Chr.21: 33624805-33624805 on Build GRCh38
rs763784702 Chr.21: 33624616-33624616 on Build GRCh38
rs752865721 Chr.21: 33624677-33624677 on Build GRCh38
rs774868273 Chr.21: 33624793-33624793 on Build GRCh38
rs114404532 Chr.21: 33624613-33624613 on Build GRCh38
rs746128901 Chr.21: 33624813-33624814 on Build GRCh38
rs767738473 Chr.21: 33624675-33624675 on Build GRCh38
rs760751058 Chr.21: 33624682-33624682 on Build GRCh38
rs560815978 Chr.21: 33624654-33624654 on Build GRCh38
rs183243463 Chr.21: 33624612-33624612 on Build GRCh38
rs201072452 Chr.21: 33624708-33624708 on Build GRCh38
rs935782214 Chr.21: 33624982-33624982 on Build GRCh38
rs111248812 Chr.21: 33624729-33624729 on Build GRCh38
rs768068825 Chr.21: 33624639-33624639 on Build GRCh38
rs757226936 Chr.21: 33624704-33624704 on Build GRCh38
rs779702938 Chr.21: 33624738-33624738 on Build GRCh38
rs1011023848 Chr.21: 33625031-33625031 on Build GRCh38
rs765422716 Chr.21: 33625058-33625058 on Build GRCh38
rs771377329 Chr.21: 33624788-33624788 on Build GRCh38
rs775976976 Chr.21: 33624807-33624807 on Build GRCh38
rs369128909 Chr.21: 33624640-33624640 on Build GRCh38
rs878857871 Chr.21: 33624897-33624897 on Build GRCh38
rs188750338 Chr.21: 33624948-33624948 on Build GRCh38
rs916885158 Chr.21: 33624920-33624920 on Build GRCh38
rs764064388 Chr.21: 33624686-33624686 on Build GRCh38
rs893906538 Chr.21: 33624983-33624983 on Build GRCh38
rs746627994 Chr.21: 33624748-33624748 on Build GRCh38
rs555878501 Chr.21: 33625015-33625015 on Build GRCh38
rs754727597 Chr.21: 33624763-33624763 on Build GRCh38
rs1028978806 Chr.21: 33625039-33625039 on Build GRCh38
rs1050242165 Chr.21: 33625001-33625001 on Build GRCh38
rs1029059237 Chr.21: 33624630-33624630 on Build GRCh38
rs756073822 Chr.21: 33625037-33625037 on Build GRCh38
rs149006249 Chr.21: 33624664-33624664 on Build GRCh38
rs760799412 Chr.21: 33624810-33624810 on Build GRCh38
rs754034368 Chr.21: 33624696-33624696 on Build GRCh38
rs112930012 Chr.21: 33624862-33624862 on Build GRCh38
rs746465045 Chr.21: 33624637-33624637 on Build GRCh38
rs866226131 Chr.21: 33624976-33624976 on Build GRCh38
rs150528653 Chr.21: 33624711-33624711 on Build GRCh38
rs143644594 Chr.21: 33624701-33624701 on Build GRCh38
rs13050238 Chr.21: 33624712-33624712 on Build GRCh38
rs759913984 Chr.21: 33624665-33624665 on Build GRCh38
rs746176837 Chr.21: 33624799-33624799 on Build GRCh38
rs904376346 Chr.21: 33624978-33624978 on Build GRCh38
rs992548489 Chr.21: 33624911-33624911 on Build GRCh38
rs770932596 Chr.21: 33624664-33624665 on Build GRCh38
rs949998050 Chr.21: 33624904-33624904 on Build GRCh38
rs889242666 Chr.21: 33625009-33625009 on Build GRCh38
rs550793039 Chr.21: 33624730-33624730 on Build GRCh38
rs112225000 Chr.21: 33624743-33624743 on Build GRCh38
rs1009377870 Chr.21: 33625032-33625032 on Build GRCh38
rs780650772 Chr.21: 33624764-33624764 on Build GRCh38
rs374426329 Chr.21: 33624646-33624646 on Build GRCh38

View Full Product Details