Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24225456-24225964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208149 c.1256G>A p.R419Q Chr.22: 24225626-24225626 on Build GRCh38
3552628 c.1261G>A p.G421S Chr.22: 24225621-24225621 on Build GRCh38
3552627 c.1311G>A p.R437R Chr.22: 24225571-24225571 on Build GRCh38
4103057 c.1317C>T p.S439S Chr.22: 24225565-24225565 on Build GRCh38
1683565 c.1275_1276insA p.N426fs*87 Chr.22: 24225606-24225607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200937485 Chr.22: 24225548-24225548 on Build GRCh38
rs972045743 Chr.22: 24225722-24225722 on Build GRCh38
rs752530955 Chr.22: 24225583-24225583 on Build GRCh38
rs562195268 Chr.22: 24225869-24225869 on Build GRCh38
rs977377508 Chr.22: 24225841-24225841 on Build GRCh38
rs529592678 Chr.22: 24225639-24225639 on Build GRCh38
rs776163245 Chr.22: 24225579-24225579 on Build GRCh38
rs368114799 Chr.22: 24225661-24225661 on Build GRCh38
rs534493457 Chr.22: 24225757-24225757 on Build GRCh38
rs1050149047 Chr.22: 24225857-24225857 on Build GRCh38
rs371281442 Chr.22: 24225565-24225565 on Build GRCh38
rs924850065 Chr.22: 24225764-24225764 on Build GRCh38
rs756289453 Chr.22: 24225588-24225588 on Build GRCh38
rs71318962 Chr.22: 24225735-24225735 on Build GRCh38
rs779215682 Chr.22: 24225622-24225622 on Build GRCh38
rs764492598 Chr.22: 24225741-24225741 on Build GRCh38
rs753960025 Chr.22: 24225690-24225690 on Build GRCh38
rs781195762 Chr.22: 24225524-24225524 on Build GRCh38
rs759124656 Chr.22: 24225581-24225581 on Build GRCh38
rs145909552 Chr.22: 24225604-24225604 on Build GRCh38
rs775196188 Chr.22: 24225660-24225660 on Build GRCh38
rs564017458 Chr.22: 24225664-24225664 on Build GRCh38
rs769456009 Chr.22: 24225644-24225644 on Build GRCh38
rs75273222 Chr.22: 24225572-24225572 on Build GRCh38
rs200073930 Chr.22: 24225618-24225618 on Build GRCh38
rs192160185 Chr.22: 24225506-24225506 on Build GRCh38
rs189178297 Chr.22: 24225723-24225723 on Build GRCh38
rs928685613 Chr.22: 24225598-24225598 on Build GRCh38
rs148922656 Chr.22: 24225554-24225554 on Build GRCh38
rs758448257 Chr.22: 24225496-24225519 on Build GRCh38
rs1044679349 Chr.22: 24225927-24225927 on Build GRCh38
rs1029261650 Chr.22: 24225695-24225695 on Build GRCh38
rs367693056 Chr.22: 24225569-24225569 on Build GRCh38
rs1021582574 Chr.22: 24225838-24225838 on Build GRCh38
rs868507043 Chr.22: 24225879-24225879 on Build GRCh38
rs748665403 Chr.22: 24225564-24225564 on Build GRCh38
rs371880180 Chr.22: 24225558-24225558 on Build GRCh38
rs183990532 Chr.22: 24225575-24225575 on Build GRCh38
rs756137997 Chr.22: 24225604-24225606 on Build GRCh38
rs774717334 Chr.22: 24225557-24225557 on Build GRCh38
rs1036855208 Chr.22: 24225740-24225740 on Build GRCh38
rs916055461 Chr.22: 24225620-24225620 on Build GRCh38
rs747475402 Chr.22: 24225627-24225627 on Build GRCh38
rs118007626 Chr.22: 24225756-24225756 on Build GRCh38
rs1052063709 Chr.22: 24225777-24225777 on Build GRCh38
rs143000629 Chr.22: 24225814-24225814 on Build GRCh38
rs568226124 Chr.22: 24225781-24225781 on Build GRCh38
rs372057047 Chr.22: 24225884-24225884 on Build GRCh38
rs756452604 Chr.22: 24225528-24225528 on Build GRCh38
rs761779669 Chr.22: 24225668-24225668 on Build GRCh38
rs925476156 Chr.22: 24225476-24225476 on Build GRCh38
rs780218835 Chr.22: 24225629-24225629 on Build GRCh38
rs940146463 Chr.22: 24225875-24225875 on Build GRCh38
rs56188355 Chr.22: 24225889-24225889 on Build GRCh38
rs901486137 Chr.22: 24225785-24225785 on Build GRCh38
rs374828667 Chr.22: 24225585-24225585 on Build GRCh38
rs772642258 Chr.22: 24225571-24225571 on Build GRCh38
rs921530830 Chr.22: 24225843-24225843 on Build GRCh38
rs1006560724 Chr.22: 24225711-24225711 on Build GRCh38
rs755235008 Chr.22: 24225619-24225619 on Build GRCh38
rs546779082 Chr.22: 24225773-24225773 on Build GRCh38
rs1023091411 Chr.22: 24225576-24225576 on Build GRCh38
rs765031471 Chr.22: 24225582-24225582 on Build GRCh38
rs1014620904 Chr.22: 24225753-24225753 on Build GRCh38
rs367843924 Chr.22: 24225617-24225617 on Build GRCh38
rs374350087 Chr.22: 24225497-24225497 on Build GRCh38
rs73167566 Chr.22: 24225553-24225553 on Build GRCh38
rs769426895 Chr.22: 24225667-24225667 on Build GRCh38
rs781271175 Chr.22: 24225503-24225504 on Build GRCh38
rs778369692 Chr.22: 24225626-24225626 on Build GRCh38
rs12627971 Chr.22: 24225523-24225523 on Build GRCh38
rs1006033470 Chr.22: 24225871-24225871 on Build GRCh38
rs573307691 Chr.22: 24225892-24225892 on Build GRCh38
rs562982518 Chr.22: 24225691-24225691 on Build GRCh38
rs557389866 Chr.22: 24225822-24225822 on Build GRCh38
rs752816688 Chr.22: 24225701-24225701 on Build GRCh38
rs751403095 Chr.22: 24225587-24225587 on Build GRCh38
rs752581530 Chr.22: 24225510-24225510 on Build GRCh38
rs988291433 Chr.22: 24225824-24225824 on Build GRCh38
rs202092652 Chr.22: 24225570-24225570 on Build GRCh38
rs140980900 Chr.22: 24225573-24225573 on Build GRCh38
rs1030702214 Chr.22: 24225521-24225521 on Build GRCh38
rs140834056 Chr.22: 24225586-24225586 on Build GRCh38
rs984702061 Chr.22: 24225555-24225555 on Build GRCh38
rs903488327 Chr.22: 24225504-24225504 on Build GRCh38
rs1048426049 Chr.22: 24225705-24225705 on Build GRCh38
rs1016185568 Chr.22: 24225715-24225715 on Build GRCh38
rs374036306 Chr.22: 24225699-24225699 on Build GRCh38
rs201765477 Chr.22: 24225631-24225631 on Build GRCh38
rs7288670 Chr.22: 24225858-24225858 on Build GRCh38
rs773460385 Chr.22: 24225860-24225860 on Build GRCh38
rs201667511 Chr.22: 24225525-24225525 on Build GRCh38
rs185866792 Chr.22: 24225485-24225485 on Build GRCh38
rs999878962 Chr.22: 24225763-24225763 on Build GRCh38
rs916070531 Chr.22: 24225747-24225747 on Build GRCh38
rs748517292 Chr.22: 24225624-24225624 on Build GRCh38
rs535139256 Chr.22: 24225791-24225791 on Build GRCh38
rs935029942 Chr.22: 24225776-24225776 on Build GRCh38
rs1036858055 Chr.22: 24225876-24225876 on Build GRCh38
rs899898018 Chr.22: 24225538-24225538 on Build GRCh38
rs142733225 Chr.22: 24225625-24225625 on Build GRCh38
rs386820187 Chr.22: 24225857-24225858 on Build GRCh38
rs372565443 Chr.22: 24225676-24225676 on Build GRCh38
rs370949394 Chr.22: 24225509-24225509 on Build GRCh38
rs759245413 Chr.22: 24225641-24225641 on Build GRCh38
rs777819007 Chr.22: 24225541-24225541 on Build GRCh38
rs962061374 Chr.22: 24225718-24225718 on Build GRCh38
rs745862633 Chr.22: 24225504-24225504 on Build GRCh38

View Full Product Details