Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25201295-25201790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBB3
Entrez Gene ID
1417
Gene Name
crystallin beta B3
Gene Aliases
CATCN2, CRYB3, CTRCT22
UniGene
Hs.533022
Gene Chromosome Location
Chr.22: 25199312-25207617 on Build GRCh38
Transcripts
NM_004076.4 XM_011529902.2 XM_017028599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6635860 c.60T>C p.L20L Chr.22: 25201456-25201456 on Build GRCh38
5646819 c.24C>T p.P8P Chr.22: 25201420-25201420 on Build GRCh38
4103122 c.15C>T p.H5H Chr.22: 25201411-25201411 on Build GRCh38
243860 c.5C>T p.A2V Chr.22: 25201401-25201401 on Build GRCh38
1032787 c.25G>A p.E9K Chr.22: 25201421-25201421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913984505 Chr.22: 25201733-25201733 on Build GRCh38
rs989555298 Chr.22: 25201732-25201732 on Build GRCh38
rs759527194 Chr.22: 25201388-25201388 on Build GRCh38
rs144393956 Chr.22: 25201402-25201402 on Build GRCh38
rs948023294 Chr.22: 25201749-25201749 on Build GRCh38
rs758170568 Chr.22: 25201523-25201523 on Build GRCh38
rs186827436 Chr.22: 25201712-25201712 on Build GRCh38
rs755555776 Chr.22: 25201405-25201405 on Build GRCh38
rs748121476 Chr.22: 25201510-25201510 on Build GRCh38
rs150327844 Chr.22: 25201549-25201549 on Build GRCh38
rs374542918 Chr.22: 25201379-25201379 on Build GRCh38
rs561701759 Chr.22: 25201594-25201594 on Build GRCh38
rs763541551 Chr.22: 25201397-25201397 on Build GRCh38
rs2269672 Chr.22: 25201364-25201364 on Build GRCh38
rs532959032 Chr.22: 25201339-25201339 on Build GRCh38
rs200388017 Chr.22: 25201742-25201743 on Build GRCh38
rs367616021 Chr.22: 25201421-25201421 on Build GRCh38
rs139163387 Chr.22: 25201404-25201404 on Build GRCh38
rs746036578 Chr.22: 25201472-25201472 on Build GRCh38
rs755696732 Chr.22: 25201399-25201400 on Build GRCh38
rs764517889 Chr.22: 25201400-25201400 on Build GRCh38
rs3064261 Chr.22: 25201744-25201745 on Build GRCh38
rs914432518 Chr.22: 25201547-25201547 on Build GRCh38
rs1021263423 Chr.22: 25201625-25201625 on Build GRCh38
rs149884520 Chr.22: 25201411-25201411 on Build GRCh38
rs980800215 Chr.22: 25201443-25201443 on Build GRCh38
rs779636937 Chr.22: 25201410-25201410 on Build GRCh38
rs765740553 Chr.22: 25201478-25201478 on Build GRCh38
rs753125280 Chr.22: 25201396-25201396 on Build GRCh38
rs749294708 Chr.22: 25201376-25201376 on Build GRCh38
rs544783292 Chr.22: 25201670-25201670 on Build GRCh38
rs780980241 Chr.22: 25201403-25201403 on Build GRCh38
rs144871285 Chr.22: 25201621-25201621 on Build GRCh38
rs751211649 Chr.22: 25201351-25201351 on Build GRCh38
rs147831812 Chr.22: 25201434-25201434 on Build GRCh38
rs755647925 Chr.22: 25201461-25201461 on Build GRCh38
rs780783860 Chr.22: 25201363-25201363 on Build GRCh38
rs187054110 Chr.22: 25201500-25201500 on Build GRCh38
rs754537825 Chr.22: 25201497-25201497 on Build GRCh38
rs761209154 Chr.22: 25201630-25201630 on Build GRCh38
rs759853170 Chr.22: 25201428-25201428 on Build GRCh38
rs749008681 Chr.22: 25201338-25201338 on Build GRCh38
rs549370220 Chr.22: 25201723-25201723 on Build GRCh38
rs35317614 Chr.22: 25201745-25201746 on Build GRCh38
rs2027889 Chr.22: 25201659-25201659 on Build GRCh38
rs989503152 Chr.22: 25201507-25201507 on Build GRCh38
rs561169675 Chr.22: 25201725-25201725 on Build GRCh38
rs995226639 Chr.22: 25201398-25201398 on Build GRCh38
rs935491684 Chr.22: 25201568-25201568 on Build GRCh38
rs544227754 Chr.22: 25201631-25201631 on Build GRCh38
rs550527178 Chr.22: 25201756-25201756 on Build GRCh38
rs983420425 Chr.22: 25201437-25201437 on Build GRCh38
rs775674447 Chr.22: 25201448-25201448 on Build GRCh38
rs572823233 Chr.22: 25201597-25201597 on Build GRCh38
rs764711964 Chr.22: 25201454-25201454 on Build GRCh38
rs1033661697 Chr.22: 25201724-25201724 on Build GRCh38
rs760815425 Chr.22: 25201382-25201382 on Build GRCh38
rs560237354 Chr.22: 25201685-25201685 on Build GRCh38
rs757004750 Chr.22: 25201362-25201362 on Build GRCh38
rs183993135 Chr.22: 25201417-25201417 on Build GRCh38
rs768642748 Chr.22: 25201412-25201412 on Build GRCh38
rs562297552 Chr.22: 25201331-25201331 on Build GRCh38
rs1012305716 Chr.22: 25201340-25201340 on Build GRCh38
rs531720937 Chr.22: 25201744-25201744 on Build GRCh38
rs751977829 Chr.22: 25201401-25201401 on Build GRCh38
rs1046900436 Chr.22: 25201544-25201544 on Build GRCh38
rs192385580 Chr.22: 25201603-25201603 on Build GRCh38
rs199594395 Chr.22: 25201361-25201361 on Build GRCh38
rs539217392 Chr.22: 25201581-25201581 on Build GRCh38
rs900157110 Chr.22: 25201643-25201643 on Build GRCh38
rs141199896 Chr.22: 25201489-25201489 on Build GRCh38
rs928040668 Chr.22: 25201764-25201764 on Build GRCh38
rs776894149 Chr.22: 25201384-25201384 on Build GRCh38
rs1004383237 Chr.22: 25201702-25201702 on Build GRCh38
rs182106380 Chr.22: 25201647-25201647 on Build GRCh38
rs887309168 Chr.22: 25201682-25201682 on Build GRCh38
rs540238430 Chr.22: 25201602-25201602 on Build GRCh38
rs749992336 Chr.22: 25201470-25201470 on Build GRCh38
rs1025526291 Chr.22: 25201703-25201703 on Build GRCh38
rs148607859 Chr.22: 25201468-25201468 on Build GRCh38
rs762265757 Chr.22: 25201467-25201467 on Build GRCh38
rs779794577 Chr.22: 25201483-25201484 on Build GRCh38
rs747920787 Chr.22: 25201419-25201419 on Build GRCh38
rs763306632 Chr.22: 25201453-25201453 on Build GRCh38
rs1014733207 Chr.22: 25201728-25201728 on Build GRCh38
rs748483757 Chr.22: 25201420-25201420 on Build GRCh38
rs72559634 Chr.22: 25201743-25201744 on Build GRCh38
rs376429738 Chr.22: 25201350-25201350 on Build GRCh38
rs779852655 Chr.22: 25201372-25201372 on Build GRCh38
rs113363513 Chr.22: 25201358-25201358 on Build GRCh38
rs771152543 Chr.22: 25201383-25201383 on Build GRCh38

View Full Product Details