Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92478347-92478840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1
Entrez Gene ID
2672
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
GFI-1, GFI1A, SCN2, ZNF163
UniGene
Hs.73172
Gene Chromosome Location
Chr.1: 92473043-92486876 on Build GRCh38
Transcripts
NM_001127215.1 NM_001127216.1 NM_005263.3 XM_005270749.3 XM_011541245.2 XM_011541246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408120 c.941G>C p.C314S Chr.1: 92478737-92478737 on Build GRCh38
6867249 c.1006G>A p.D336N Chr.1: 92478672-92478672 on Build GRCh38
5847491 c.961T>C p.F321L Chr.1: 92478717-92478717 on Build GRCh38
7096435 c.1080C>G p.F360L Chr.1: 92478598-92478598 on Build GRCh38
4392428 c.967A>C p.R323R Chr.1: 92478711-92478711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020047718 Chr.1: 92478616-92478616 on Build GRCh38
rs775847745 Chr.1: 92478736-92478736 on Build GRCh38
rs901002678 Chr.1: 92478491-92478491 on Build GRCh38
rs376338971 Chr.1: 92478758-92478767 on Build GRCh38
rs769183564 Chr.1: 92478748-92478748 on Build GRCh38
rs781299744 Chr.1: 92478687-92478687 on Build GRCh38
rs35896485 Chr.1: 92478794-92478795 on Build GRCh38
rs753715649 Chr.1: 92478797-92478797 on Build GRCh38
rs754658728 Chr.1: 92478795-92478795 on Build GRCh38
rs184448271 Chr.1: 92478394-92478394 on Build GRCh38
rs1001142272 Chr.1: 92478425-92478425 on Build GRCh38
rs770961178 Chr.1: 92478674-92478674 on Build GRCh38
rs545182784 Chr.1: 92478501-92478501 on Build GRCh38
rs779414850 Chr.1: 92478790-92478791 on Build GRCh38
rs376042061 Chr.1: 92478740-92478740 on Build GRCh38
rs764527821 Chr.1: 92478566-92478566 on Build GRCh38
rs931023354 Chr.1: 92478381-92478381 on Build GRCh38
rs759781375 Chr.1: 92478546-92478546 on Build GRCh38
rs568345533 Chr.1: 92478419-92478419 on Build GRCh38
rs766059250 Chr.1: 92478796-92478796 on Build GRCh38
rs775825511 Chr.1: 92478559-92478559 on Build GRCh38
rs1014266969 Chr.1: 92478464-92478464 on Build GRCh38
rs370438369 Chr.1: 92478750-92478750 on Build GRCh38
rs559993011 Chr.1: 92478769-92478769 on Build GRCh38
rs747988798 Chr.1: 92478797-92478797 on Build GRCh38
rs907917339 Chr.1: 92478462-92478463 on Build GRCh38
rs746141709 Chr.1: 92478712-92478712 on Build GRCh38
rs545888848 Chr.1: 92478438-92478438 on Build GRCh38
rs546029934 Chr.1: 92478769-92478787 on Build GRCh38
rs117252159 Chr.1: 92478422-92478422 on Build GRCh38
rs757990501 Chr.1: 92478787-92478788 on Build GRCh38
rs774792393 Chr.1: 92478749-92478749 on Build GRCh38
rs973065856 Chr.1: 92478632-92478632 on Build GRCh38
rs373356460 Chr.1: 92478598-92478598 on Build GRCh38
rs758599873 Chr.1: 92478625-92478625 on Build GRCh38
rs764707446 Chr.1: 92478767-92478768 on Build GRCh38
rs886046555 Chr.1: 92478758-92478761 on Build GRCh38
rs754264573 Chr.1: 92478775-92478776 on Build GRCh38
rs966866159 Chr.1: 92478531-92478531 on Build GRCh38
rs1040474905 Chr.1: 92478471-92478471 on Build GRCh38
rs371078453 Chr.1: 92478757-92478758 on Build GRCh38
rs747130172 Chr.1: 92478666-92478666 on Build GRCh38
rs764728437 Chr.1: 92478763-92478764 on Build GRCh38
rs766980817 Chr.1: 92478765-92478765 on Build GRCh38
rs952336709 Chr.1: 92478807-92478807 on Build GRCh38
rs755765258 Chr.1: 92478604-92478604 on Build GRCh38
rs767221774 Chr.1: 92478595-92478595 on Build GRCh38
rs755613674 Chr.1: 92478419-92478420 on Build GRCh38
rs139144506 Chr.1: 92478646-92478646 on Build GRCh38
rs759849584 Chr.1: 92478761-92478762 on Build GRCh38
rs746453276 Chr.1: 92478791-92478792 on Build GRCh38
rs760305848 Chr.1: 92478795-92478795 on Build GRCh38
rs1001068366 Chr.1: 92478538-92478538 on Build GRCh38
rs1021087871 Chr.1: 92478517-92478517 on Build GRCh38
rs200902533 Chr.1: 92478758-92478763 on Build GRCh38
rs189638200 Chr.1: 92478574-92478574 on Build GRCh38
rs768494519 Chr.1: 92478758-92478759 on Build GRCh38
rs372208699 Chr.1: 92478561-92478561 on Build GRCh38
rs927026116 Chr.1: 92478786-92478786 on Build GRCh38
rs575393960 Chr.1: 92478423-92478423 on Build GRCh38
rs372553729 Chr.1: 92478730-92478730 on Build GRCh38
rs1048127558 Chr.1: 92478406-92478406 on Build GRCh38
rs1032416552 Chr.1: 92478539-92478539 on Build GRCh38
rs112121970 Chr.1: 92478530-92478532 on Build GRCh38
rs80337126 Chr.1: 92478631-92478631 on Build GRCh38
rs918426594 Chr.1: 92478376-92478376 on Build GRCh38
rs765560374 Chr.1: 92478552-92478552 on Build GRCh38
rs762267083 Chr.1: 92478444-92478444 on Build GRCh38
rs144192494 Chr.1: 92478800-92478800 on Build GRCh38
rs751075166 Chr.1: 92478571-92478571 on Build GRCh38
rs766365921 Chr.1: 92478759-92478760 on Build GRCh38
rs932513740 Chr.1: 92478493-92478493 on Build GRCh38
rs779282255 Chr.1: 92478649-92478649 on Build GRCh38

View Full Product Details