Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29272128-29272636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EWSR1
Entrez Gene ID
2130
Gene Name
EWS RNA binding protein 1
Gene Aliases
EWS, EWS-FLI1, bK984G1.4
UniGene
Hs.374477
Gene Chromosome Location
Chr.22: 29268009-29300525 on Build GRCh38
Transcripts
NM_001163285.1 NM_001163286.1 NM_001163287.1 NM_005243.3 NM_013986.3 XM_005261389.3 XM_005261390.4 XM_011529995.2 XM_011529996.2 XM_011529997.1 XM_011529998.1 XM_011529999.2 XM_011530000.1 XM_011530001.2 XM_011530002.2 XM_017028644.1 XM_017028645.1 XM_017028646.1 XM_017028647.1 XM_017028648.1 XM_017028649.1 XM_017028650.1 XM_017028651.1 XM_017028652.1 XM_017028653.1 XM_017028654.1 XM_017028655.1 XM_017028656.1 XM_017028657.1 XM_017028658.1 XM_017028659.1 XM_017028660.1 XM_017028661.1 XM_017028662.1 XM_017028663.1 XM_017028664.1 XM_017028665.1 XM_017028666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1153984 c.66C>T p.T22T Chr.22: 29272395-29272395 on Build GRCh38
478851 c.53A>G p.Y18C Chr.22: 29272382-29272382 on Build GRCh38
4582286 c.65C>T p.T22I Chr.22: 29272394-29272394 on Build GRCh38
4103282 c.67G>A p.A23T Chr.22: 29272396-29272396 on Build GRCh38
1136842 c.53A>G p.Y18C Chr.22: 29272382-29272382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558819034 Chr.22: 29272175-29272175 on Build GRCh38
rs781641485 Chr.22: 29272372-29272372 on Build GRCh38
rs189461448 Chr.22: 29272539-29272539 on Build GRCh38
rs560101493 Chr.22: 29272466-29272466 on Build GRCh38
rs923040938 Chr.22: 29272568-29272568 on Build GRCh38
rs766632221 Chr.22: 29272338-29272338 on Build GRCh38
rs368813757 Chr.22: 29272203-29272203 on Build GRCh38
rs556889251 Chr.22: 29272305-29272305 on Build GRCh38
rs766835367 Chr.22: 29272464-29272464 on Build GRCh38
rs753568405 Chr.22: 29272342-29272342 on Build GRCh38
rs752720273 Chr.22: 29272167-29272167 on Build GRCh38
rs776845481 Chr.22: 29272264-29272264 on Build GRCh38
rs377398751 Chr.22: 29272332-29272332 on Build GRCh38
rs773819889 Chr.22: 29272461-29272461 on Build GRCh38
rs947629103 Chr.22: 29272583-29272583 on Build GRCh38
rs763137483 Chr.22: 29272250-29272250 on Build GRCh38
rs887617813 Chr.22: 29272589-29272589 on Build GRCh38
rs964770249 Chr.22: 29272157-29272157 on Build GRCh38
rs763801144 Chr.22: 29272206-29272206 on Build GRCh38
rs748165509 Chr.22: 29272215-29272215 on Build GRCh38
rs575156065 Chr.22: 29272472-29272472 on Build GRCh38
rs140505486 Chr.22: 29272229-29272229 on Build GRCh38
rs764603459 Chr.22: 29272180-29272180 on Build GRCh38
rs778030232 Chr.22: 29272536-29272536 on Build GRCh38
rs770806014 Chr.22: 29272398-29272398 on Build GRCh38
rs369093316 Chr.22: 29272317-29272317 on Build GRCh38
rs547085912 Chr.22: 29272458-29272458 on Build GRCh38
rs541858841 Chr.22: 29272502-29272502 on Build GRCh38
rs931707838 Chr.22: 29272575-29272575 on Build GRCh38
rs774427133 Chr.22: 29272440-29272440 on Build GRCh38
rs776256103 Chr.22: 29272405-29272405 on Build GRCh38
rs764920378 Chr.22: 29272470-29272470 on Build GRCh38
rs374252787 Chr.22: 29272453-29272453 on Build GRCh38
rs200665732 Chr.22: 29272275-29272275 on Build GRCh38
rs768331633 Chr.22: 29272209-29272209 on Build GRCh38
rs761540841 Chr.22: 29272330-29272330 on Build GRCh38
rs567998920 Chr.22: 29272295-29272295 on Build GRCh38
rs757936434 Chr.22: 29272477-29272477 on Build GRCh38
rs762363230 Chr.22: 29272284-29272284 on Build GRCh38
rs147500022 Chr.22: 29272581-29272581 on Build GRCh38
rs6006084 Chr.22: 29272210-29272210 on Build GRCh38
rs530800854 Chr.22: 29272577-29272577 on Build GRCh38
rs752116070 Chr.22: 29272355-29272357 on Build GRCh38
rs371367792 Chr.22: 29272204-29272204 on Build GRCh38
rs750786554 Chr.22: 29272194-29272194 on Build GRCh38
rs377732811 Chr.22: 29272395-29272395 on Build GRCh38
rs1045462749 Chr.22: 29272276-29272276 on Build GRCh38
rs969825379 Chr.22: 29272561-29272561 on Build GRCh38
rs757729009 Chr.22: 29272185-29272185 on Build GRCh38
rs201537508 Chr.22: 29272274-29272274 on Build GRCh38
rs757511243 Chr.22: 29272359-29272359 on Build GRCh38
rs973933631 Chr.22: 29272527-29272527 on Build GRCh38
rs542135443 Chr.22: 29272465-29272465 on Build GRCh38
rs752106445 Chr.22: 29272182-29272182 on Build GRCh38
rs768830118 Chr.22: 29272228-29272228 on Build GRCh38
rs777722463 Chr.22: 29272396-29272396 on Build GRCh38
rs1030758644 Chr.22: 29272181-29272181 on Build GRCh38
rs934984595 Chr.22: 29272534-29272534 on Build GRCh38
rs34328838 Chr.22: 29272337-29272338 on Build GRCh38
rs571961306 Chr.22: 29272444-29272451 on Build GRCh38
rs950721618 Chr.22: 29272197-29272197 on Build GRCh38
rs747122752 Chr.22: 29272566-29272566 on Build GRCh38
rs199543598 Chr.22: 29272290-29272290 on Build GRCh38
rs535098541 Chr.22: 29272301-29272301 on Build GRCh38
rs979668761 Chr.22: 29272496-29272496 on Build GRCh38
rs201792944 Chr.22: 29272226-29272226 on Build GRCh38
rs1039276995 Chr.22: 29272594-29272594 on Build GRCh38
rs750321493 Chr.22: 29272292-29272292 on Build GRCh38
rs781602544 Chr.22: 29272190-29272190 on Build GRCh38
rs138408577 Chr.22: 29272599-29272599 on Build GRCh38
rs775729620 Chr.22: 29272244-29272244 on Build GRCh38
rs200382526 Chr.22: 29272200-29272200 on Build GRCh38
rs769936524 Chr.22: 29272236-29272236 on Build GRCh38
rs539480242 Chr.22: 29272322-29272322 on Build GRCh38
rs756465092 Chr.22: 29272471-29272472 on Build GRCh38
rs765446727 Chr.22: 29272165-29272165 on Build GRCh38
rs772125233 Chr.22: 29272224-29272224 on Build GRCh38
rs778627427 Chr.22: 29272361-29272361 on Build GRCh38
rs746797640 Chr.22: 29272397-29272397 on Build GRCh38
rs981606565 Chr.22: 29272567-29272567 on Build GRCh38
rs768850800 Chr.22: 29272435-29272435 on Build GRCh38
rs934360766 Chr.22: 29272580-29272580 on Build GRCh38
rs763045571 Chr.22: 29272173-29272173 on Build GRCh38
rs900672731 Chr.22: 29272604-29272608 on Build GRCh38
rs1035042314 Chr.22: 29272382-29272382 on Build GRCh38
rs997544985 Chr.22: 29272179-29272179 on Build GRCh38
rs761057257 Chr.22: 29272463-29272463 on Build GRCh38
rs369885313 Chr.22: 29272343-29272343 on Build GRCh38
rs777129655 Chr.22: 29272480-29272480 on Build GRCh38
rs374284762 Chr.22: 29272363-29272363 on Build GRCh38
rs1006556071 Chr.22: 29272608-29272608 on Build GRCh38
rs201834106 Chr.22: 29272444-29272444 on Build GRCh38
rs867467747 Chr.22: 29272506-29272506 on Build GRCh38

View Full Product Details