Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30285710-30286199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
UniGene
Hs.444950
Gene Chromosome Location
Chr.22: 30285118-30289627 on Build GRCh38
Transcripts
NM_001037666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425750 c.751A>T p.S251C Chr.22: 30286091-30286091 on Build GRCh38
4829834 c.800G>A p.R267H Chr.22: 30286042-30286042 on Build GRCh38
3553261 c.909C>T p.F303F Chr.22: 30285844-30285844 on Build GRCh38
4801333 c.777G>T p.S259S Chr.22: 30286065-30286065 on Build GRCh38
4636051 c.903C>G p.F301L Chr.22: 30285850-30285850 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776262884 Chr.22: 30285989-30285989 on Build GRCh38
rs1000404870 Chr.22: 30286064-30286064 on Build GRCh38
rs772758847 Chr.22: 30285977-30285977 on Build GRCh38
rs891910469 Chr.22: 30286101-30286101 on Build GRCh38
rs769129971 Chr.22: 30285837-30285838 on Build GRCh38
rs1021389156 Chr.22: 30286027-30286027 on Build GRCh38
rs751205602 Chr.22: 30285822-30285823 on Build GRCh38
rs765264145 Chr.22: 30285993-30285993 on Build GRCh38
rs6147585 Chr.22: 30285808-30285809 on Build GRCh38
rs1021423520 Chr.22: 30285744-30285744 on Build GRCh38
rs772350946 Chr.22: 30286106-30286106 on Build GRCh38
rs906024231 Chr.22: 30286038-30286038 on Build GRCh38
rs377703825 Chr.22: 30285935-30285935 on Build GRCh38
rs55833321 Chr.22: 30285829-30285830 on Build GRCh38
rs760831027 Chr.22: 30285804-30285805 on Build GRCh38
rs756171722 Chr.22: 30285933-30285933 on Build GRCh38
rs572104208 Chr.22: 30286036-30286036 on Build GRCh38
rs868093308 Chr.22: 30285807-30285808 on Build GRCh38
rs940163335 Chr.22: 30285960-30285960 on Build GRCh38
rs376300447 Chr.22: 30285983-30285983 on Build GRCh38
rs753767117 Chr.22: 30285804-30285804 on Build GRCh38
rs756306909 Chr.22: 30286043-30286043 on Build GRCh38
rs748712648 Chr.22: 30285766-30285766 on Build GRCh38
rs769048839 Chr.22: 30285857-30285857 on Build GRCh38
rs369445412 Chr.22: 30285796-30285796 on Build GRCh38
rs748636183 Chr.22: 30286072-30286072 on Build GRCh38
rs765176150 Chr.22: 30285917-30285917 on Build GRCh38
rs954554338 Chr.22: 30285839-30285839 on Build GRCh38
rs370019964 Chr.22: 30285938-30285938 on Build GRCh38
rs200317782 Chr.22: 30285988-30285988 on Build GRCh38
rs776349713 Chr.22: 30286141-30286141 on Build GRCh38
rs757402615 Chr.22: 30285836-30285836 on Build GRCh38
rs756796691 Chr.22: 30285826-30285827 on Build GRCh38
rs754938761 Chr.22: 30285896-30285896 on Build GRCh38
rs765442258 Chr.22: 30285989-30285991 on Build GRCh38
rs188064023 Chr.22: 30286017-30286017 on Build GRCh38
rs891849043 Chr.22: 30286005-30286005 on Build GRCh38
rs748558260 Chr.22: 30286117-30286123 on Build GRCh38
rs35151307 Chr.22: 30285809-30285810 on Build GRCh38
rs376001918 Chr.22: 30286001-30286001 on Build GRCh38
rs780915705 Chr.22: 30285828-30285829 on Build GRCh38
rs374186822 Chr.22: 30285883-30285883 on Build GRCh38
rs370649027 Chr.22: 30286016-30286016 on Build GRCh38
rs758164626 Chr.22: 30285819-30285820 on Build GRCh38
rs371123714 Chr.22: 30286142-30286142 on Build GRCh38
rs769966102 Chr.22: 30286085-30286085 on Build GRCh38
rs775313464 Chr.22: 30286149-30286149 on Build GRCh38
rs757569133 Chr.22: 30285930-30285930 on Build GRCh38
rs778164967 Chr.22: 30286108-30286108 on Build GRCh38
rs202028147 Chr.22: 30286000-30286000 on Build GRCh38
rs115548734 Chr.22: 30285830-30285830 on Build GRCh38
rs753853345 Chr.22: 30285894-30285894 on Build GRCh38
rs779487292 Chr.22: 30286015-30286015 on Build GRCh38
rs749861752 Chr.22: 30285834-30285835 on Build GRCh38
rs955294917 Chr.22: 30285895-30285895 on Build GRCh38
rs765935276 Chr.22: 30285889-30285889 on Build GRCh38
rs56979117 Chr.22: 30285808-30285808 on Build GRCh38
rs750642300 Chr.22: 30286037-30286037 on Build GRCh38
rs745481482 Chr.22: 30286146-30286146 on Build GRCh38
rs747278480 Chr.22: 30285835-30285835 on Build GRCh38
rs867318734 Chr.22: 30285833-30285834 on Build GRCh38
rs140150664 Chr.22: 30285806-30285807 on Build GRCh38
rs761583978 Chr.22: 30285871-30285871 on Build GRCh38
rs769373208 Chr.22: 30286145-30286145 on Build GRCh38
rs565934560 Chr.22: 30286172-30286172 on Build GRCh38
rs772421093 Chr.22: 30285973-30285973 on Build GRCh38
rs201713389 Chr.22: 30286128-30286128 on Build GRCh38
rs757768327 Chr.22: 30285925-30285925 on Build GRCh38
rs778807317 Chr.22: 30285834-30285834 on Build GRCh38
rs753908989 Chr.22: 30286065-30286065 on Build GRCh38
rs769138635 Chr.22: 30285961-30285961 on Build GRCh38
rs201496296 Chr.22: 30285833-30285833 on Build GRCh38
rs780677610 Chr.22: 30285830-30285831 on Build GRCh38
rs781277636 Chr.22: 30285931-30285931 on Build GRCh38
rs941241835 Chr.22: 30285981-30285981 on Build GRCh38
rs373022461 Chr.22: 30285965-30285965 on Build GRCh38
rs760113556 Chr.22: 30285886-30285886 on Build GRCh38
rs956018813 Chr.22: 30285757-30285757 on Build GRCh38
rs763930328 Chr.22: 30285821-30285822 on Build GRCh38
rs759609513 Chr.22: 30285990-30285990 on Build GRCh38
rs752424095 Chr.22: 30285922-30285922 on Build GRCh38
rs745933272 Chr.22: 30285838-30285838 on Build GRCh38
rs774763697 Chr.22: 30285964-30285964 on Build GRCh38
rs370723725 Chr.22: 30285984-30285984 on Build GRCh38
rs749746879 Chr.22: 30285856-30285856 on Build GRCh38
rs767070750 Chr.22: 30285882-30285882 on Build GRCh38
rs547927578 Chr.22: 30286098-30286098 on Build GRCh38
rs796431006 Chr.22: 30285805-30285805 on Build GRCh38
rs770698252 Chr.22: 30286133-30286133 on Build GRCh38
rs1010060439 Chr.22: 30285729-30285729 on Build GRCh38
rs774675821 Chr.22: 30285861-30285861 on Build GRCh38
rs1029346314 Chr.22: 30286153-30286153 on Build GRCh38
rs754982494 Chr.22: 30286066-30286066 on Build GRCh38
rs769847466 Chr.22: 30285841-30285841 on Build GRCh38
rs775990870 Chr.22: 30285854-30285854 on Build GRCh38
rs780165118 Chr.22: 30286046-30286046 on Build GRCh38
rs968568439 Chr.22: 30285752-30285752 on Build GRCh38
rs1029674366 Chr.22: 30286152-30286152 on Build GRCh38
rs746045837 Chr.22: 30285932-30285932 on Build GRCh38
rs796556705 Chr.22: 30285805-30285808 on Build GRCh38
rs756140777 Chr.22: 30285734-30285734 on Build GRCh38
rs1050352549 Chr.22: 30286171-30286200 on Build GRCh38
rs200533086 Chr.22: 30285808-30285808 on Build GRCh38

View Full Product Details