Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38538203-38538703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMC1
Entrez Gene ID
11144
Gene Name
DNA meiotic recombinase 1
Gene Aliases
DMC1H, LIM15, dJ199H16.1
UniGene
Hs.339396
Gene Chromosome Location
Chr.22: 38509154-38570286 on Build GRCh38
Transcripts
NM_001278208.1 NM_007068.3 XM_006724112.2 XM_011529834.1 XM_011529835.2 XM_011529837.1 XM_011529838.1 XM_017028542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726312 c.600G>T p.M200I Chr.22: 38538599-38538599 on Build GRCh38
4977707 c.666C>T p.I222I Chr.22: 38538404-38538404 on Build GRCh38
7140302 c.667G>T p.D223Y Chr.22: 38538403-38538403 on Build GRCh38
1714393 c.631C>T p.H211Y Chr.22: 38538568-38538568 on Build GRCh38
4677999 c.596A>G p.Q199R Chr.22: 38538603-38538603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762406860 Chr.22: 38538303-38538304 on Build GRCh38
rs773347078 Chr.22: 38538418-38538418 on Build GRCh38
rs767965966 Chr.22: 38538372-38538372 on Build GRCh38
rs957140253 Chr.22: 38538566-38538566 on Build GRCh38
rs1047145080 Chr.22: 38538439-38538439 on Build GRCh38
rs11570425 Chr.22: 38538456-38538456 on Build GRCh38
rs2227914 Chr.22: 38538601-38538601 on Build GRCh38
rs941482256 Chr.22: 38538452-38538452 on Build GRCh38
rs766473627 Chr.22: 38538493-38538495 on Build GRCh38
rs760981205 Chr.22: 38538417-38538417 on Build GRCh38
rs761687068 Chr.22: 38538250-38538250 on Build GRCh38
rs138352828 Chr.22: 38538350-38538350 on Build GRCh38
rs762330092 Chr.22: 38538662-38538662 on Build GRCh38
rs201348870 Chr.22: 38538404-38538404 on Build GRCh38
rs767781951 Chr.22: 38538274-38538274 on Build GRCh38
rs749245221 Chr.22: 38538567-38538567 on Build GRCh38
rs4987166 Chr.22: 38538472-38538472 on Build GRCh38
rs774652856 Chr.22: 38538440-38538440 on Build GRCh38
rs1000731986 Chr.22: 38538362-38538362 on Build GRCh38
rs199821104 Chr.22: 38538391-38538391 on Build GRCh38
rs756601168 Chr.22: 38538308-38538308 on Build GRCh38
rs763162657 Chr.22: 38538255-38538255 on Build GRCh38
rs557897016 Chr.22: 38538253-38538253 on Build GRCh38
rs760936356 Chr.22: 38538612-38538612 on Build GRCh38
rs773429160 Chr.22: 38538416-38538416 on Build GRCh38
rs4987165 Chr.22: 38538494-38538494 on Build GRCh38
rs199712228 Chr.22: 38538329-38538329 on Build GRCh38
rs1053316108 Chr.22: 38538656-38538656 on Build GRCh38
rs779570436 Chr.22: 38538520-38538520 on Build GRCh38
rs998431755 Chr.22: 38538477-38538477 on Build GRCh38
rs201179906 Chr.22: 38538288-38538288 on Build GRCh38
rs368393201 Chr.22: 38538248-38538248 on Build GRCh38
rs200187131 Chr.22: 38538294-38538294 on Build GRCh38
rs780934367 Chr.22: 38538541-38538541 on Build GRCh38
rs374584727 Chr.22: 38538589-38538589 on Build GRCh38
rs752982276 Chr.22: 38538461-38538461 on Build GRCh38
rs929137334 Chr.22: 38538438-38538438 on Build GRCh38
rs754302372 Chr.22: 38538629-38538629 on Build GRCh38
rs189723693 Chr.22: 38538638-38538638 on Build GRCh38
rs772154761 Chr.22: 38538599-38538599 on Build GRCh38
rs764445641 Chr.22: 38538625-38538625 on Build GRCh38
rs1001407956 Chr.22: 38538426-38538426 on Build GRCh38
rs751890214 Chr.22: 38538267-38538267 on Build GRCh38
rs549690822 Chr.22: 38538531-38538531 on Build GRCh38
rs11570424 Chr.22: 38538482-38538482 on Build GRCh38
rs774320642 Chr.22: 38538577-38538577 on Build GRCh38
rs766701089 Chr.22: 38538258-38538258 on Build GRCh38
rs777796944 Chr.22: 38538553-38538553 on Build GRCh38
rs4987164 Chr.22: 38538510-38538510 on Build GRCh38
rs369538413 Chr.22: 38538546-38538546 on Build GRCh38
rs765308228 Chr.22: 38538251-38538251 on Build GRCh38
rs867267644 Chr.22: 38538355-38538355 on Build GRCh38
rs974898588 Chr.22: 38538602-38538602 on Build GRCh38
rs757835564 Chr.22: 38538517-38538517 on Build GRCh38
rs771098959 Chr.22: 38538576-38538576 on Build GRCh38
rs755315236 Chr.22: 38538271-38538271 on Build GRCh38
rs746644230 Chr.22: 38538379-38538379 on Build GRCh38
rs757972386 Chr.22: 38538349-38538349 on Build GRCh38
rs769615441 Chr.22: 38538594-38538594 on Build GRCh38
rs759710579 Chr.22: 38538596-38538596 on Build GRCh38
rs143874492 Chr.22: 38538397-38538397 on Build GRCh38
rs376521904 Chr.22: 38538545-38538545 on Build GRCh38
rs748020628 Chr.22: 38538403-38538403 on Build GRCh38
rs763113866 Chr.22: 38538424-38538424 on Build GRCh38
rs926122282 Chr.22: 38538620-38538620 on Build GRCh38
rs778302919 Chr.22: 38538315-38538315 on Build GRCh38
rs766582513 Chr.22: 38538425-38538425 on Build GRCh38
rs374790630 Chr.22: 38538450-38538450 on Build GRCh38
rs764582783 Chr.22: 38538498-38538498 on Build GRCh38
rs981328386 Chr.22: 38538669-38538671 on Build GRCh38
rs775196006 Chr.22: 38538677-38538677 on Build GRCh38
rs754560985 Chr.22: 38538534-38538534 on Build GRCh38

View Full Product Details