Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41694613-41695128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf46
Entrez Gene ID
79640
Gene Name
chromosome 22 open reading frame 46
Gene Aliases
-
UniGene
Hs.517653
Gene Chromosome Location
Chr.22: 41688939-41698136 on Build GRCh38
Transcripts
NM_001142964.1 XM_011530373.2 XM_017028939.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3766715 c.653C>T p.T218I Chr.22: 41694839-41694839 on Build GRCh38
3766714 c.653C>T p.T218I Chr.22: 41694839-41694839 on Build GRCh38
3785562 c.776T>C p.L259P Chr.22: 41694962-41694962 on Build GRCh38
3964355 c.722A>C p.E241A Chr.22: 41694908-41694908 on Build GRCh38
445060 c.837_838insAG p.G282fs*>19 Chr.22: 41695023-41695024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920136296 Chr.22: 41695012-41695012 on Build GRCh38
rs899707233 Chr.22: 41694738-41694738 on Build GRCh38
rs942913908 Chr.22: 41694781-41694781 on Build GRCh38
rs67262039 Chr.22: 41695024-41695025 on Build GRCh38
rs377699274 Chr.22: 41694650-41694650 on Build GRCh38
rs531405111 Chr.22: 41694736-41694736 on Build GRCh38
rs764346480 Chr.22: 41695067-41695067 on Build GRCh38
rs879188428 Chr.22: 41694950-41694950 on Build GRCh38
rs552569500 Chr.22: 41695025-41695025 on Build GRCh38
rs149658084 Chr.22: 41694946-41694946 on Build GRCh38
rs867720582 Chr.22: 41695094-41695094 on Build GRCh38
rs35942153 Chr.22: 41694679-41694680 on Build GRCh38
rs11913145 Chr.22: 41695068-41695068 on Build GRCh38
rs549620761 Chr.22: 41694765-41694765 on Build GRCh38
rs370311364 Chr.22: 41694693-41694693 on Build GRCh38
rs922813038 Chr.22: 41694839-41694839 on Build GRCh38
rs73161347 Chr.22: 41695059-41695059 on Build GRCh38
rs183606851 Chr.22: 41694911-41694911 on Build GRCh38
rs35117663 Chr.22: 41695023-41695024 on Build GRCh38
rs372424033 Chr.22: 41695027-41695028 on Build GRCh38
rs1054494092 Chr.22: 41695017-41695017 on Build GRCh38
rs763690927 Chr.22: 41694776-41694776 on Build GRCh38
rs548598486 Chr.22: 41694990-41694990 on Build GRCh38
rs967426368 Chr.22: 41694921-41694921 on Build GRCh38
rs397764640 Chr.22: 41695028-41695029 on Build GRCh38
rs530229784 Chr.22: 41694849-41694849 on Build GRCh38
rs369922685 Chr.22: 41695023-41695023 on Build GRCh38
rs368948094 Chr.22: 41694741-41694741 on Build GRCh38
rs928769197 Chr.22: 41694968-41694968 on Build GRCh38
rs563764552 Chr.22: 41694651-41694651 on Build GRCh38
rs931583836 Chr.22: 41695034-41695034 on Build GRCh38
rs753380230 Chr.22: 41694879-41694879 on Build GRCh38
rs1046733577 Chr.22: 41694684-41694684 on Build GRCh38
rs79249497 Chr.22: 41695009-41695009 on Build GRCh38
rs758892288 Chr.22: 41694811-41694811 on Build GRCh38
rs957387102 Chr.22: 41694782-41694782 on Build GRCh38
rs563052129 Chr.22: 41694787-41694787 on Build GRCh38
rs886818367 Chr.22: 41694723-41694723 on Build GRCh38
rs200813002 Chr.22: 41694982-41694982 on Build GRCh38
rs958899421 Chr.22: 41694995-41694995 on Build GRCh38
rs1007951260 Chr.22: 41694726-41694726 on Build GRCh38
rs982214131 Chr.22: 41694659-41694659 on Build GRCh38

View Full Product Details