Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10289936-10290479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
UniGene
Hs.590080
Gene Chromosome Location
Chr.3: 10285750-10292947 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035904 c.28C>A p.L10I Chr.3: 10290153-10290153 on Build GRCh38
5863135 c.14G>A p.G5E Chr.3: 10290167-10290167 on Build GRCh38
5863136 c.8C>T p.S3F Chr.3: 10290173-10290173 on Build GRCh38
727483 c.87C>T p.S29S Chr.3: 10290094-10290094 on Build GRCh38
4552122 c.54G>A p.L18L Chr.3: 10290127-10290127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542958062 Chr.3: 10290061-10290061 on Build GRCh38
rs1044968334 Chr.3: 10290407-10290407 on Build GRCh38
rs375723549 Chr.3: 10290059-10290059 on Build GRCh38
rs769018720 Chr.3: 10290128-10290128 on Build GRCh38
rs986636054 Chr.3: 10290310-10290310 on Build GRCh38
rs374058751 Chr.3: 10290395-10290395 on Build GRCh38
rs774004361 Chr.3: 10290064-10290064 on Build GRCh38
rs754766217 Chr.3: 10290010-10290010 on Build GRCh38
rs533262550 Chr.3: 10290182-10290182 on Build GRCh38
rs540101934 Chr.3: 10290141-10290141 on Build GRCh38
rs575291178 Chr.3: 10290325-10290325 on Build GRCh38
rs1042772512 Chr.3: 10290351-10290351 on Build GRCh38
rs535903821 Chr.3: 10290339-10290339 on Build GRCh38
rs1007543943 Chr.3: 10289960-10289960 on Build GRCh38
rs758478747 Chr.3: 10290119-10290119 on Build GRCh38
rs1003604407 Chr.3: 10290159-10290159 on Build GRCh38
rs770322294 Chr.3: 10290214-10290214 on Build GRCh38
rs923729630 Chr.3: 10290338-10290338 on Build GRCh38
rs1017550114 Chr.3: 10289976-10289976 on Build GRCh38
rs564745430 Chr.3: 10290414-10290414 on Build GRCh38
rs770262839 Chr.3: 10290388-10290388 on Build GRCh38
rs762719219 Chr.3: 10290060-10290060 on Build GRCh38
rs375923081 Chr.3: 10290109-10290109 on Build GRCh38
rs780582809 Chr.3: 10290120-10290120 on Build GRCh38
rs546885979 Chr.3: 10290279-10290279 on Build GRCh38
rs36023991 Chr.3: 10290292-10290293 on Build GRCh38
rs1012526184 Chr.3: 10290084-10290084 on Build GRCh38
rs189014950 Chr.3: 10290225-10290225 on Build GRCh38
rs939202324 Chr.3: 10290418-10290418 on Build GRCh38
rs1039113200 Chr.3: 10290336-10290336 on Build GRCh38
rs111516350 Chr.3: 10290228-10290228 on Build GRCh38
rs982669377 Chr.3: 10290022-10290022 on Build GRCh38
rs147996515 Chr.3: 10290162-10290162 on Build GRCh38
rs770928203 Chr.3: 10290301-10290301 on Build GRCh38
rs893819769 Chr.3: 10290209-10290209 on Build GRCh38
rs368923730 Chr.3: 10290074-10290074 on Build GRCh38
rs962318703 Chr.3: 10290177-10290177 on Build GRCh38
rs367817894 Chr.3: 10290305-10290305 on Build GRCh38
rs768945629 Chr.3: 10290058-10290058 on Build GRCh38
rs895332050 Chr.3: 10290389-10290389 on Build GRCh38
rs890575323 Chr.3: 10290169-10290169 on Build GRCh38
rs755490508 Chr.3: 10290275-10290275 on Build GRCh38
rs986318841 Chr.3: 10290363-10290363 on Build GRCh38
rs745412337 Chr.3: 10290212-10290212 on Build GRCh38
rs191554698 Chr.3: 10290428-10290428 on Build GRCh38
rs575581521 Chr.3: 10290001-10290001 on Build GRCh38
rs200551646 Chr.3: 10290095-10290095 on Build GRCh38
rs1021596833 Chr.3: 10290013-10290013 on Build GRCh38
rs1019080068 Chr.3: 10290372-10290372 on Build GRCh38
rs1003798795 Chr.3: 10290380-10290380 on Build GRCh38
rs149396238 Chr.3: 10290110-10290110 on Build GRCh38
rs774664563 Chr.3: 10290161-10290162 on Build GRCh38
rs946765697 Chr.3: 10290341-10290341 on Build GRCh38
rs750654631 Chr.3: 10290192-10290192 on Build GRCh38
rs966921454 Chr.3: 10290021-10290021 on Build GRCh38
rs370910573 Chr.3: 10290187-10290187 on Build GRCh38
rs186245596 Chr.3: 10290313-10290313 on Build GRCh38
rs780022128 Chr.3: 10290200-10290200 on Build GRCh38
rs753048882 Chr.3: 10290332-10290332 on Build GRCh38
rs775924171 Chr.3: 10290027-10290027 on Build GRCh38
rs748344380 Chr.3: 10290244-10290244 on Build GRCh38
rs986688276 Chr.3: 10290451-10290451 on Build GRCh38
rs962764802 Chr.3: 10290406-10290406 on Build GRCh38
rs527311766 Chr.3: 10290268-10290268 on Build GRCh38
rs1014945166 Chr.3: 10290175-10290175 on Build GRCh38
rs200739485 Chr.3: 10289993-10289993 on Build GRCh38
rs745943870 Chr.3: 10290117-10290117 on Build GRCh38
rs369411848 Chr.3: 10290452-10290452 on Build GRCh38
rs942508501 Chr.3: 10290316-10290316 on Build GRCh38
rs999458603 Chr.3: 10290034-10290034 on Build GRCh38
rs763897355 Chr.3: 10290098-10290098 on Build GRCh38
rs567909330 Chr.3: 10290255-10290255 on Build GRCh38
rs749612662 Chr.3: 10290040-10290042 on Build GRCh38
rs955002510 Chr.3: 10290450-10290450 on Build GRCh38
rs755548560 Chr.3: 10290206-10290206 on Build GRCh38
rs753640208 Chr.3: 10290172-10290172 on Build GRCh38
rs747511968 Chr.3: 10290205-10290205 on Build GRCh38
rs747522812 Chr.3: 10290121-10290121 on Build GRCh38
rs371597404 Chr.3: 10290405-10290405 on Build GRCh38
rs559789290 Chr.3: 10290165-10290165 on Build GRCh38
rs777169175 Chr.3: 10290131-10290131 on Build GRCh38
rs367945789 Chr.3: 10290398-10290398 on Build GRCh38
rs759055681 Chr.3: 10290140-10290140 on Build GRCh38
rs758485535 Chr.3: 10290197-10290197 on Build GRCh38
rs557285398 Chr.3: 10290369-10290369 on Build GRCh38
rs1040707192 Chr.3: 10289994-10289994 on Build GRCh38
rs781642540 Chr.3: 10290212-10290212 on Build GRCh38
rs180917811 Chr.3: 10290248-10290248 on Build GRCh38
rs376547309 Chr.3: 10290415-10290415 on Build GRCh38
rs1035181985 Chr.3: 10290251-10290251 on Build GRCh38
rs373150301 Chr.3: 10290183-10290183 on Build GRCh38
rs754016454 Chr.3: 10290107-10290107 on Build GRCh38
rs542292966 Chr.3: 10290298-10290298 on Build GRCh38
rs959101091 Chr.3: 10290053-10290053 on Build GRCh38
rs565109081 Chr.3: 10290338-10290339 on Build GRCh38
rs1027637484 Chr.3: 10290343-10290343 on Build GRCh38
rs933865089 Chr.3: 10290149-10290149 on Build GRCh38
rs779987102 Chr.3: 10289982-10289982 on Build GRCh38
rs1036185171 Chr.3: 10290419-10290419 on Build GRCh38
rs1034794983 Chr.3: 10290046-10290046 on Build GRCh38
rs368309734 Chr.3: 10290344-10290344 on Build GRCh38
rs771527525 Chr.3: 10290078-10290079 on Build GRCh38
rs370367548 Chr.3: 10290136-10290136 on Build GRCh38
rs759334712 Chr.3: 10290067-10290067 on Build GRCh38
rs760559023 Chr.3: 10290156-10290156 on Build GRCh38
rs990523055 Chr.3: 10290083-10290083 on Build GRCh38
rs778060399 Chr.3: 10290396-10290396 on Build GRCh38
rs183593317 Chr.3: 10290216-10290216 on Build GRCh38
rs943806206 Chr.3: 10290238-10290238 on Build GRCh38
rs775447610 Chr.3: 10290147-10290147 on Build GRCh38
rs1053446670 Chr.3: 10290269-10290269 on Build GRCh38
rs1056694723 Chr.3: 10290382-10290382 on Build GRCh38
rs767339804 Chr.3: 10290440-10290440 on Build GRCh38
rs375935115 Chr.3: 10290291-10290291 on Build GRCh38
rs753727460 Chr.3: 10290005-10290005 on Build GRCh38
rs1011165478 Chr.3: 10290012-10290012 on Build GRCh38
rs137923291 Chr.3: 10290431-10290432 on Build GRCh38
rs199668506 Chr.3: 10290097-10290097 on Build GRCh38

View Full Product Details