Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32530173-32530667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI1
Entrez Gene ID
51143
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Gene Aliases
DLC-A, DNCLI1, LIC1
UniGene
Hs.529495
Gene Chromosome Location
Chr.3: 32525971-32570874 on Build GRCh38
Transcripts
NM_016141.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425848 c.1105G>T p.A369S Chr.3: 32530496-32530496 on Build GRCh38
5943612 c.1141-4delT p.? Chr.3: 32530332-32530332 on Build GRCh38
7336528 c.1142C>T p.S381F Chr.3: 32530327-32530327 on Build GRCh38
6328378 c.1185+5G>A p.? Chr.3: 32530279-32530279 on Build GRCh38
3974374 c.1159C>T p.P387S Chr.3: 32530310-32530310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766598323 Chr.3: 32530374-32530374 on Build GRCh38
rs765696176 Chr.3: 32530518-32530518 on Build GRCh38
rs765317815 Chr.3: 32530357-32530357 on Build GRCh38
rs71630522 Chr.3: 32530348-32530348 on Build GRCh38
rs780856138 Chr.3: 32530236-32530236 on Build GRCh38
rs777817388 Chr.3: 32530247-32530247 on Build GRCh38
rs761941642 Chr.3: 32530349-32530349 on Build GRCh38
rs1010501713 Chr.3: 32530419-32530419 on Build GRCh38
rs972007472 Chr.3: 32530643-32530643 on Build GRCh38
rs138771608 Chr.3: 32530635-32530635 on Build GRCh38
rs769665852 Chr.3: 32530246-32530246 on Build GRCh38
rs759161224 Chr.3: 32530450-32530450 on Build GRCh38
rs760136933 Chr.3: 32530485-32530487 on Build GRCh38
rs763055683 Chr.3: 32530546-32530546 on Build GRCh38
rs764625805 Chr.3: 32530346-32530346 on Build GRCh38
rs985209253 Chr.3: 32530473-32530473 on Build GRCh38
rs555665546 Chr.3: 32530271-32530271 on Build GRCh38
rs200363387 Chr.3: 32530311-32530311 on Build GRCh38
rs776926946 Chr.3: 32530498-32530498 on Build GRCh38
rs776809607 Chr.3: 32530331-32530332 on Build GRCh38
rs778315903 Chr.3: 32530440-32530440 on Build GRCh38
rs992552496 Chr.3: 32530593-32530593 on Build GRCh38
rs778616005 Chr.3: 32530331-32530331 on Build GRCh38
rs759113533 Chr.3: 32530302-32530302 on Build GRCh38
rs546953226 Chr.3: 32530332-32530334 on Build GRCh38
rs758582608 Chr.3: 32530368-32530368 on Build GRCh38
rs748870550 Chr.3: 32530248-32530248 on Build GRCh38
rs771672471 Chr.3: 32530464-32530464 on Build GRCh38
rs755644381 Chr.3: 32530571-32530574 on Build GRCh38
rs1049716978 Chr.3: 32530212-32530212 on Build GRCh38
rs35467710 Chr.3: 32530332-32530332 on Build GRCh38
rs997889746 Chr.3: 32530378-32530378 on Build GRCh38
rs1012318392 Chr.3: 32530199-32530199 on Build GRCh38
rs112341904 Chr.3: 32530618-32530618 on Build GRCh38
rs767882610 Chr.3: 32530555-32530559 on Build GRCh38
rs967921348 Chr.3: 32530582-32530582 on Build GRCh38
rs751789308 Chr.3: 32530568-32530568 on Build GRCh38
rs773191391 Chr.3: 32530529-32530529 on Build GRCh38
rs766572988 Chr.3: 32530565-32530565 on Build GRCh38
rs775179366 Chr.3: 32530468-32530468 on Build GRCh38
rs766916517 Chr.3: 32530351-32530352 on Build GRCh38
rs141219462 Chr.3: 32530640-32530640 on Build GRCh38
rs777911437 Chr.3: 32530430-32530430 on Build GRCh38
rs1029324091 Chr.3: 32530384-32530384 on Build GRCh38
rs374523173 Chr.3: 32530243-32530243 on Build GRCh38
rs869079067 Chr.3: 32530347-32530348 on Build GRCh38
rs745333328 Chr.3: 32530442-32530442 on Build GRCh38
rs762316181 Chr.3: 32530514-32530514 on Build GRCh38
rs1019596653 Chr.3: 32530382-32530382 on Build GRCh38
rs773941494 Chr.3: 32530272-32530272 on Build GRCh38
rs140662500 Chr.3: 32530485-32530485 on Build GRCh38
rs769924692 Chr.3: 32530349-32530349 on Build GRCh38
rs939271292 Chr.3: 32530622-32530622 on Build GRCh38
rs755965236 Chr.3: 32530427-32530427 on Build GRCh38
rs753209491 Chr.3: 32530567-32530568 on Build GRCh38
rs752473059 Chr.3: 32530421-32530421 on Build GRCh38
rs761827813 Chr.3: 32530332-32530333 on Build GRCh38
rs115854413 Chr.3: 32530364-32530364 on Build GRCh38
rs548434410 Chr.3: 32530387-32530387 on Build GRCh38
rs751855841 Chr.3: 32530445-32530449 on Build GRCh38
rs569176229 Chr.3: 32530317-32530317 on Build GRCh38
rs532136591 Chr.3: 32530200-32530200 on Build GRCh38
rs201889867 Chr.3: 32530332-32530332 on Build GRCh38
rs143895523 Chr.3: 32530471-32530471 on Build GRCh38
rs541437600 Chr.3: 32530238-32530238 on Build GRCh38
rs749251070 Chr.3: 32530431-32530431 on Build GRCh38
rs978296713 Chr.3: 32530589-32530589 on Build GRCh38
rs549502785 Chr.3: 32530334-32530334 on Build GRCh38

View Full Product Details