Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38062087-38062598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
UniGene
Hs.714499
Gene Chromosome Location
Chr.3: 38038595-38122741 on Build GRCh38
Transcripts
NM_001321153.1 NM_007335.3 NM_007337.3 XM_006713438.3 XM_006713439.3 XM_006713440.3 XM_011534317.2 XM_011534318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203687 c.727G>A p.V243I Chr.3: 38062222-38062222 on Build GRCh38
5948933 c.839C>A p.P280H Chr.3: 38062334-38062334 on Build GRCh38
4508077 c.764C>T p.S255L Chr.3: 38062259-38062259 on Build GRCh38
4509901 c.820C>T p.L274L Chr.3: 38062315-38062315 on Build GRCh38
3593101 c.873+1G>A p.? Chr.3: 38062369-38062369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201476620 Chr.3: 38062133-38062133 on Build GRCh38
rs374147987 Chr.3: 38062170-38062170 on Build GRCh38
rs528457273 Chr.3: 38062390-38062390 on Build GRCh38
rs776231381 Chr.3: 38062229-38062229 on Build GRCh38
rs940058841 Chr.3: 38062193-38062193 on Build GRCh38
rs116202356 Chr.3: 38062285-38062285 on Build GRCh38
rs759076042 Chr.3: 38062391-38062391 on Build GRCh38
rs369795790 Chr.3: 38062477-38062477 on Build GRCh38
rs375270548 Chr.3: 38062363-38062363 on Build GRCh38
rs749458835 Chr.3: 38062324-38062324 on Build GRCh38
rs148212000 Chr.3: 38062222-38062222 on Build GRCh38
rs372876972 Chr.3: 38062539-38062539 on Build GRCh38
rs751988743 Chr.3: 38062166-38062166 on Build GRCh38
rs369375450 Chr.3: 38062210-38062210 on Build GRCh38
rs365331 Chr.3: 38062299-38062299 on Build GRCh38
rs922642053 Chr.3: 38062518-38062518 on Build GRCh38
rs754061070 Chr.3: 38062407-38062407 on Build GRCh38
rs752142620 Chr.3: 38062277-38062277 on Build GRCh38
rs761489333 Chr.3: 38062122-38062122 on Build GRCh38
rs141610254 Chr.3: 38062438-38062438 on Build GRCh38
rs768029521 Chr.3: 38062538-38062538 on Build GRCh38
rs139083277 Chr.3: 38062544-38062544 on Build GRCh38
rs115509072 Chr.3: 38062528-38062528 on Build GRCh38
rs1016538653 Chr.3: 38062306-38062306 on Build GRCh38
rs143108244 Chr.3: 38062570-38062570 on Build GRCh38
rs940089771 Chr.3: 38062466-38062466 on Build GRCh38
rs756436066 Chr.3: 38062319-38062319 on Build GRCh38
rs909850671 Chr.3: 38062456-38062456 on Build GRCh38
rs776704507 Chr.3: 38062360-38062360 on Build GRCh38
rs753574986 Chr.3: 38062258-38062258 on Build GRCh38
rs769948262 Chr.3: 38062196-38062196 on Build GRCh38
rs375721069 Chr.3: 38062557-38062557 on Build GRCh38
rs976841304 Chr.3: 38062510-38062510 on Build GRCh38
rs762600849 Chr.3: 38062261-38062261 on Build GRCh38
rs532010312 Chr.3: 38062467-38062467 on Build GRCh38
rs889007660 Chr.3: 38062276-38062276 on Build GRCh38
rs551103158 Chr.3: 38062519-38062519 on Build GRCh38
rs369208018 Chr.3: 38062397-38062397 on Build GRCh38
rs760485634 Chr.3: 38062221-38062221 on Build GRCh38
rs375717966 Chr.3: 38062401-38062401 on Build GRCh38
rs418495 Chr.3: 38062298-38062298 on Build GRCh38
rs749941183 Chr.3: 38062409-38062410 on Build GRCh38
rs934839045 Chr.3: 38062507-38062507 on Build GRCh38
rs765051280 Chr.3: 38062242-38062242 on Build GRCh38
rs979021102 Chr.3: 38062482-38062482 on Build GRCh38
rs755377750 Chr.3: 38062284-38062284 on Build GRCh38
rs757363624 Chr.3: 38062417-38062417 on Build GRCh38
rs780652256 Chr.3: 38062165-38062165 on Build GRCh38
rs764851225 Chr.3: 38062127-38062127 on Build GRCh38
rs746980614 Chr.3: 38062357-38062357 on Build GRCh38
rs368760298 Chr.3: 38062354-38062354 on Build GRCh38
rs762655651 Chr.3: 38062365-38062365 on Build GRCh38
rs756490994 Chr.3: 38062402-38062402 on Build GRCh38
rs750009204 Chr.3: 38062128-38062128 on Build GRCh38
rs75961453 Chr.3: 38062358-38062358 on Build GRCh38
rs551628345 Chr.3: 38062233-38062233 on Build GRCh38
rs369065647 Chr.3: 38062558-38062558 on Build GRCh38
rs747997784 Chr.3: 38062361-38062361 on Build GRCh38
rs900965895 Chr.3: 38062220-38062220 on Build GRCh38
rs765922410 Chr.3: 38062368-38062368 on Build GRCh38
rs542508168 Chr.3: 38062292-38062292 on Build GRCh38
rs367725661 Chr.3: 38062216-38062216 on Build GRCh38
rs757904431 Chr.3: 38062503-38062503 on Build GRCh38
rs777961309 Chr.3: 38062206-38062206 on Build GRCh38
rs933866875 Chr.3: 38062480-38062480 on Build GRCh38
rs1048858433 Chr.3: 38062116-38062116 on Build GRCh38
rs748378780 Chr.3: 38062194-38062194 on Build GRCh38
rs769201752 Chr.3: 38062395-38062395 on Build GRCh38
rs745588809 Chr.3: 38062397-38062398 on Build GRCh38
rs771070045 Chr.3: 38062352-38062352 on Build GRCh38
rs1035393211 Chr.3: 38062367-38062367 on Build GRCh38
rs531939236 Chr.3: 38062422-38062422 on Build GRCh38
rs867106300 Chr.3: 38062150-38062150 on Build GRCh38
rs781376106 Chr.3: 38062179-38062179 on Build GRCh38
rs866395576 Chr.3: 38062134-38062134 on Build GRCh38
rs775515217 Chr.3: 38062217-38062217 on Build GRCh38
rs779182127 Chr.3: 38062532-38062532 on Build GRCh38
rs772145521 Chr.3: 38062407-38062407 on Build GRCh38
rs146192252 Chr.3: 38062499-38062499 on Build GRCh38
rs778016787 Chr.3: 38062320-38062320 on Build GRCh38
rs966443323 Chr.3: 38062315-38062315 on Build GRCh38
rs977774630 Chr.3: 38062345-38062345 on Build GRCh38
rs774315901 Chr.3: 38062233-38062234 on Build GRCh38
rs761685604 Chr.3: 38062239-38062239 on Build GRCh38
rs765759075 Chr.3: 38062271-38062271 on Build GRCh38

View Full Product Details