Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 43366623-43367126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO10
Entrez Gene ID
55129
Gene Name
anoctamin 10
Gene Aliases
SCAR10, TMEM16K
UniGene
Hs.656657
Gene Chromosome Location
Chr.3: 43366324-43691594 on Build GRCh38
Transcripts
NM_001204831.1 NM_001204832.1 NM_001204833.1 NM_001204834.1 NM_018075.3 XM_011533887.2 XM_017006716.1 XM_017006717.1 XM_017006719.1 XM_017006720.1 XM_017006721.1 XM_017006722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005911 c.1963G>A p.G655R Chr.3: 43366926-43366926 on Build GRCh38
4474914 c.1952C>T p.P651L Chr.3: 43366937-43366937 on Build GRCh38
4117702 c.1939C>G p.L647V Chr.3: 43366950-43366950 on Build GRCh38
1196466 c.1931C>A p.T644N Chr.3: 43366958-43366958 on Build GRCh38
5617185 c.1965G>T p.G655G Chr.3: 43366924-43366924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142565970 Chr.3: 43366670-43366670 on Build GRCh38
rs762148327 Chr.3: 43367002-43367002 on Build GRCh38
rs571804558 Chr.3: 43366995-43366995 on Build GRCh38
rs377647762 Chr.3: 43366896-43366896 on Build GRCh38
rs768832226 Chr.3: 43366994-43366994 on Build GRCh38
rs35884024 Chr.3: 43366715-43366716 on Build GRCh38
rs925663453 Chr.3: 43366876-43366876 on Build GRCh38
rs951493321 Chr.3: 43366741-43366741 on Build GRCh38
rs888413940 Chr.3: 43367020-43367020 on Build GRCh38
rs988373136 Chr.3: 43367101-43367101 on Build GRCh38
rs759676008 Chr.3: 43366957-43366957 on Build GRCh38
rs73831273 Chr.3: 43366660-43366660 on Build GRCh38
rs111871863 Chr.3: 43366727-43366727 on Build GRCh38
rs374056456 Chr.3: 43366871-43366871 on Build GRCh38
rs567669993 Chr.3: 43367006-43367006 on Build GRCh38
rs556549778 Chr.3: 43366831-43366832 on Build GRCh38
rs904271838 Chr.3: 43366669-43366669 on Build GRCh38
rs1017133929 Chr.3: 43366999-43366999 on Build GRCh38
rs371303743 Chr.3: 43366858-43366858 on Build GRCh38
rs913107733 Chr.3: 43366658-43366658 on Build GRCh38
rs774571169 Chr.3: 43366833-43366833 on Build GRCh38
rs776954907 Chr.3: 43366815-43366815 on Build GRCh38
rs560810994 Chr.3: 43366887-43366888 on Build GRCh38
rs553086954 Chr.3: 43367043-43367043 on Build GRCh38
rs777841466 Chr.3: 43366929-43366929 on Build GRCh38
rs996982235 Chr.3: 43367048-43367048 on Build GRCh38
rs538163972 Chr.3: 43366845-43366845 on Build GRCh38
rs560234921 Chr.3: 43366824-43366824 on Build GRCh38
rs964174647 Chr.3: 43366793-43366793 on Build GRCh38
rs952837822 Chr.3: 43366753-43366753 on Build GRCh38
rs913158259 Chr.3: 43366832-43366832 on Build GRCh38
rs866263881 Chr.3: 43366956-43366956 on Build GRCh38
rs751112898 Chr.3: 43366870-43366870 on Build GRCh38
rs200245234 Chr.3: 43366889-43366889 on Build GRCh38
rs369088166 Chr.3: 43366664-43366664 on Build GRCh38
rs1025814851 Chr.3: 43366734-43366734 on Build GRCh38
rs756170502 Chr.3: 43366927-43366927 on Build GRCh38
rs771052268 Chr.3: 43366826-43366826 on Build GRCh38
rs955339704 Chr.3: 43367081-43367081 on Build GRCh38
rs928230995 Chr.3: 43366835-43366835 on Build GRCh38
rs762952996 Chr.3: 43366918-43366920 on Build GRCh38
rs559978110 Chr.3: 43367010-43367010 on Build GRCh38
rs147989825 Chr.3: 43366935-43366935 on Build GRCh38
rs759727363 Chr.3: 43366874-43366874 on Build GRCh38
rs1017410577 Chr.3: 43366780-43366780 on Build GRCh38
rs887376698 Chr.3: 43366981-43366981 on Build GRCh38
rs1025591959 Chr.3: 43366742-43366742 on Build GRCh38
rs747927889 Chr.3: 43366701-43366701 on Build GRCh38
rs975809344 Chr.3: 43366811-43366811 on Build GRCh38
rs1004150292 Chr.3: 43367067-43367067 on Build GRCh38
rs544065871 Chr.3: 43366682-43366682 on Build GRCh38
rs749989628 Chr.3: 43366856-43366856 on Build GRCh38
rs529595858 Chr.3: 43366726-43366726 on Build GRCh38
rs73831275 Chr.3: 43367053-43367053 on Build GRCh38
rs1005722503 Chr.3: 43366779-43366779 on Build GRCh38
rs766059921 Chr.3: 43366868-43366868 on Build GRCh38
rs73831274 Chr.3: 43366710-43366710 on Build GRCh38
rs760388836 Chr.3: 43366914-43366914 on Build GRCh38
rs747153209 Chr.3: 43366963-43366963 on Build GRCh38
rs536682474 Chr.3: 43367028-43367028 on Build GRCh38
rs945790500 Chr.3: 43366659-43366659 on Build GRCh38
rs775797184 Chr.3: 43366962-43366962 on Build GRCh38
rs764369738 Chr.3: 43367027-43367027 on Build GRCh38
rs773525056 Chr.3: 43367013-43367013 on Build GRCh38
rs982405254 Chr.3: 43366832-43366832 on Build GRCh38
rs765756827 Chr.3: 43367026-43367026 on Build GRCh38
rs141722980 Chr.3: 43366905-43366906 on Build GRCh38
rs988103087 Chr.3: 43367087-43367087 on Build GRCh38
rs762538192 Chr.3: 43367015-43367015 on Build GRCh38
rs185583931 Chr.3: 43366846-43366846 on Build GRCh38
rs775593449 Chr.3: 43366926-43366926 on Build GRCh38
rs896056845 Chr.3: 43366723-43366723 on Build GRCh38
rs956966324 Chr.3: 43366839-43366839 on Build GRCh38

View Full Product Details