Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45943401-45943901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR6
Entrez Gene ID
10663
Gene Name
C-X-C motif chemokine receptor 6
Gene Aliases
BONZO, CD186, STRL33, TYMSTR
UniGene
Hs.34526
Gene Chromosome Location
Chr.3: 45940688-45948354 on Build GRCh38
Transcripts
NM_006564.1 XM_011533290.2 XM_011533291.2 NM_024513.3 XM_006713333.3 XM_006713334.3 XM_011534111.2

Gene Symbol
FYCO1
Entrez Gene ID
79443
Gene Name
FYVE and coiled-coil domain containing 1
Gene Aliases
CATC2, CTRCT18, RUFY3, ZFYVE7
UniGene
Hs.200227
Gene Chromosome Location
Chr.3: 45917899-45995824 on Build GRCh38
Transcripts
NM_006564.1 XM_011533290.2 XM_011533291.2 NM_024513.3 XM_006713333.3 XM_006713334.3 XM_011534111.2

SNP Detail

SNP ID Chromosome Location
rs543546527 Chr.3: 45943676-45943676 on Build GRCh38
rs369064798 Chr.3: 45943730-45943730 on Build GRCh38
rs1042795763 Chr.3: 45943494-45943494 on Build GRCh38
rs959232769 Chr.3: 45943738-45943738 on Build GRCh38
rs1003491524 Chr.3: 45943717-45943717 on Build GRCh38
rs973112413 Chr.3: 45943622-45943622 on Build GRCh38
rs554390639 Chr.3: 45943604-45943604 on Build GRCh38
rs978099365 Chr.3: 45943850-45943850 on Build GRCh38
rs536451108 Chr.3: 45943474-45943474 on Build GRCh38
rs924193069 Chr.3: 45943499-45943499 on Build GRCh38
rs915294757 Chr.3: 45943760-45943760 on Build GRCh38
rs1007259124 Chr.3: 45943619-45943619 on Build GRCh38
rs1051191028 Chr.3: 45943603-45943603 on Build GRCh38
rs560001345 Chr.3: 45943846-45943846 on Build GRCh38
rs1034844188 Chr.3: 45943726-45943726 on Build GRCh38
rs6785091 Chr.3: 45943855-45943855 on Build GRCh38
rs763754678 Chr.3: 45943704-45943704 on Build GRCh38
rs577192784 Chr.3: 45943758-45943758 on Build GRCh38
rs950646688 Chr.3: 45943713-45943713 on Build GRCh38
rs915226107 Chr.3: 45943470-45943470 on Build GRCh38
rs541035859 Chr.3: 45943815-45943815 on Build GRCh38
rs867697852 Chr.3: 45943509-45943509 on Build GRCh38
rs759885605 Chr.3: 45943606-45943606 on Build GRCh38
rs983400643 Chr.3: 45943538-45943538 on Build GRCh38
rs949539528 Chr.3: 45943801-45943801 on Build GRCh38
rs934258837 Chr.3: 45943572-45943572 on Build GRCh38
rs907781525 Chr.3: 45943577-45943577 on Build GRCh38
rs55642383 Chr.3: 45943631-45943631 on Build GRCh38
rs565018443 Chr.3: 45943712-45943712 on Build GRCh38

View Full Product Details