Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112122139-112122636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
UniGene
Hs.434247
Gene Chromosome Location
Chr.3: 112086335-112131095 on Build GRCh38
Transcripts
NM_001190259.1 NM_001190260.1 NM_152785.4 XM_005247272.2 XM_011512627.2 XM_011512628.2 XM_011512629.1 XM_011512630.1

Gene Symbol
GCSAM
Entrez Gene ID
257144
Gene Name
germinal center associated signaling and motility
Gene Aliases
GCAT2, GCET2, HGAL
UniGene
Hs.49614
Gene Chromosome Location
Chr.3: 112120841-112133305 on Build GRCh38
Transcripts
NM_001190259.1 NM_001190260.1 NM_152785.4 XM_005247272.2 XM_011512627.2 XM_011512628.2 XM_011512629.1 XM_011512630.1

SNP Detail

SNP ID Chromosome Location
rs965421243 Chr.3: 112122443-112122443 on Build GRCh38
rs146326621 Chr.3: 112122464-112122464 on Build GRCh38
rs919319752 Chr.3: 112122338-112122338 on Build GRCh38
rs34977417 Chr.3: 112122501-112122502 on Build GRCh38
rs967135683 Chr.3: 112122187-112122187 on Build GRCh38
rs779802453 Chr.3: 112122526-112122526 on Build GRCh38
rs972272259 Chr.3: 112122328-112122328 on Build GRCh38
rs930733318 Chr.3: 112122349-112122349 on Build GRCh38
rs1020130319 Chr.3: 112122514-112122514 on Build GRCh38
rs1492489 Chr.3: 112122456-112122456 on Build GRCh38
rs1492490 Chr.3: 112122276-112122276 on Build GRCh38
rs780559680 Chr.3: 112122438-112122438 on Build GRCh38
rs186869598 Chr.3: 112122241-112122241 on Build GRCh38
rs532561613 Chr.3: 112122485-112122485 on Build GRCh38
rs751392781 Chr.3: 112122232-112122232 on Build GRCh38
rs938441465 Chr.3: 112122271-112122271 on Build GRCh38
rs925617046 Chr.3: 112122194-112122194 on Build GRCh38
rs888730764 Chr.3: 112122365-112122371 on Build GRCh38
rs530185511 Chr.3: 112122307-112122307 on Build GRCh38
rs561141565 Chr.3: 112122291-112122291 on Build GRCh38
rs541267315 Chr.3: 112122290-112122290 on Build GRCh38
rs1049020768 Chr.3: 112122376-112122376 on Build GRCh38
rs905238731 Chr.3: 112122439-112122439 on Build GRCh38
rs902900559 Chr.3: 112122575-112122575 on Build GRCh38
rs986826630 Chr.3: 112122245-112122245 on Build GRCh38
rs914449733 Chr.3: 112122543-112122543 on Build GRCh38
rs953646387 Chr.3: 112122210-112122210 on Build GRCh38
rs896824578 Chr.3: 112122289-112122289 on Build GRCh38
rs968676513 Chr.3: 112122586-112122586 on Build GRCh38
rs1027636350 Chr.3: 112122363-112122364 on Build GRCh38
rs1492488 Chr.3: 112122499-112122499 on Build GRCh38
rs999980178 Chr.3: 112122597-112122597 on Build GRCh38
rs978382082 Chr.3: 112122192-112122192 on Build GRCh38

View Full Product Details