Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114159563-114160061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
UniGene
Hs.121478
Gene Chromosome Location
Chr.3: 114127623-114199407 on Build GRCh38
Transcripts
NM_000796.5 NM_001282563.2 NM_001290809.1 NM_033663.5 XM_017005829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752819 c.307T>A p.C103S Chr.3: 114159831-114159831 on Build GRCh38
7365456 c.297C>A p.S99R Chr.3: 114159841-114159841 on Build GRCh38
1250701 c.356T>C p.L119P Chr.3: 114159782-114159782 on Build GRCh38
6162626 c.310G>T p.D104Y Chr.3: 114159828-114159828 on Build GRCh38
4112401 c.345A>G p.T115T Chr.3: 114159793-114159793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530441506 Chr.3: 114159814-114159814 on Build GRCh38
rs755320341 Chr.3: 114159860-114159860 on Build GRCh38
rs978566165 Chr.3: 114159716-114159716 on Build GRCh38
rs200269629 Chr.3: 114159788-114159788 on Build GRCh38
rs201447792 Chr.3: 114159777-114159777 on Build GRCh38
rs763221391 Chr.3: 114159705-114159705 on Build GRCh38
rs779415461 Chr.3: 114160008-114160008 on Build GRCh38
rs200206712 Chr.3: 114159748-114159748 on Build GRCh38
rs754200808 Chr.3: 114159842-114159842 on Build GRCh38
rs371687884 Chr.3: 114159584-114159584 on Build GRCh38
rs200206609 Chr.3: 114159862-114159862 on Build GRCh38
rs1037870110 Chr.3: 114159841-114159841 on Build GRCh38
rs747608272 Chr.3: 114159745-114159745 on Build GRCh38
rs757184273 Chr.3: 114159914-114159914 on Build GRCh38
rs568782509 Chr.3: 114159997-114159997 on Build GRCh38
rs183915085 Chr.3: 114159695-114159695 on Build GRCh38
rs867827307 Chr.3: 114159906-114159906 on Build GRCh38
rs200616464 Chr.3: 114159719-114159719 on Build GRCh38
rs202135599 Chr.3: 114159729-114159729 on Build GRCh38
rs1044090111 Chr.3: 114159942-114159942 on Build GRCh38
rs1046229245 Chr.3: 114159659-114159659 on Build GRCh38
rs769054339 Chr.3: 114159978-114159978 on Build GRCh38
rs761232724 Chr.3: 114159837-114159837 on Build GRCh38
rs898941936 Chr.3: 114159587-114159587 on Build GRCh38
rs538290387 Chr.3: 114159948-114159948 on Build GRCh38
rs1019737055 Chr.3: 114159711-114159711 on Build GRCh38
rs199709036 Chr.3: 114159888-114159888 on Build GRCh38
rs759285810 Chr.3: 114159775-114159775 on Build GRCh38
rs201756825 Chr.3: 114159833-114159833 on Build GRCh38
rs199811701 Chr.3: 114159731-114159731 on Build GRCh38
rs149054363 Chr.3: 114159933-114159933 on Build GRCh38
rs768904348 Chr.3: 114159791-114159791 on Build GRCh38
rs967291549 Chr.3: 114159966-114159966 on Build GRCh38
rs571201566 Chr.3: 114159938-114159938 on Build GRCh38
rs202231573 Chr.3: 114159825-114159825 on Build GRCh38
rs948353610 Chr.3: 114159935-114159935 on Build GRCh38
rs777619426 Chr.3: 114159746-114159746 on Build GRCh38
rs201831382 Chr.3: 114159739-114159739 on Build GRCh38
rs200381524 Chr.3: 114159900-114159900 on Build GRCh38
rs201570903 Chr.3: 114159878-114159878 on Build GRCh38
rs920334839 Chr.3: 114159952-114159952 on Build GRCh38
rs760966434 Chr.3: 114159590-114159590 on Build GRCh38
rs745631217 Chr.3: 114159916-114159916 on Build GRCh38
rs866112130 Chr.3: 114160025-114160025 on Build GRCh38
rs867402333 Chr.3: 114159851-114159851 on Build GRCh38
rs1005037577 Chr.3: 114159691-114159691 on Build GRCh38
rs200053265 Chr.3: 114159717-114159717 on Build GRCh38
rs200897022 Chr.3: 114159767-114159767 on Build GRCh38
rs550566777 Chr.3: 114159976-114159976 on Build GRCh38
rs868178187 Chr.3: 114159932-114159932 on Build GRCh38
rs958113454 Chr.3: 114159996-114159996 on Build GRCh38
rs898813811 Chr.3: 114159701-114159701 on Build GRCh38
rs756246784 Chr.3: 114159812-114159812 on Build GRCh38
rs1047985632 Chr.3: 114159989-114159989 on Build GRCh38
rs71653627 Chr.3: 114159620-114159620 on Build GRCh38
rs767711870 Chr.3: 114159826-114159826 on Build GRCh38
rs1019806695 Chr.3: 114159943-114159943 on Build GRCh38
rs886224882 Chr.3: 114159678-114159678 on Build GRCh38
rs202055756 Chr.3: 114159772-114159772 on Build GRCh38
rs114388605 Chr.3: 114159713-114159713 on Build GRCh38
rs7632503 Chr.3: 114159736-114159736 on Build GRCh38
rs201092150 Chr.3: 114159808-114159808 on Build GRCh38
rs199528192 Chr.3: 114159854-114159854 on Build GRCh38
rs200516160 Chr.3: 114159708-114159708 on Build GRCh38
rs759823322 Chr.3: 114159918-114159918 on Build GRCh38
rs770828404 Chr.3: 114159754-114159754 on Build GRCh38
rs886057789 Chr.3: 114159836-114159836 on Build GRCh38
rs144960688 Chr.3: 114159706-114159706 on Build GRCh38
rs371424110 Chr.3: 114159883-114159883 on Build GRCh38
rs200553872 Chr.3: 114159876-114159876 on Build GRCh38
rs201249206 Chr.3: 114159903-114159903 on Build GRCh38
rs780482392 Chr.3: 114159726-114159726 on Build GRCh38
rs199595684 Chr.3: 114159915-114159915 on Build GRCh38
rs752349435 Chr.3: 114159871-114159871 on Build GRCh38
rs764829977 Chr.3: 114159861-114159861 on Build GRCh38
rs766862523 Chr.3: 114159839-114159839 on Build GRCh38
rs369256865 Chr.3: 114159707-114159707 on Build GRCh38
rs774740867 Chr.3: 114159805-114159805 on Build GRCh38

View Full Product Details