Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126419913-126420312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP100
Entrez Gene ID
348807
Gene Name
cilia and flagella associated protein 100
Gene Aliases
CCDC37, MIA1
UniGene
Hs.591305
Gene Chromosome Location
Chr.3: 126394893-126436556 on Build GRCh38
Transcripts
NM_182628.2 XM_006713623.2 XM_017006320.1 XM_017006321.1 XM_017006322.1 XM_017006323.1 XM_017006324.1 XM_017006325.1 XM_017006326.1 XM_017006327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5452891 c.948G>A p.W316* Chr.3: 126420014-126420014 on Build GRCh38
4138400 c.1014C>A p.S338R Chr.3: 126420161-126420161 on Build GRCh38
6988616 c.976C>T p.P326S Chr.3: 126420123-126420123 on Build GRCh38
279650 c.972G>T p.K324N Chr.3: 126420119-126420119 on Build GRCh38
293184 c.996G>A p.T332T Chr.3: 126420143-126420143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771585406 Chr.3: 126420159-126420159 on Build GRCh38
rs769809248 Chr.3: 126420170-126420170 on Build GRCh38
rs778577840 Chr.3: 126420230-126420230 on Build GRCh38
rs751551024 Chr.3: 126420160-126420161 on Build GRCh38
rs774688196 Chr.3: 126419950-126419950 on Build GRCh38
rs763155055 Chr.3: 126420198-126420198 on Build GRCh38
rs775441794 Chr.3: 126420247-126420247 on Build GRCh38
rs768092890 Chr.3: 126420032-126420032 on Build GRCh38
rs753309017 Chr.3: 126420278-126420278 on Build GRCh38
rs11714780 Chr.3: 126420102-126420102 on Build GRCh38
rs773611862 Chr.3: 126420088-126420088 on Build GRCh38
rs757972509 Chr.3: 126420125-126420126 on Build GRCh38
rs761214124 Chr.3: 126420037-126420037 on Build GRCh38
rs779509260 Chr.3: 126420137-126420148 on Build GRCh38
rs375786267 Chr.3: 126420148-126420148 on Build GRCh38
rs755583014 Chr.3: 126420152-126420152 on Build GRCh38
rs201666882 Chr.3: 126420043-126420043 on Build GRCh38
rs781077095 Chr.3: 126420280-126420280 on Build GRCh38
rs767475855 Chr.3: 126420212-126420212 on Build GRCh38
rs757733856 Chr.3: 126420237-126420237 on Build GRCh38
rs11714759 Chr.3: 126420171-126420171 on Build GRCh38
rs146000789 Chr.3: 126420164-126420164 on Build GRCh38
rs373762802 Chr.3: 126420045-126420045 on Build GRCh38
rs777519725 Chr.3: 126419988-126419988 on Build GRCh38
rs764739308 Chr.3: 126419959-126419959 on Build GRCh38
rs200743173 Chr.3: 126420076-126420076 on Build GRCh38
rs774704801 Chr.3: 126420027-126420027 on Build GRCh38
rs752898491 Chr.3: 126420139-126420139 on Build GRCh38
rs373852507 Chr.3: 126420143-126420143 on Build GRCh38
rs190717627 Chr.3: 126420031-126420031 on Build GRCh38
rs746443821 Chr.3: 126419937-126419937 on Build GRCh38
rs374555935 Chr.3: 126420077-126420077 on Build GRCh38
rs371860709 Chr.3: 126419959-126419959 on Build GRCh38
rs762452330 Chr.3: 126419954-126419954 on Build GRCh38
rs964229607 Chr.3: 126419933-126419933 on Build GRCh38
rs761549040 Chr.3: 126420204-126420204 on Build GRCh38
rs765871561 Chr.3: 126420218-126420218 on Build GRCh38
rs942090974 Chr.3: 126419958-126419958 on Build GRCh38
rs6795815 Chr.3: 126420181-126420181 on Build GRCh38
rs111556812 Chr.3: 126420054-126420054 on Build GRCh38
rs770463887 Chr.3: 126420001-126420001 on Build GRCh38
rs916627413 Chr.3: 126420235-126420235 on Build GRCh38
rs1030210346 Chr.3: 126420131-126420131 on Build GRCh38
rs571233699 Chr.3: 126419941-126419941 on Build GRCh38
rs780750708 Chr.3: 126420066-126420066 on Build GRCh38
rs757177256 Chr.3: 126420064-126420064 on Build GRCh38
rs768786171 Chr.3: 126420260-126420260 on Build GRCh38
rs756023195 Chr.3: 126420214-126420214 on Build GRCh38
rs765585207 Chr.3: 126420126-126420126 on Build GRCh38
rs746431953 Chr.3: 126420242-126420242 on Build GRCh38
rs1047120775 Chr.3: 126420254-126420254 on Build GRCh38
rs760532731 Chr.3: 126420273-126420273 on Build GRCh38
rs923474937 Chr.3: 126420149-126420149 on Build GRCh38
rs776018764 Chr.3: 126420241-126420241 on Build GRCh38
rs2271630 Chr.3: 126419935-126419935 on Build GRCh38
rs369591163 Chr.3: 126420090-126420090 on Build GRCh38
rs781053138 Chr.3: 126420147-126420147 on Build GRCh38
rs746557093 Chr.3: 126419994-126419994 on Build GRCh38
rs754111487 Chr.3: 126419954-126419955 on Build GRCh38
rs764573230 Chr.3: 126419940-126419942 on Build GRCh38
rs779585872 Chr.3: 126420153-126420153 on Build GRCh38
rs55997846 Chr.3: 126420243-126420243 on Build GRCh38
rs373731054 Chr.3: 126419987-126419987 on Build GRCh38
rs113527043 Chr.3: 126420145-126420145 on Build GRCh38
rs778917672 Chr.3: 126419980-126419980 on Build GRCh38
rs144751770 Chr.3: 126420106-126420106 on Build GRCh38
rs774518439 Chr.3: 126420266-126420266 on Build GRCh38
rs375845011 Chr.3: 126420269-126420269 on Build GRCh38
rs767847846 Chr.3: 126419956-126419956 on Build GRCh38
rs750401050 Chr.3: 126420213-126420213 on Build GRCh38
rs750077115 Chr.3: 126420032-126420033 on Build GRCh38
rs111705111 Chr.3: 126420142-126420142 on Build GRCh38
rs142532946 Chr.3: 126420242-126420242 on Build GRCh38
rs772978195 Chr.3: 126420270-126420270 on Build GRCh38
rs572580590 Chr.3: 126420010-126420010 on Build GRCh38
rs202168304 Chr.3: 126420156-126420156 on Build GRCh38
rs749333683 Chr.3: 126420255-126420255 on Build GRCh38
rs191763154 Chr.3: 126420275-126420275 on Build GRCh38
rs745319464 Chr.3: 126420068-126420068 on Build GRCh38
rs570287700 Chr.3: 126420002-126420002 on Build GRCh38
rs772363155 Chr.3: 126420083-126420083 on Build GRCh38
rs951007664 Chr.3: 126420252-126420252 on Build GRCh38
rs773553711 Chr.3: 126420033-126420033 on Build GRCh38
rs766623334 Chr.3: 126420041-126420041 on Build GRCh38
rs551358419 Chr.3: 126419940-126419940 on Build GRCh38
rs372846486 Chr.3: 126420268-126420268 on Build GRCh38
rs368926593 Chr.3: 126420203-126420203 on Build GRCh38
rs761531040 Chr.3: 126419958-126419959 on Build GRCh38
rs868061166 Chr.3: 126420124-126420125 on Build GRCh38
rs755237057 Chr.3: 126419978-126419978 on Build GRCh38
rs111320520 Chr.3: 126419946-126419946 on Build GRCh38
rs745581400 Chr.3: 126420007-126420007 on Build GRCh38
rs751505171 Chr.3: 126420057-126420057 on Build GRCh38
rs776862604 Chr.3: 126420092-126420092 on Build GRCh38
rs747406810 Chr.3: 126420231-126420231 on Build GRCh38
rs368552983 Chr.3: 126419944-126419944 on Build GRCh38
rs564907952 Chr.3: 126420223-126420223 on Build GRCh38
rs561868340 Chr.3: 126419936-126419936 on Build GRCh38
rs760084651 Chr.3: 126420167-126420167 on Build GRCh38
rs370457546 Chr.3: 126420091-126420091 on Build GRCh38
rs1877967 Chr.3: 126420046-126420046 on Build GRCh38
rs752557161 Chr.3: 126420049-126420049 on Build GRCh38
rs758085734 Chr.3: 126420052-126420052 on Build GRCh38
rs377186686 Chr.3: 126420219-126420219 on Build GRCh38
rs565242404 Chr.3: 126419964-126419964 on Build GRCh38
rs59283185 Chr.3: 126420157-126420157 on Build GRCh38

View Full Product Details