Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 180619725-180620223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC39
Entrez Gene ID
339829
Gene Name
coiled-coil domain containing 39
Gene Aliases
CILD14, FAP59
UniGene
Hs.712820
Gene Chromosome Location
Chr.3: 180614008-180679495 on Build GRCh38
Transcripts
NM_181426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738881 c.2057A>G p.N686S Chr.3: 180619912-180619912 on Build GRCh38
6622140 c.2120A>C p.N707T Chr.3: 180619849-180619849 on Build GRCh38
6622141 c.2079C>T p.Y693Y Chr.3: 180619890-180619890 on Build GRCh38
1041653 c.2047G>A p.A683T Chr.3: 180619922-180619922 on Build GRCh38
6163841 c.2058C>A p.N686K Chr.3: 180619911-180619911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772234860 Chr.3: 180619767-180619767 on Build GRCh38
rs748321086 Chr.3: 180619813-180619813 on Build GRCh38
rs764617529 Chr.3: 180619932-180619932 on Build GRCh38
rs527502014 Chr.3: 180619927-180619927 on Build GRCh38
rs761167681 Chr.3: 180619846-180619848 on Build GRCh38
rs751147958 Chr.3: 180619964-180619964 on Build GRCh38
rs377564125 Chr.3: 180619786-180619787 on Build GRCh38
rs953349340 Chr.3: 180619860-180619860 on Build GRCh38
rs771560076 Chr.3: 180619918-180619918 on Build GRCh38
rs904974192 Chr.3: 180620044-180620044 on Build GRCh38
rs375292349 Chr.3: 180619766-180619766 on Build GRCh38
rs971075780 Chr.3: 180619857-180619857 on Build GRCh38
rs201586263 Chr.3: 180619912-180619912 on Build GRCh38
rs556121787 Chr.3: 180619787-180619787 on Build GRCh38
rs780247255 Chr.3: 180619877-180619877 on Build GRCh38
rs561979601 Chr.3: 180619814-180619814 on Build GRCh38
rs949551804 Chr.3: 180619956-180619956 on Build GRCh38
rs373905898 Chr.3: 180619982-180619982 on Build GRCh38
rs753833834 Chr.3: 180619833-180619834 on Build GRCh38
rs765555238 Chr.3: 180619842-180619842 on Build GRCh38
rs763893153 Chr.3: 180619787-180619787 on Build GRCh38
rs770727088 Chr.3: 180619801-180619801 on Build GRCh38
rs762287443 Chr.3: 180619952-180619952 on Build GRCh38
rs754224849 Chr.3: 180619837-180619837 on Build GRCh38
rs781385113 Chr.3: 180620001-180620001 on Build GRCh38
rs1039006472 Chr.3: 180620122-180620122 on Build GRCh38
rs745596147 Chr.3: 180620015-180620015 on Build GRCh38
rs930977227 Chr.3: 180620057-180620057 on Build GRCh38
rs779059515 Chr.3: 180619762-180619762 on Build GRCh38
rs369900592 Chr.3: 180619768-180619768 on Build GRCh38
rs749284137 Chr.3: 180620009-180620009 on Build GRCh38
rs764725359 Chr.3: 180619916-180619917 on Build GRCh38
rs541172213 Chr.3: 180620038-180620038 on Build GRCh38
rs766064478 Chr.3: 180619993-180619993 on Build GRCh38
rs1004504294 Chr.3: 180620153-180620153 on Build GRCh38
rs769886847 Chr.3: 180619889-180619889 on Build GRCh38
rs749995752 Chr.3: 180619933-180619934 on Build GRCh38
rs770888279 Chr.3: 180619909-180619909 on Build GRCh38
rs376782159 Chr.3: 180619890-180619890 on Build GRCh38
rs965192057 Chr.3: 180619800-180619800 on Build GRCh38
rs748174105 Chr.3: 180619769-180619769 on Build GRCh38
rs777800918 Chr.3: 180619894-180619894 on Build GRCh38
rs371256070 Chr.3: 180620175-180620181 on Build GRCh38
rs989194150 Chr.3: 180619940-180619940 on Build GRCh38
rs113196242 Chr.3: 180620067-180620067 on Build GRCh38
rs925538032 Chr.3: 180619906-180619906 on Build GRCh38
rs757259586 Chr.3: 180620010-180620010 on Build GRCh38
rs769849329 Chr.3: 180619783-180619783 on Build GRCh38
rs1052385589 Chr.3: 180620073-180620073 on Build GRCh38
rs774013177 Chr.3: 180619807-180619807 on Build GRCh38
rs913667094 Chr.3: 180620028-180620028 on Build GRCh38
rs780429898 Chr.3: 180619950-180619952 on Build GRCh38
rs1024870119 Chr.3: 180619855-180619855 on Build GRCh38
rs777162306 Chr.3: 180619951-180619951 on Build GRCh38
rs750746675 Chr.3: 180619966-180619966 on Build GRCh38
rs148793330 Chr.3: 180620173-180620179 on Build GRCh38
rs778776316 Chr.3: 180620014-180620014 on Build GRCh38
rs62292364 Chr.3: 180619949-180619949 on Build GRCh38
rs988316404 Chr.3: 180619914-180619914 on Build GRCh38
rs980451572 Chr.3: 180619867-180619867 on Build GRCh38
rs373567562 Chr.3: 180619868-180619868 on Build GRCh38
rs190816323 Chr.3: 180620093-180620093 on Build GRCh38
rs764529780 Chr.3: 180619835-180619835 on Build GRCh38
rs751710659 Chr.3: 180619986-180619986 on Build GRCh38
rs750707206 Chr.3: 180619848-180619848 on Build GRCh38
rs761177022 Chr.3: 180619824-180619824 on Build GRCh38
rs775083303 Chr.3: 180619925-180619925 on Build GRCh38
rs899134514 Chr.3: 180620193-180620193 on Build GRCh38
rs752148109 Chr.3: 180619822-180619822 on Build GRCh38
rs758649358 Chr.3: 180619973-180619973 on Build GRCh38
rs767117097 Chr.3: 180619816-180619816 on Build GRCh38
rs186307085 Chr.3: 180619789-180619789 on Build GRCh38
rs752965627 Chr.3: 180620019-180620023 on Build GRCh38
rs541780919 Chr.3: 180619811-180619811 on Build GRCh38
rs377395103 Chr.3: 180619991-180619991 on Build GRCh38
rs887023525 Chr.3: 180620072-180620072 on Build GRCh38
rs943651328 Chr.3: 180620119-180620119 on Build GRCh38
rs201554599 Chr.3: 180620043-180620043 on Build GRCh38

View Full Product Details