Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116518606-116519147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD58
Entrez Gene ID
965
Gene Name
CD58 molecule
Gene Aliases
LFA-3, LFA3, ag3
UniGene
Hs.34341
Gene Chromosome Location
Chr.1: 116487140-116571093 on Build GRCh38
Transcripts
NM_001144822.1 NM_001779.2 NR_026665.1 XM_017002869.1

SNP Detail

SNP ID Chromosome Location
rs769782430 Chr.1: 116519085-116519085 on Build GRCh38
rs888729529 Chr.1: 116518702-116518702 on Build GRCh38
rs879248002 Chr.1: 116518679-116518679 on Build GRCh38
rs969386933 Chr.1: 116518738-116518738 on Build GRCh38
rs565638201 Chr.1: 116518729-116518729 on Build GRCh38
rs551421497 Chr.1: 116519080-116519084 on Build GRCh38
rs559155884 Chr.1: 116518862-116518862 on Build GRCh38
rs974035948 Chr.1: 116518756-116518756 on Build GRCh38
rs537633249 Chr.1: 116518803-116518803 on Build GRCh38
rs888623071 Chr.1: 116519032-116519032 on Build GRCh38
rs761668072 Chr.1: 116518924-116518924 on Build GRCh38
rs913141672 Chr.1: 116518813-116518813 on Build GRCh38
rs531377720 Chr.1: 116518673-116518673 on Build GRCh38
rs947333520 Chr.1: 116518849-116518849 on Build GRCh38
rs375907540 Chr.1: 116518982-116518982 on Build GRCh38
rs139790985 Chr.1: 116518961-116518961 on Build GRCh38
rs532448378 Chr.1: 116518965-116518965 on Build GRCh38
rs145970247 Chr.1: 116518930-116518930 on Build GRCh38
rs369968095 Chr.1: 116518705-116518706 on Build GRCh38
rs563436606 Chr.1: 116519087-116519087 on Build GRCh38
rs930151652 Chr.1: 116518997-116518997 on Build GRCh38
rs183871651 Chr.1: 116518939-116518939 on Build GRCh38
rs114784750 Chr.1: 116518765-116518765 on Build GRCh38
rs750318125 Chr.1: 116519024-116519024 on Build GRCh38
rs754156942 Chr.1: 116518766-116518766 on Build GRCh38
rs10802190 Chr.1: 116518762-116518762 on Build GRCh38
rs997705965 Chr.1: 116518922-116518922 on Build GRCh38
rs140150510 Chr.1: 116518703-116518703 on Build GRCh38
rs181139888 Chr.1: 116518699-116518699 on Build GRCh38
rs982164716 Chr.1: 116518754-116518754 on Build GRCh38
rs550317137 Chr.1: 116519054-116519054 on Build GRCh38
rs995525885 Chr.1: 116519030-116519030 on Build GRCh38
rs1048113906 Chr.1: 116518972-116518973 on Build GRCh38
rs1004345914 Chr.1: 116518983-116518983 on Build GRCh38
rs978707221 Chr.1: 116518870-116518870 on Build GRCh38
rs775740735 Chr.1: 116518695-116518695 on Build GRCh38
rs574759690 Chr.1: 116519049-116519049 on Build GRCh38
rs962707663 Chr.1: 116518747-116518747 on Build GRCh38
rs547398985 Chr.1: 116518705-116518705 on Build GRCh38
rs910857692 Chr.1: 116518921-116518921 on Build GRCh38
rs118001644 Chr.1: 116518755-116518755 on Build GRCh38
rs368099891 Chr.1: 116518852-116518852 on Build GRCh38
rs372234574 Chr.1: 116518866-116518866 on Build GRCh38
rs945221126 Chr.1: 116518810-116518810 on Build GRCh38
rs549128018 Chr.1: 116518801-116518801 on Build GRCh38
rs866940674 Chr.1: 116519118-116519118 on Build GRCh38
rs549609857 Chr.1: 116518678-116518678 on Build GRCh38
rs541883244 Chr.1: 116519050-116519050 on Build GRCh38
rs1024019014 Chr.1: 116518719-116518719 on Build GRCh38

View Full Product Details