Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117865562-117866061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs182491737 Chr.1: 117865702-117865702 on Build GRCh38
rs938090041 Chr.1: 117865659-117865659 on Build GRCh38
rs200524703 Chr.1: 117865732-117865732 on Build GRCh38
rs535181671 Chr.1: 117865599-117865599 on Build GRCh38
rs893244111 Chr.1: 117865587-117865587 on Build GRCh38
rs907711479 Chr.1: 117865615-117865615 on Build GRCh38
rs565924898 Chr.1: 117865743-117865743 on Build GRCh38
rs927085094 Chr.1: 117866016-117866016 on Build GRCh38
rs552844422 Chr.1: 117865591-117865591 on Build GRCh38
rs539641688 Chr.1: 117865827-117865827 on Build GRCh38
rs190037886 Chr.1: 117865614-117865614 on Build GRCh38
rs1027335581 Chr.1: 117865976-117865976 on Build GRCh38
rs941806184 Chr.1: 117865621-117865621 on Build GRCh38
rs938114989 Chr.1: 117866022-117866022 on Build GRCh38
rs79357772 Chr.1: 117865606-117865606 on Build GRCh38
rs990530755 Chr.1: 117865695-117865695 on Build GRCh38
rs576638417 Chr.1: 117865803-117865803 on Build GRCh38
rs537869134 Chr.1: 117865981-117865981 on Build GRCh38
rs117401979 Chr.1: 117865887-117865887 on Build GRCh38
rs893179532 Chr.1: 117865846-117865846 on Build GRCh38
rs983168024 Chr.1: 117865998-117865998 on Build GRCh38
rs1037530204 Chr.1: 117865650-117865650 on Build GRCh38
rs983372703 Chr.1: 117865957-117865957 on Build GRCh38
rs1035337350 Chr.1: 117865931-117865931 on Build GRCh38
rs908058743 Chr.1: 117865742-117865742 on Build GRCh38
rs369649850 Chr.1: 117865959-117865962 on Build GRCh38
rs963285953 Chr.1: 117865979-117865979 on Build GRCh38
rs778617332 Chr.1: 117865897-117865897 on Build GRCh38
rs1020759599 Chr.1: 117865870-117865870 on Build GRCh38
rs141344381 Chr.1: 117865988-117865988 on Build GRCh38
rs1055550099 Chr.1: 117865675-117865675 on Build GRCh38
rs920323211 Chr.1: 117865730-117865730 on Build GRCh38
rs563264520 Chr.1: 117865959-117865959 on Build GRCh38
rs768793525 Chr.1: 117865923-117865923 on Build GRCh38
rs868655437 Chr.1: 117865713-117865713 on Build GRCh38
rs535862869 Chr.1: 117865665-117865665 on Build GRCh38
rs1001029405 Chr.1: 117865848-117865848 on Build GRCh38
rs1014258712 Chr.1: 117865745-117865745 on Build GRCh38
rs17037690 Chr.1: 117865653-117865653 on Build GRCh38
rs747631838 Chr.1: 117865942-117865942 on Build GRCh38
rs1014881326 Chr.1: 117865972-117865972 on Build GRCh38
rs944921065 Chr.1: 117865764-117865764 on Build GRCh38
rs17037692 Chr.1: 117865835-117865835 on Build GRCh38
rs780858865 Chr.1: 117865864-117865864 on Build GRCh38
rs375928317 Chr.1: 117865853-117865853 on Build GRCh38
rs748872843 Chr.1: 117866009-117866009 on Build GRCh38
rs187609340 Chr.1: 117866025-117866025 on Build GRCh38
rs541843594 Chr.1: 117865941-117865941 on Build GRCh38
rs558375855 Chr.1: 117865797-117865797 on Build GRCh38
rs1010721167 Chr.1: 117865596-117865597 on Build GRCh38
rs951765463 Chr.1: 117865980-117865980 on Build GRCh38
rs1003412460 Chr.1: 117865908-117865908 on Build GRCh38
rs954799866 Chr.1: 117865598-117865598 on Build GRCh38
rs543301234 Chr.1: 117865741-117865741 on Build GRCh38
rs185668664 Chr.1: 117865597-117865597 on Build GRCh38
rs74113066 Chr.1: 117865736-117865736 on Build GRCh38
rs114280785 Chr.1: 117866010-117866010 on Build GRCh38

View Full Product Details