Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40354360-40354874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA9
Entrez Gene ID
55584
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
HSA243342, NACHRA9
UniGene
Hs.272278
Gene Chromosome Location
Chr.4: 40335329-40355217 on Build GRCh38
Transcripts
NM_017581.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
51589 c.1311C>A p.D437E Chr.4: 40354391-40354391 on Build GRCh38
4124641 c.1440G>T p.*480Y Chr.4: 40354520-40354520 on Build GRCh38
4124640 c.1289A>G p.Y430C Chr.4: 40354369-40354369 on Build GRCh38
1665421 c.1306_1307delAA p.K436fs*11 Chr.4: 40354386-40354387 on Build GRCh38
3603651 c.1383G>A p.W461* Chr.4: 40354463-40354463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763435160 Chr.4: 40354488-40354488 on Build GRCh38
rs770473967 Chr.4: 40354441-40354441 on Build GRCh38
rs370016019 Chr.4: 40354410-40354410 on Build GRCh38
rs151059510 Chr.4: 40354759-40354759 on Build GRCh38
rs749110452 Chr.4: 40354604-40354604 on Build GRCh38
rs756088452 Chr.4: 40354503-40354503 on Build GRCh38
rs778435499 Chr.4: 40354388-40354388 on Build GRCh38
rs753133730 Chr.4: 40354385-40354385 on Build GRCh38
rs200608832 Chr.4: 40354449-40354449 on Build GRCh38
rs4861327 Chr.4: 40354697-40354697 on Build GRCh38
rs146751601 Chr.4: 40354513-40354513 on Build GRCh38
rs6819816 Chr.4: 40354763-40354763 on Build GRCh38
rs759080788 Chr.4: 40354450-40354450 on Build GRCh38
rs550444282 Chr.4: 40354496-40354496 on Build GRCh38
rs759102706 Chr.4: 40354547-40354547 on Build GRCh38
rs564504462 Chr.4: 40354426-40354426 on Build GRCh38
rs752236042 Chr.4: 40354480-40354480 on Build GRCh38
rs889848486 Chr.4: 40354820-40354820 on Build GRCh38
rs545777062 Chr.4: 40354791-40354791 on Build GRCh38
rs777517452 Chr.4: 40354526-40354526 on Build GRCh38
rs566123376 Chr.4: 40354584-40354584 on Build GRCh38
rs948405911 Chr.4: 40354636-40354636 on Build GRCh38
rs10009228 Chr.4: 40354405-40354405 on Build GRCh38
rs746125534 Chr.4: 40354398-40354398 on Build GRCh38
rs925888166 Chr.4: 40354689-40354689 on Build GRCh38
rs115863896 Chr.4: 40354663-40354663 on Build GRCh38
rs143447364 Chr.4: 40354436-40354436 on Build GRCh38
rs559200096 Chr.4: 40354834-40354834 on Build GRCh38
rs750519738 Chr.4: 40354499-40354499 on Build GRCh38
rs976982211 Chr.4: 40354492-40354492 on Build GRCh38
rs1003330176 Chr.4: 40354582-40354582 on Build GRCh38
rs1052918557 Chr.4: 40354705-40354705 on Build GRCh38
rs953391533 Chr.4: 40354415-40354415 on Build GRCh38
rs918411635 Chr.4: 40354829-40354829 on Build GRCh38
rs758621226 Chr.4: 40354387-40354387 on Build GRCh38
rs373316531 Chr.4: 40354562-40354562 on Build GRCh38
rs769176565 Chr.4: 40354548-40354548 on Build GRCh38
rs776369420 Chr.4: 40354544-40354544 on Build GRCh38
rs1044783303 Chr.4: 40354522-40354522 on Build GRCh38
rs780380156 Chr.4: 40354504-40354504 on Build GRCh38
rs1018979149 Chr.4: 40354758-40354758 on Build GRCh38
rs181978594 Chr.4: 40354397-40354397 on Build GRCh38
rs764743328 Chr.4: 40354384-40354384 on Build GRCh38
rs865863614 Chr.4: 40354839-40354839 on Build GRCh38
rs145376238 Chr.4: 40354629-40354629 on Build GRCh38
rs139982841 Chr.4: 40354435-40354435 on Build GRCh38
rs868864375 Chr.4: 40354802-40354802 on Build GRCh38
rs77225431 Chr.4: 40354537-40354537 on Build GRCh38
rs547079792 Chr.4: 40354753-40354753 on Build GRCh38
rs757479961 Chr.4: 40354394-40354394 on Build GRCh38
rs771890826 Chr.4: 40354514-40354514 on Build GRCh38
rs139871234 Chr.4: 40354589-40354589 on Build GRCh38
rs371532896 Chr.4: 40354498-40354498 on Build GRCh38
rs752153552 Chr.4: 40354495-40354495 on Build GRCh38
rs760571804 Chr.4: 40354437-40354437 on Build GRCh38
rs775492724 Chr.4: 40354534-40354534 on Build GRCh38
rs35867374 Chr.4: 40354411-40354412 on Build GRCh38
rs764550636 Chr.4: 40354493-40354493 on Build GRCh38
rs764950252 Chr.4: 40354459-40354459 on Build GRCh38
rs747986258 Chr.4: 40354412-40354412 on Build GRCh38
rs752100723 Chr.4: 40354393-40354393 on Build GRCh38

View Full Product Details