Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42110764-42111262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND4
Entrez Gene ID
389206
Gene Name
BEN domain containing 4
Gene Aliases
CCDC4
UniGene
Hs.120591
Gene Chromosome Location
Chr.4: 42110853-42152878 on Build GRCh38
Transcripts
NM_001159547.1 NM_207406.3 XM_017008185.1

SNP Detail

SNP ID Chromosome Location
rs1043539330 Chr.4: 42111176-42111176 on Build GRCh38
rs576991854 Chr.4: 42110959-42110959 on Build GRCh38
rs916906049 Chr.4: 42110979-42110979 on Build GRCh38
rs904333827 Chr.4: 42111179-42111179 on Build GRCh38
rs948392392 Chr.4: 42110991-42110991 on Build GRCh38
rs927564548 Chr.4: 42111112-42111112 on Build GRCh38
rs965047096 Chr.4: 42111029-42111029 on Build GRCh38
rs572846978 Chr.4: 42110810-42110810 on Build GRCh38
rs879696444 Chr.4: 42110990-42110990 on Build GRCh38
rs546105127 Chr.4: 42110816-42110816 on Build GRCh38
rs192935515 Chr.4: 42111089-42111089 on Build GRCh38
rs986158413 Chr.4: 42110817-42110817 on Build GRCh38
rs1002840838 Chr.4: 42110864-42110864 on Build GRCh38
rs1053963115 Chr.4: 42111064-42111064 on Build GRCh38
rs74688673 Chr.4: 42111091-42111091 on Build GRCh38
rs1035724370 Chr.4: 42110888-42110888 on Build GRCh38
rs760303921 Chr.4: 42111067-42111067 on Build GRCh38
rs950637190 Chr.4: 42111196-42111196 on Build GRCh38
rs1010022623 Chr.4: 42110916-42110916 on Build GRCh38
rs973268168 Chr.4: 42110894-42110894 on Build GRCh38
rs1010243933 Chr.4: 42111175-42111175 on Build GRCh38
rs954736453 Chr.4: 42110814-42110814 on Build GRCh38
rs948361760 Chr.4: 42111233-42111233 on Build GRCh38
rs963494933 Chr.4: 42111125-42111125 on Build GRCh38
rs533978800 Chr.4: 42110865-42110865 on Build GRCh38
rs910674434 Chr.4: 42110857-42110857 on Build GRCh38
rs1007151813 Chr.4: 42111198-42111198 on Build GRCh38
rs758980551 Chr.4: 42111211-42111211 on Build GRCh38
rs550589272 Chr.4: 42110986-42110986 on Build GRCh38
rs976437490 Chr.4: 42111041-42111041 on Build GRCh38
rs552431133 Chr.4: 42111170-42111170 on Build GRCh38
rs10018324 Chr.4: 42111232-42111232 on Build GRCh38
rs1009837110 Chr.4: 42111076-42111076 on Build GRCh38
rs13756 Chr.4: 42111224-42111224 on Build GRCh38
rs891875104 Chr.4: 42111173-42111173 on Build GRCh38
rs7659700 Chr.4: 42110890-42110890 on Build GRCh38
rs532699913 Chr.4: 42111103-42111103 on Build GRCh38
rs572899497 Chr.4: 42110996-42110996 on Build GRCh38
rs1018628344 Chr.4: 42111000-42111000 on Build GRCh38
rs754448686 Chr.4: 42110947-42110947 on Build GRCh38
rs1057100381 Chr.4: 42111166-42111166 on Build GRCh38
rs973620428 Chr.4: 42111140-42111141 on Build GRCh38
rs778555408 Chr.4: 42110987-42110987 on Build GRCh38
rs559588111 Chr.4: 42111180-42111180 on Build GRCh38
rs1030561521 Chr.4: 42111087-42111087 on Build GRCh38
rs747647162 Chr.4: 42111049-42111049 on Build GRCh38
rs919128848 Chr.4: 42110906-42110906 on Build GRCh38
rs143248234 Chr.4: 42111066-42111066 on Build GRCh38
rs575251402 Chr.4: 42111079-42111079 on Build GRCh38
rs938666660 Chr.4: 42111155-42111155 on Build GRCh38
rs541815247 Chr.4: 42111052-42111052 on Build GRCh38
rs951141159 Chr.4: 42111059-42111059 on Build GRCh38
rs771735976 Chr.4: 42110811-42110811 on Build GRCh38

View Full Product Details