Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46036322-46036848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG1
Entrez Gene ID
2565
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Gene Aliases
-
UniGene
Hs.375051
Gene Chromosome Location
Chr.4: 46035769-46124065 on Build GRCh38
Transcripts
NM_173536.3 XM_017007990.1

SNP Detail

SNP ID Chromosome Location
rs1021649901 Chr.4: 46036495-46036495 on Build GRCh38
rs371067021 Chr.4: 46036379-46036379 on Build GRCh38
rs182127955 Chr.4: 46036628-46036628 on Build GRCh38
rs954244768 Chr.4: 46036512-46036512 on Build GRCh38
rs1000557429 Chr.4: 46036630-46036630 on Build GRCh38
rs562645378 Chr.4: 46036786-46036786 on Build GRCh38
rs984385008 Chr.4: 46036649-46036649 on Build GRCh38
rs890222943 Chr.4: 46036546-46036546 on Build GRCh38
rs978684320 Chr.4: 46036375-46036375 on Build GRCh38
rs767802603 Chr.4: 46036698-46036698 on Build GRCh38
rs751986390 Chr.4: 46036361-46036361 on Build GRCh38
rs1020326451 Chr.4: 46036564-46036565 on Build GRCh38
rs1047394562 Chr.4: 46036540-46036540 on Build GRCh38
rs1003151504 Chr.4: 46036461-46036461 on Build GRCh38
rs929070415 Chr.4: 46036637-46036637 on Build GRCh38
rs917337138 Chr.4: 46036405-46036405 on Build GRCh38
rs891911921 Chr.4: 46036470-46036470 on Build GRCh38
rs1047447880 Chr.4: 46036773-46036773 on Build GRCh38
rs544075618 Chr.4: 46036775-46036775 on Build GRCh38
rs531020035 Chr.4: 46036660-46036660 on Build GRCh38
rs1038903591 Chr.4: 46036741-46036741 on Build GRCh38
rs966033042 Chr.4: 46036805-46036805 on Build GRCh38
rs993180507 Chr.4: 46036536-46036536 on Build GRCh38
rs750915911 Chr.4: 46036617-46036617 on Build GRCh38
rs551349614 Chr.4: 46036547-46036550 on Build GRCh38
rs117266754 Chr.4: 46036693-46036693 on Build GRCh38
rs1000037817 Chr.4: 46036788-46036788 on Build GRCh38
rs908882889 Chr.4: 46036657-46036657 on Build GRCh38
rs950106865 Chr.4: 46036413-46036413 on Build GRCh38
rs563774281 Chr.4: 46036488-46036488 on Build GRCh38
rs890108143 Chr.4: 46036774-46036774 on Build GRCh38
rs907030457 Chr.4: 46036460-46036460 on Build GRCh38
rs917222635 Chr.4: 46036694-46036694 on Build GRCh38
rs1038795165 Chr.4: 46036776-46036776 on Build GRCh38
rs1007336918 Chr.4: 46036798-46036798 on Build GRCh38
rs145361126 Chr.4: 46036726-46036726 on Build GRCh38
rs185936551 Chr.4: 46036806-46036806 on Build GRCh38
rs1011561846 Chr.4: 46036487-46036487 on Build GRCh38
rs918853928 Chr.4: 46036752-46036752 on Build GRCh38
rs12374299 Chr.4: 46036728-46036728 on Build GRCh38
rs941634330 Chr.4: 46036463-46036463 on Build GRCh38
rs948427269 Chr.4: 46036668-46036668 on Build GRCh38
rs755407394 Chr.4: 46036578-46036578 on Build GRCh38
rs189862671 Chr.4: 46036518-46036518 on Build GRCh38
rs766482419 Chr.4: 46036820-46036820 on Build GRCh38
rs988897421 Chr.4: 46036402-46036402 on Build GRCh38
rs1019846021 Chr.4: 46036800-46036800 on Build GRCh38
rs900374634 Chr.4: 46036787-46036787 on Build GRCh38
rs867852704 Chr.4: 46036404-46036404 on Build GRCh38
rs956113674 Chr.4: 46036688-46036688 on Build GRCh38
rs965567436 Chr.4: 46036606-46036606 on Build GRCh38
rs906011435 Chr.4: 46036438-46036438 on Build GRCh38
rs554213559 Chr.4: 46036591-46036591 on Build GRCh38
rs796823278 Chr.4: 46036366-46036366 on Build GRCh38
rs965006732 Chr.4: 46036370-46036370 on Build GRCh38
rs896966875 Chr.4: 46036573-46036573 on Build GRCh38
rs1017421017 Chr.4: 46036365-46036365 on Build GRCh38
rs1044559380 Chr.4: 46036434-46036434 on Build GRCh38
rs941534134 Chr.4: 46036730-46036730 on Build GRCh38
rs796671990 Chr.4: 46036399-46036399 on Build GRCh38
rs1032928285 Chr.4: 46036680-46036680 on Build GRCh38
rs1017473457 Chr.4: 46036626-46036626 on Build GRCh38
rs1038607859 Chr.4: 46036474-46036474 on Build GRCh38
rs930183051 Chr.4: 46036762-46036762 on Build GRCh38

View Full Product Details