Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108812055-108812567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL25A1
Entrez Gene ID
84570
Gene Name
collagen type XXV alpha 1
Gene Aliases
AMY, CFEOM5, CLAC, CLAC-P, CLACP
UniGene
Hs.658842
Gene Chromosome Location
Chr.4: 108808725-109302737 on Build GRCh38
Transcripts
NM_198721.3 NR_045756.2 XM_011532333.2 XM_011532334.2 XM_011532335.2 XM_011532338.2 XM_011532355.2 XM_011532356.2 XM_011532358.2 XM_017008735.1 XM_017008736.1 XM_017008737.1

SNP Detail

SNP ID Chromosome Location
rs373463930 Chr.4: 108812314-108812317 on Build GRCh38
rs1010365534 Chr.4: 108812407-108812407 on Build GRCh38
rs768008704 Chr.4: 108812448-108812449 on Build GRCh38
rs992838751 Chr.4: 108812514-108812514 on Build GRCh38
rs759082531 Chr.4: 108812448-108812448 on Build GRCh38
rs75520549 Chr.4: 108812430-108812430 on Build GRCh38
rs951504604 Chr.4: 108812536-108812536 on Build GRCh38
rs942933959 Chr.4: 108812157-108812157 on Build GRCh38
rs555327040 Chr.4: 108812460-108812460 on Build GRCh38
rs747150865 Chr.4: 108812381-108812381 on Build GRCh38
rs1015135521 Chr.4: 108812470-108812470 on Build GRCh38
rs552654655 Chr.4: 108812533-108812533 on Build GRCh38
rs1002509333 Chr.4: 108812312-108812315 on Build GRCh38
rs569741324 Chr.4: 108812413-108812413 on Build GRCh38
rs539917571 Chr.4: 108812090-108812090 on Build GRCh38
rs900185268 Chr.4: 108812199-108812199 on Build GRCh38
rs866169176 Chr.4: 108812465-108812465 on Build GRCh38
rs1034216371 Chr.4: 108812456-108812456 on Build GRCh38
rs903101988 Chr.4: 108812277-108812277 on Build GRCh38
rs551433885 Chr.4: 108812225-108812225 on Build GRCh38
rs918702196 Chr.4: 108812534-108812534 on Build GRCh38
rs570891790 Chr.4: 108812078-108812078 on Build GRCh38
rs149447255 Chr.4: 108812412-108812412 on Build GRCh38
rs1039957398 Chr.4: 108812192-108812192 on Build GRCh38
rs997173049 Chr.4: 108812203-108812203 on Build GRCh38
rs962613181 Chr.4: 108812504-108812504 on Build GRCh38
rs1035777253 Chr.4: 108812316-108812316 on Build GRCh38
rs1030519895 Chr.4: 108812211-108812211 on Build GRCh38
rs773749103 Chr.4: 108812418-108812418 on Build GRCh38
rs891534313 Chr.4: 108812338-108812338 on Build GRCh38
rs532918866 Chr.4: 108812103-108812103 on Build GRCh38
rs960053398 Chr.4: 108812459-108812459 on Build GRCh38

View Full Product Details