Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 109760384-109760896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFI
Entrez Gene ID
3426
Gene Name
complement factor I
Gene Aliases
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF
UniGene
Hs.312485
Gene Chromosome Location
Chr.4: 109731221-109802225 on Build GRCh38
Transcripts
NM_000204.4 NM_001318057.1 XM_005262976.1 XM_006714209.1 XM_006714210.3 XM_011531920.1 XM_017008164.1 XM_017008165.1 XM_017008166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4328713 c.767G>A p.C256Y Chr.4: 109760528-109760528 on Build GRCh38
3940696 c.673G>A p.D225N Chr.4: 109760622-109760622 on Build GRCh38
234535 c.678C>T p.F226F Chr.4: 109760617-109760617 on Build GRCh38
3916942 c.714G>A p.M238I Chr.4: 109760581-109760581 on Build GRCh38
6272163 c.772+6C>A p.? Chr.4: 109760517-109760517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200095323 Chr.4: 109760564-109760564 on Build GRCh38
rs771896276 Chr.4: 109760600-109760600 on Build GRCh38
rs190001845 Chr.4: 109760625-109760625 on Build GRCh38
rs376590059 Chr.4: 109760486-109760486 on Build GRCh38
rs746106072 Chr.4: 109760406-109760406 on Build GRCh38
rs146444258 Chr.4: 109760576-109760576 on Build GRCh38
rs968849744 Chr.4: 109760707-109760707 on Build GRCh38
rs1039232123 Chr.4: 109760871-109760871 on Build GRCh38
rs34676321 Chr.4: 109760598-109760599 on Build GRCh38
rs887541371 Chr.4: 109760490-109760490 on Build GRCh38
rs111403504 Chr.4: 109760759-109760759 on Build GRCh38
rs768453639 Chr.4: 109760786-109760786 on Build GRCh38
rs750561676 Chr.4: 109760557-109760557 on Build GRCh38
rs952228361 Chr.4: 109760441-109760441 on Build GRCh38
rs1056454412 Chr.4: 109760726-109760726 on Build GRCh38
rs764182605 Chr.4: 109760531-109760531 on Build GRCh38
rs775537507 Chr.4: 109760548-109760548 on Build GRCh38
rs753572959 Chr.4: 109760830-109760830 on Build GRCh38
rs992073161 Chr.4: 109760783-109760783 on Build GRCh38
rs377528991 Chr.4: 109760633-109760633 on Build GRCh38
rs1006269350 Chr.4: 109760493-109760493 on Build GRCh38
rs749140698 Chr.4: 109760566-109760566 on Build GRCh38
rs758703842 Chr.4: 109760418-109760418 on Build GRCh38
rs373891906 Chr.4: 109760590-109760590 on Build GRCh38
rs773811629 Chr.4: 109760506-109760506 on Build GRCh38
rs1056423109 Chr.4: 109760436-109760436 on Build GRCh38
rs759244323 Chr.4: 109760507-109760507 on Build GRCh38
rs897855515 Chr.4: 109760741-109760742 on Build GRCh38
rs763953082 Chr.4: 109760638-109760638 on Build GRCh38
rs144145014 Chr.4: 109760599-109760599 on Build GRCh38
rs753946575 Chr.4: 109760547-109760547 on Build GRCh38
rs770616029 Chr.4: 109760500-109760500 on Build GRCh38
rs941778606 Chr.4: 109760850-109760850 on Build GRCh38
rs769192209 Chr.4: 109760429-109760429 on Build GRCh38
rs929088133 Chr.4: 109760792-109760792 on Build GRCh38
rs976423733 Chr.4: 109760474-109760474 on Build GRCh38
rs200881799 Chr.4: 109760535-109760535 on Build GRCh38
rs769739581 Chr.4: 109760514-109760514 on Build GRCh38
rs565632838 Chr.4: 109760838-109760838 on Build GRCh38
rs766156625 Chr.4: 109760824-109760824 on Build GRCh38
rs760518975 Chr.4: 109760796-109760796 on Build GRCh38
rs748808959 Chr.4: 109760499-109760499 on Build GRCh38
rs4382037 Chr.4: 109760462-109760462 on Build GRCh38
rs374153717 Chr.4: 109760422-109760422 on Build GRCh38
rs35933786 Chr.4: 109760588-109760589 on Build GRCh38
rs559804742 Chr.4: 109760756-109760756 on Build GRCh38
rs747475434 Chr.4: 109760424-109760424 on Build GRCh38
rs939294354 Chr.4: 109760714-109760714 on Build GRCh38
rs370795796 Chr.4: 109760592-109760592 on Build GRCh38
rs144048103 Chr.4: 109760569-109760569 on Build GRCh38
rs927302190 Chr.4: 109760758-109760758 on Build GRCh38
rs761185925 Chr.4: 109760746-109760746 on Build GRCh38
rs1021635917 Chr.4: 109760695-109760695 on Build GRCh38
rs758650703 Chr.4: 109760560-109760560 on Build GRCh38
rs775409716 Chr.4: 109760612-109760612 on Build GRCh38
rs980617230 Chr.4: 109760729-109760729 on Build GRCh38
rs121964916 Chr.4: 109760567-109760567 on Build GRCh38
rs199688124 Chr.4: 109760523-109760523 on Build GRCh38
rs775462739 Chr.4: 109760522-109760522 on Build GRCh38
rs767376435 Chr.4: 109760515-109760515 on Build GRCh38
rs201442703 Chr.4: 109760565-109760565 on Build GRCh38
rs937924790 Chr.4: 109760415-109760415 on Build GRCh38
rs146560961 Chr.4: 109760727-109760727 on Build GRCh38
rs896311655 Chr.4: 109760440-109760440 on Build GRCh38
rs1017833829 Chr.4: 109760448-109760448 on Build GRCh38
rs746016932 Chr.4: 109760452-109760452 on Build GRCh38
rs776373631 Chr.4: 109760670-109760670 on Build GRCh38
rs986300200 Chr.4: 109760446-109760446 on Build GRCh38
rs1007701868 Chr.4: 109760855-109760855 on Build GRCh38
rs77218589 Chr.4: 109760538-109760538 on Build GRCh38
rs370378650 Chr.4: 109760671-109760671 on Build GRCh38
rs765371768 Chr.4: 109760553-109760553 on Build GRCh38
rs765006760 Chr.4: 109760414-109760414 on Build GRCh38

View Full Product Details