Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174677184-174677720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRA3
Entrez Gene ID
8001
Gene Name
glycine receptor alpha 3
Gene Aliases
-
UniGene
Hs.413099
Gene Chromosome Location
Chr.4: 174642011-174829568 on Build GRCh38
Transcripts
NM_001042543.2 NM_006529.3 XM_011532267.2 XM_011532268.2 XM_017008630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6220841 c.817G>A p.V273I Chr.4: 174677188-174677188 on Build GRCh38
6671734 c.751C>T p.R251* Chr.4: 174677254-174677254 on Build GRCh38
7137178 c.723G>A p.T241T Chr.4: 174677282-174677282 on Build GRCh38
5615396 c.802G>T p.V268F Chr.4: 174677203-174677203 on Build GRCh38
733090 c.812C>A p.S271Y Chr.4: 174677193-174677193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768917914 Chr.4: 174677299-174677299 on Build GRCh38
rs774589195 Chr.4: 174677268-174677268 on Build GRCh38
rs781100845 Chr.4: 174677271-174677271 on Build GRCh38
rs1006466614 Chr.4: 174677346-174677346 on Build GRCh38
rs749253586 Chr.4: 174677283-174677283 on Build GRCh38
rs959573979 Chr.4: 174677354-174677354 on Build GRCh38
rs773539362 Chr.4: 174677327-174677327 on Build GRCh38
rs564086334 Chr.4: 174677345-174677345 on Build GRCh38
rs865795166 Chr.4: 174677253-174677253 on Build GRCh38
rs548629373 Chr.4: 174677650-174677650 on Build GRCh38
rs1040160475 Chr.4: 174677521-174677521 on Build GRCh38
rs1018797499 Chr.4: 174677368-174677368 on Build GRCh38
rs116414284 Chr.4: 174677282-174677282 on Build GRCh38
rs541882435 Chr.4: 174677222-174677222 on Build GRCh38
rs776968068 Chr.4: 174677301-174677301 on Build GRCh38
rs990087437 Chr.4: 174677486-174677486 on Build GRCh38
rs1054180974 Chr.4: 174677541-174677541 on Build GRCh38
rs915532128 Chr.4: 174677545-174677545 on Build GRCh38
rs200026813 Chr.4: 174677278-174677278 on Build GRCh38
rs867988051 Chr.4: 174677683-174677683 on Build GRCh38
rs1006599462 Chr.4: 174677688-174677688 on Build GRCh38
rs567437148 Chr.4: 174677655-174677655 on Build GRCh38
rs201620617 Chr.4: 174677269-174677269 on Build GRCh38
rs78196152 Chr.4: 174677377-174677377 on Build GRCh38
rs115009641 Chr.4: 174677446-174677446 on Build GRCh38
rs779270533 Chr.4: 174677526-174677527 on Build GRCh38
rs971117745 Chr.4: 174677423-174677423 on Build GRCh38
rs752656878 Chr.4: 174677622-174677622 on Build GRCh38
rs759891940 Chr.4: 174677302-174677302 on Build GRCh38
rs775660210 Chr.4: 174677291-174677291 on Build GRCh38
rs774668313 Chr.4: 174677425-174677425 on Build GRCh38
rs762287278 Chr.4: 174677210-174677210 on Build GRCh38
rs771987128 Chr.4: 174677288-174677288 on Build GRCh38
rs992296240 Chr.4: 174677368-174677368 on Build GRCh38
rs765591766 Chr.4: 174677308-174677308 on Build GRCh38
rs749930503 Chr.4: 174677254-174677254 on Build GRCh38
rs149146668 Chr.4: 174677277-174677277 on Build GRCh38
rs79980656 Chr.4: 174677303-174677303 on Build GRCh38
rs901961249 Chr.4: 174677580-174677580 on Build GRCh38
rs951591480 Chr.4: 174677440-174677440 on Build GRCh38
rs910130254 Chr.4: 174677500-174677500 on Build GRCh38
rs924547206 Chr.4: 174677239-174677239 on Build GRCh38
rs765792225 Chr.4: 174677241-174677241 on Build GRCh38
rs949846781 Chr.4: 174677579-174677579 on Build GRCh38
rs917018626 Chr.4: 174677384-174677384 on Build GRCh38
rs181593623 Chr.4: 174677344-174677344 on Build GRCh38
rs1034305940 Chr.4: 174677347-174677347 on Build GRCh38
rs964146892 Chr.4: 174677386-174677386 on Build GRCh38
rs910427869 Chr.4: 174677490-174677490 on Build GRCh38
rs905664183 Chr.4: 174677409-174677409 on Build GRCh38
rs759631125 Chr.4: 174677439-174677439 on Build GRCh38
rs6848286 Chr.4: 174677520-174677520 on Build GRCh38
rs1026257475 Chr.4: 174677339-174677339 on Build GRCh38
rs561852492 Chr.4: 174677404-174677404 on Build GRCh38
rs940974153 Chr.4: 174677267-174677267 on Build GRCh38
rs924291478 Chr.4: 174677445-174677445 on Build GRCh38
rs541969992 Chr.4: 174677385-174677385 on Build GRCh38
rs993480249 Chr.4: 174677309-174677309 on Build GRCh38
rs760828688 Chr.4: 174677656-174677656 on Build GRCh38
rs533049169 Chr.4: 174677349-174677349 on Build GRCh38
rs552163628 Chr.4: 174677405-174677405 on Build GRCh38

View Full Product Details