Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 177440123-177440633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
UniGene
Hs.207776
Gene Chromosome Location
Chr.4: 177430774-177442503 on Build GRCh38
Transcripts
NM_000027.3 NM_001171988.1 NR_033655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428359 c.250G>T p.E84* Chr.4: 177440304-177440304 on Build GRCh38
222968 c.146C>T p.S49F Chr.4: 177440408-177440408 on Build GRCh38
3974818 c.233G>A p.S78N Chr.4: 177440321-177440321 on Build GRCh38
7209451 c.201G>A p.E67E Chr.4: 177440353-177440353 on Build GRCh38
3602216 c.215C>T p.S72F Chr.4: 177440339-177440339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765186052 Chr.4: 177440338-177440338 on Build GRCh38
rs748002607 Chr.4: 177440316-177440316 on Build GRCh38
rs907791659 Chr.4: 177440480-177440480 on Build GRCh38
rs367922948 Chr.4: 177440444-177440444 on Build GRCh38
rs375077753 Chr.4: 177440232-177440232 on Build GRCh38
rs386833419 Chr.4: 177440362-177440362 on Build GRCh38
rs770392499 Chr.4: 177440475-177440475 on Build GRCh38
rs554359822 Chr.4: 177440385-177440385 on Build GRCh38
rs984748851 Chr.4: 177440176-177440177 on Build GRCh38
rs776196445 Chr.4: 177440304-177440304 on Build GRCh38
rs751165674 Chr.4: 177440603-177440603 on Build GRCh38
rs746506859 Chr.4: 177440280-177440280 on Build GRCh38
rs143210437 Chr.4: 177440404-177440404 on Build GRCh38
rs999695070 Chr.4: 177440335-177440335 on Build GRCh38
rs373209495 Chr.4: 177440350-177440350 on Build GRCh38
rs150912713 Chr.4: 177440278-177440278 on Build GRCh38
rs757523971 Chr.4: 177440262-177440262 on Build GRCh38
rs75639723 Chr.4: 177440577-177440577 on Build GRCh38
rs373224954 Chr.4: 177440558-177440558 on Build GRCh38
rs1054743624 Chr.4: 177440426-177440426 on Build GRCh38
rs200195689 Chr.4: 177440371-177440371 on Build GRCh38
rs760862994 Chr.4: 177440468-177440468 on Build GRCh38
rs747800631 Chr.4: 177440431-177440431 on Build GRCh38
rs772979226 Chr.4: 177440241-177440241 on Build GRCh38
rs372881453 Chr.4: 177440250-177440250 on Build GRCh38
rs761536667 Chr.4: 177440238-177440238 on Build GRCh38
rs780842670 Chr.4: 177440425-177440425 on Build GRCh38
rs121964907 Chr.4: 177440375-177440375 on Build GRCh38
rs757480668 Chr.4: 177440377-177440377 on Build GRCh38
rs747908061 Chr.4: 177440466-177440466 on Build GRCh38
rs746939020 Chr.4: 177440462-177440467 on Build GRCh38
rs750795098 Chr.4: 177440383-177440383 on Build GRCh38
rs531988617 Chr.4: 177440555-177440555 on Build GRCh38
rs757065230 Chr.4: 177440330-177440330 on Build GRCh38
rs74448573 Chr.4: 177440219-177440219 on Build GRCh38
rs1040247749 Chr.4: 177440258-177440258 on Build GRCh38
rs139812241 Chr.4: 177440291-177440291 on Build GRCh38
rs949735021 Chr.4: 177440512-177440512 on Build GRCh38
rs944664746 Chr.4: 177440504-177440504 on Build GRCh38
rs769505733 Chr.4: 177440465-177440466 on Build GRCh38
rs762870187 Chr.4: 177440344-177440344 on Build GRCh38
rs533715399 Chr.4: 177440157-177440157 on Build GRCh38
rs199523988 Chr.4: 177440242-177440242 on Build GRCh38
rs936963111 Chr.4: 177440214-177440214 on Build GRCh38
rs750634899 Chr.4: 177440376-177440377 on Build GRCh38
rs769452319 Chr.4: 177440440-177440440 on Build GRCh38
rs780480789 Chr.4: 177440393-177440393 on Build GRCh38
rs768772316 Chr.4: 177440465-177440465 on Build GRCh38
rs746411193 Chr.4: 177440269-177440269 on Build GRCh38
rs562442577 Chr.4: 177440595-177440595 on Build GRCh38
rs796922197 Chr.4: 177440466-177440467 on Build GRCh38
rs765934592 Chr.4: 177440357-177440357 on Build GRCh38
rs992715539 Chr.4: 177440394-177440394 on Build GRCh38
rs201174810 Chr.4: 177440465-177440465 on Build GRCh38
rs769423815 Chr.4: 177440334-177440334 on Build GRCh38
rs564681166 Chr.4: 177440469-177440469 on Build GRCh38
rs754066653 Chr.4: 177440374-177440374 on Build GRCh38
rs910142863 Chr.4: 177440176-177440176 on Build GRCh38
rs751071150 Chr.4: 177440395-177440395 on Build GRCh38
rs754176418 Chr.4: 177440305-177440306 on Build GRCh38
rs3792607 Chr.4: 177440467-177440467 on Build GRCh38
rs758531447 Chr.4: 177440455-177440456 on Build GRCh38
rs537759103 Chr.4: 177440240-177440240 on Build GRCh38
rs768292136 Chr.4: 177440224-177440224 on Build GRCh38
rs758886928 Chr.4: 177440387-177440387 on Build GRCh38
rs760773119 Chr.4: 177440366-177440366 on Build GRCh38
rs34241758 Chr.4: 177440260-177440260 on Build GRCh38
rs779157725 Chr.4: 177440353-177440356 on Build GRCh38
rs752824210 Chr.4: 177440251-177440251 on Build GRCh38
rs376346895 Chr.4: 177440255-177440255 on Build GRCh38
rs1041415731 Chr.4: 177440524-177440524 on Build GRCh38
rs121964909 Chr.4: 177440340-177440340 on Build GRCh38
rs958977553 Chr.4: 177440488-177440488 on Build GRCh38
rs992178203 Chr.4: 177440500-177440500 on Build GRCh38
rs560547845 Chr.4: 177440379-177440379 on Build GRCh38
rs762639905 Chr.4: 177440467-177440468 on Build GRCh38
rs758907500 Chr.4: 177440274-177440274 on Build GRCh38
rs764053416 Chr.4: 177440275-177440275 on Build GRCh38
rs386833420 Chr.4: 177440353-177440354 on Build GRCh38
rs754261269 Chr.4: 177440261-177440261 on Build GRCh38
rs750098973 Chr.4: 177440396-177440396 on Build GRCh38
rs749057212 Chr.4: 177440462-177440462 on Build GRCh38
rs558350852 Chr.4: 177440165-177440165 on Build GRCh38
rs113432122 Chr.4: 177440466-177440467 on Build GRCh38
rs903884327 Chr.4: 177440306-177440306 on Build GRCh38
rs1040597958 Chr.4: 177440565-177440565 on Build GRCh38

View Full Product Details