Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186194268-186194786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3131077 c.251A>G p.Y84C Chr.4: 186194536-186194536 on Build GRCh38
5866148 c.281T>C p.V94A Chr.4: 186194566-186194566 on Build GRCh38
5819202 c.311A>C p.N104T Chr.4: 186194596-186194596 on Build GRCh38
5435723 c.304C>T p.L102F Chr.4: 186194589-186194589 on Build GRCh38
6099779 c.242C>G p.T81R Chr.4: 186194527-186194527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919817600 Chr.4: 186194435-186194435 on Build GRCh38
rs780201946 Chr.4: 186194608-186194608 on Build GRCh38
rs143272248 Chr.4: 186194587-186194587 on Build GRCh38
rs751065501 Chr.4: 186194581-186194581 on Build GRCh38
rs199476182 Chr.4: 186194613-186194613 on Build GRCh38
rs755855410 Chr.4: 186194597-186194597 on Build GRCh38
rs747109853 Chr.4: 186194610-186194610 on Build GRCh38
rs1047455760 Chr.4: 186194391-186194391 on Build GRCh38
rs559133074 Chr.4: 186194539-186194539 on Build GRCh38
rs781047997 Chr.4: 186194520-186194520 on Build GRCh38
rs917747804 Chr.4: 186194366-186194366 on Build GRCh38
rs777576473 Chr.4: 186194598-186194598 on Build GRCh38
rs959342530 Chr.4: 186194319-186194319 on Build GRCh38
rs138717663 Chr.4: 186194639-186194639 on Build GRCh38
rs757146398 Chr.4: 186194573-186194574 on Build GRCh38
rs576168140 Chr.4: 186194367-186194367 on Build GRCh38
rs775367222 Chr.4: 186194653-186194653 on Build GRCh38
rs763283418 Chr.4: 186194642-186194642 on Build GRCh38
rs62350517 Chr.4: 186194623-186194623 on Build GRCh38
rs189095281 Chr.4: 186194337-186194337 on Build GRCh38
rs192920673 Chr.4: 186194555-186194555 on Build GRCh38
rs992040327 Chr.4: 186194325-186194325 on Build GRCh38
rs564082243 Chr.4: 186194440-186194440 on Build GRCh38
rs113384324 Chr.4: 186194751-186194751 on Build GRCh38
rs10013653 Chr.4: 186194478-186194478 on Build GRCh38
rs770459499 Chr.4: 186194631-186194631 on Build GRCh38
rs911209750 Chr.4: 186194632-186194632 on Build GRCh38
rs886059281 Chr.4: 186194596-186194596 on Build GRCh38
rs142775374 Chr.4: 186194567-186194567 on Build GRCh38
rs72646245 Chr.4: 186194697-186194697 on Build GRCh38
rs746112010 Chr.4: 186194528-186194528 on Build GRCh38
rs199476186 Chr.4: 186194538-186194538 on Build GRCh38
rs770610205 Chr.4: 186194645-186194645 on Build GRCh38
rs1031102027 Chr.4: 186194517-186194519 on Build GRCh38
rs372144743 Chr.4: 186194494-186194494 on Build GRCh38
rs138173968 Chr.4: 186194525-186194525 on Build GRCh38
rs773881089 Chr.4: 186194646-186194646 on Build GRCh38
rs764551849 Chr.4: 186194456-186194456 on Build GRCh38
rs759033778 Chr.4: 186194585-186194585 on Build GRCh38
rs937588379 Chr.4: 186194757-186194757 on Build GRCh38
rs201610304 Chr.4: 186194549-186194549 on Build GRCh38
rs199476187 Chr.4: 186194568-186194568 on Build GRCh38
rs751069999 Chr.4: 186194502-186194502 on Build GRCh38
rs368058295 Chr.4: 186194479-186194479 on Build GRCh38
rs377042245 Chr.4: 186194548-186194548 on Build GRCh38
rs972611604 Chr.4: 186194427-186194427 on Build GRCh38
rs752486517 Chr.4: 186194521-186194521 on Build GRCh38
rs890686622 Chr.4: 186194713-186194713 on Build GRCh38
rs754919490 Chr.4: 186194504-186194504 on Build GRCh38
rs767122178 Chr.4: 186194470-186194470 on Build GRCh38
rs1000040559 Chr.4: 186194510-186194510 on Build GRCh38
rs199476185 Chr.4: 186194522-186194522 on Build GRCh38
rs765859742 Chr.4: 186194546-186194546 on Build GRCh38
rs998797061 Chr.4: 186194345-186194357 on Build GRCh38
rs7682918 Chr.4: 186194687-186194687 on Build GRCh38
rs72646244 Chr.4: 186194651-186194651 on Build GRCh38
rs772955821 Chr.4: 186194564-186194564 on Build GRCh38
rs542832725 Chr.4: 186194722-186194722 on Build GRCh38
rs768629035 Chr.4: 186194621-186194621 on Build GRCh38
rs932249242 Chr.4: 186194688-186194688 on Build GRCh38
rs764071280 Chr.4: 186194660-186194660 on Build GRCh38
rs753751184 Chr.4: 186194546-186194548 on Build GRCh38
rs185497433 Chr.4: 186194627-186194627 on Build GRCh38
rs528660295 Chr.4: 186194717-186194717 on Build GRCh38
rs904780354 Chr.4: 186194754-186194754 on Build GRCh38
rs745684760 Chr.4: 186194648-186194648 on Build GRCh38
rs534643986 Chr.4: 186194599-186194599 on Build GRCh38
rs759029388 Chr.4: 186194648-186194648 on Build GRCh38
rs996427635 Chr.4: 186194472-186194472 on Build GRCh38
rs777116249 Chr.4: 186194462-186194462 on Build GRCh38
rs1050153147 Chr.4: 186194471-186194471 on Build GRCh38
rs1041271685 Chr.4: 186194488-186194488 on Build GRCh38
rs1051998298 Chr.4: 186194695-186194695 on Build GRCh38
rs758628871 Chr.4: 186194602-186194602 on Build GRCh38
rs201905182 Chr.4: 186194658-186194658 on Build GRCh38
rs770301714 Chr.4: 186194557-186194557 on Build GRCh38
rs961208459 Chr.4: 186194574-186194574 on Build GRCh38
rs773690118 Chr.4: 186194640-186194640 on Build GRCh38
rs1037409917 Chr.4: 186194670-186194670 on Build GRCh38
rs752645954 Chr.4: 186194595-186194595 on Build GRCh38

View Full Product Details