Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 98863197-98863746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1
Entrez Gene ID
1105
Gene Name
chromodomain helicase DNA binding protein 1
Gene Aliases
-
UniGene
Hs.643465
Gene Chromosome Location
Chr.5: 98855204-98929014 on Build GRCh38
Transcripts
NM_001270.2 XM_005271866.4 XM_005271867.4 XM_011543112.2 XM_017008991.1 XM_017008992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634390 c.4358A>T p.K1453I Chr.5: 98863477-98863477 on Build GRCh38
3697356 c.4378G>C p.E1460Q Chr.5: 98863457-98863457 on Build GRCh38
6627804 c.4371T>C p.H1457H Chr.5: 98863464-98863464 on Build GRCh38
1439339 c.4259G>T p.R1420I Chr.5: 98863576-98863576 on Build GRCh38
6470613 c.4299G>T p.R1433S Chr.5: 98863536-98863536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142476918 Chr.5: 98863272-98863272 on Build GRCh38
rs376028777 Chr.5: 98863449-98863449 on Build GRCh38
rs759148663 Chr.5: 98863451-98863451 on Build GRCh38
rs557050743 Chr.5: 98863222-98863222 on Build GRCh38
rs1046265171 Chr.5: 98863339-98863339 on Build GRCh38
rs866864356 Chr.5: 98863276-98863276 on Build GRCh38
rs369338956 Chr.5: 98863509-98863509 on Build GRCh38
rs1050758860 Chr.5: 98863709-98863709 on Build GRCh38
rs745759312 Chr.5: 98863633-98863633 on Build GRCh38
rs941590818 Chr.5: 98863646-98863646 on Build GRCh38
rs943568515 Chr.5: 98863609-98863609 on Build GRCh38
rs1050812535 Chr.5: 98863258-98863258 on Build GRCh38
rs772248298 Chr.5: 98863698-98863698 on Build GRCh38
rs923538584 Chr.5: 98863678-98863678 on Build GRCh38
rs77326655 Chr.5: 98863357-98863357 on Build GRCh38
rs774178705 Chr.5: 98863628-98863638 on Build GRCh38
rs766810280 Chr.5: 98863436-98863436 on Build GRCh38
rs761746662 Chr.5: 98863633-98863633 on Build GRCh38
rs535838826 Chr.5: 98863331-98863331 on Build GRCh38
rs773447369 Chr.5: 98863392-98863392 on Build GRCh38
rs974985709 Chr.5: 98863669-98863669 on Build GRCh38
rs146876564 Chr.5: 98863690-98863690 on Build GRCh38
rs924329545 Chr.5: 98863711-98863711 on Build GRCh38
rs1012793761 Chr.5: 98863347-98863348 on Build GRCh38
rs762002945 Chr.5: 98863372-98863372 on Build GRCh38
rs162139 Chr.5: 98863719-98863719 on Build GRCh38
rs576976052 Chr.5: 98863662-98863662 on Build GRCh38
rs933680693 Chr.5: 98863239-98863239 on Build GRCh38
rs928013036 Chr.5: 98863426-98863426 on Build GRCh38
rs759848686 Chr.5: 98863658-98863658 on Build GRCh38
rs118187541 Chr.5: 98863349-98863349 on Build GRCh38
rs1017483904 Chr.5: 98863226-98863226 on Build GRCh38
rs763716752 Chr.5: 98863397-98863398 on Build GRCh38
rs899509950 Chr.5: 98863234-98863238 on Build GRCh38
rs35183210 Chr.5: 98863345-98863345 on Build GRCh38
rs796917408 Chr.5: 98863357-98863357 on Build GRCh38
rs1022290059 Chr.5: 98863319-98863319 on Build GRCh38
rs978184481 Chr.5: 98863379-98863379 on Build GRCh38
rs762675109 Chr.5: 98863391-98863391 on Build GRCh38
rs909387418 Chr.5: 98863570-98863570 on Build GRCh38
rs780992886 Chr.5: 98863621-98863621 on Build GRCh38
rs10069735 Chr.5: 98863373-98863373 on Build GRCh38
rs757014470 Chr.5: 98863612-98863612 on Build GRCh38
rs751296619 Chr.5: 98863554-98863554 on Build GRCh38
rs1022869897 Chr.5: 98863347-98863347 on Build GRCh38
rs161941 Chr.5: 98863500-98863500 on Build GRCh38
rs556246355 Chr.5: 98863348-98863348 on Build GRCh38
rs752444165 Chr.5: 98863463-98863463 on Build GRCh38
rs764789985 Chr.5: 98863453-98863453 on Build GRCh38
rs774582041 Chr.5: 98863368-98863368 on Build GRCh38
rs1032861321 Chr.5: 98863319-98863320 on Build GRCh38
rs754305052 Chr.5: 98863448-98863448 on Build GRCh38
rs964559666 Chr.5: 98863266-98863266 on Build GRCh38
rs893004366 Chr.5: 98863332-98863332 on Build GRCh38
rs999063837 Chr.5: 98863284-98863284 on Build GRCh38
rs202018159 Chr.5: 98863344-98863344 on Build GRCh38
rs113061054 Chr.5: 98863358-98863359 on Build GRCh38

View Full Product Details