Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127046484-127046992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
UniGene
Hs.49573
Gene Chromosome Location
Chr.5: 127040752-127073539 on Build GRCh38
Transcripts
NM_001164479.1

SNP Detail

SNP ID Chromosome Location
rs1037526293 Chr.5: 127046511-127046511 on Build GRCh38
rs1032550985 Chr.5: 127046673-127046673 on Build GRCh38
rs571348381 Chr.5: 127046550-127046550 on Build GRCh38
rs6595747 Chr.5: 127046829-127046829 on Build GRCh38
rs1032174585 Chr.5: 127046770-127046770 on Build GRCh38
rs757294406 Chr.5: 127046686-127046686 on Build GRCh38
rs931761021 Chr.5: 127046523-127046523 on Build GRCh38
rs1037487875 Chr.5: 127046964-127046964 on Build GRCh38
rs1038529570 Chr.5: 127046624-127046624 on Build GRCh38
rs754935219 Chr.5: 127046729-127046729 on Build GRCh38
rs941756711 Chr.5: 127046962-127046962 on Build GRCh38
rs986254059 Chr.5: 127046812-127046812 on Build GRCh38
rs920387419 Chr.5: 127046788-127046788 on Build GRCh38
rs886202600 Chr.5: 127046576-127046576 on Build GRCh38
rs750299135 Chr.5: 127046712-127046712 on Build GRCh38
rs907562851 Chr.5: 127046868-127046868 on Build GRCh38
rs1008203527 Chr.5: 127046718-127046718 on Build GRCh38
rs925145987 Chr.5: 127046510-127046510 on Build GRCh38
rs1012433104 Chr.5: 127046931-127046931 on Build GRCh38
rs973205673 Chr.5: 127046772-127046772 on Build GRCh38
rs971111645 Chr.5: 127046688-127046688 on Build GRCh38
rs528148079 Chr.5: 127046547-127046547 on Build GRCh38
rs911671177 Chr.5: 127046824-127046824 on Build GRCh38
rs940509840 Chr.5: 127046509-127046509 on Build GRCh38
rs902767970 Chr.5: 127046858-127046858 on Build GRCh38
rs1028998766 Chr.5: 127046679-127046679 on Build GRCh38
rs779073316 Chr.5: 127046521-127046521 on Build GRCh38
rs931840594 Chr.5: 127046801-127046801 on Build GRCh38
rs898697682 Chr.5: 127046631-127046631 on Build GRCh38
rs990719011 Chr.5: 127046781-127046781 on Build GRCh38
rs917582862 Chr.5: 127046540-127046540 on Build GRCh38
rs566529478 Chr.5: 127046855-127046855 on Build GRCh38
rs937923297 Chr.5: 127046519-127046519 on Build GRCh38
rs186843211 Chr.5: 127046896-127046896 on Build GRCh38
rs781106911 Chr.5: 127046709-127046709 on Build GRCh38
rs575727435 Chr.5: 127046953-127046953 on Build GRCh38
rs59500308 Chr.5: 127046549-127046549 on Build GRCh38
rs748536795 Chr.5: 127046609-127046610 on Build GRCh38
rs893938605 Chr.5: 127046890-127046890 on Build GRCh38
rs559049275 Chr.5: 127046952-127046952 on Build GRCh38
rs755784066 Chr.5: 127046728-127046728 on Build GRCh38
rs1050183727 Chr.5: 127046537-127046537 on Build GRCh38
rs1012713157 Chr.5: 127046680-127046680 on Build GRCh38
rs966754266 Chr.5: 127046733-127046733 on Build GRCh38
rs745935550 Chr.5: 127046589-127046589 on Build GRCh38
rs190416132 Chr.5: 127046689-127046689 on Build GRCh38
rs559627960 Chr.5: 127046882-127046882 on Build GRCh38
rs959502270 Chr.5: 127046778-127046778 on Build GRCh38
rs149086704 Chr.5: 127046623-127046623 on Build GRCh38
rs181602995 Chr.5: 127046761-127046761 on Build GRCh38
rs961969593 Chr.5: 127046734-127046734 on Build GRCh38
rs552906804 Chr.5: 127046832-127046832 on Build GRCh38
rs997099758 Chr.5: 127046648-127046648 on Build GRCh38
rs979510649 Chr.5: 127046769-127046769 on Build GRCh38

View Full Product Details