Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132076000-132076520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2
Entrez Gene ID
1437
Gene Name
colony stimulating factor 2
Gene Aliases
GMCSF
UniGene
Hs.1349
Gene Chromosome Location
Chr.5: 132073792-132076170 on Build GRCh38
Transcripts
NM_000758.3

SNP Detail

SNP ID Chromosome Location
rs967535212 Chr.5: 132076250-132076250 on Build GRCh38
rs970151998 Chr.5: 132076379-132076379 on Build GRCh38
rs1050372817 Chr.5: 132076342-132076342 on Build GRCh38
rs2069631 Chr.5: 132076499-132076499 on Build GRCh38
rs552081079 Chr.5: 132076369-132076369 on Build GRCh38
rs568046823 Chr.5: 132076478-132076478 on Build GRCh38
rs761737216 Chr.5: 132076029-132076029 on Build GRCh38
rs1053680387 Chr.5: 132076251-132076251 on Build GRCh38
rs780298705 Chr.5: 132076346-132076346 on Build GRCh38
rs151158919 Chr.5: 132076082-132076082 on Build GRCh38
rs996607194 Chr.5: 132076340-132076340 on Build GRCh38
rs900965167 Chr.5: 132076326-132076326 on Build GRCh38
rs566950612 Chr.5: 132076294-132076294 on Build GRCh38
rs150342985 Chr.5: 132076416-132076416 on Build GRCh38
rs374188903 Chr.5: 132076165-132076165 on Build GRCh38
rs140299486 Chr.5: 132076376-132076376 on Build GRCh38
rs926457767 Chr.5: 132076228-132076228 on Build GRCh38
rs774380302 Chr.5: 132076337-132076337 on Build GRCh38
rs1014232822 Chr.5: 132076370-132076370 on Build GRCh38
rs892793056 Chr.5: 132076218-132076218 on Build GRCh38
rs930159810 Chr.5: 132076081-132076081 on Build GRCh38
rs543138441 Chr.5: 132076080-132076083 on Build GRCh38
rs547052960 Chr.5: 132076270-132076270 on Build GRCh38
rs1040726557 Chr.5: 132076500-132076500 on Build GRCh38
rs917301919 Chr.5: 132076136-132076136 on Build GRCh38
rs926460469 Chr.5: 132076401-132076401 on Build GRCh38
rs985991504 Chr.5: 132076285-132076285 on Build GRCh38
rs909163421 Chr.5: 132076286-132076286 on Build GRCh38
rs869057099 Chr.5: 132076081-132076083 on Build GRCh38
rs796166785 Chr.5: 132076473-132076473 on Build GRCh38
rs13306292 Chr.5: 132076104-132076104 on Build GRCh38
rs25883 Chr.5: 132076239-132076239 on Build GRCh38
rs1015831858 Chr.5: 132076350-132076350 on Build GRCh38
rs984372689 Chr.5: 132076079-132076079 on Build GRCh38
rs1055446232 Chr.5: 132076142-132076142 on Build GRCh38
rs913786941 Chr.5: 132076484-132076484 on Build GRCh38
rs665253 Chr.5: 132076030-132076030 on Build GRCh38
rs981050750 Chr.5: 132076176-132076176 on Build GRCh38
rs544892992 Chr.5: 132076137-132076137 on Build GRCh38
rs111691590 Chr.5: 132076264-132076264 on Build GRCh38
rs953117638 Chr.5: 132076075-132076075 on Build GRCh38
rs992933671 Chr.5: 132076117-132076117 on Build GRCh38
rs961341094 Chr.5: 132076114-132076114 on Build GRCh38
rs1040780530 Chr.5: 132076322-132076322 on Build GRCh38
rs1016041510 Chr.5: 132076099-132076099 on Build GRCh38
rs76944607 Chr.5: 132076475-132076475 on Build GRCh38
rs530134693 Chr.5: 132076269-132076269 on Build GRCh38
rs374482921 Chr.5: 132076331-132076331 on Build GRCh38
rs980195606 Chr.5: 132076380-132076380 on Build GRCh38
rs962680127 Chr.5: 132076302-132076302 on Build GRCh38
rs1030590214 Chr.5: 132076259-132076259 on Build GRCh38
rs1006231606 Chr.5: 132076349-132076349 on Build GRCh38
rs930059457 Chr.5: 132076344-132076344 on Build GRCh38
rs532688480 Chr.5: 132076295-132076295 on Build GRCh38
rs937517297 Chr.5: 132076366-132076366 on Build GRCh38

View Full Product Details