Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137939447-137939956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13B
Entrez Gene ID
51306
Gene Name
family with sequence similarity 13 member B
Gene Aliases
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61
UniGene
Hs.567453
Gene Chromosome Location
Chr.5: 137937909-138051961 on Build GRCh38
Transcripts
NM_001101800.2 NM_001101801.2 NM_016603.3 XM_005272007.1 XM_006714647.1 XM_006714648.1 XM_006714649.1 XM_011543448.1 XM_011543449.1 XM_011543450.1 XM_017009549.1 XM_017009550.1 XM_017009551.1 NM_001258448.1 NM_001258449.1 NM_001300921.1 NM_014386.3 XM_017009343.1

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
UniGene
Hs.716884
Gene Chromosome Location
Chr.5: 137889420-137942747 on Build GRCh38
Transcripts
NM_001101800.2 NM_001101801.2 NM_016603.3 XM_005272007.1 XM_006714647.1 XM_006714648.1 XM_006714649.1 XM_011543448.1 XM_011543449.1 XM_011543450.1 XM_017009549.1 XM_017009550.1 XM_017009551.1 NM_001258448.1 NM_001258449.1 NM_001300921.1 NM_014386.3 XM_017009343.1

SNP Detail

SNP ID Chromosome Location
rs961524272 Chr.5: 137939501-137939501 on Build GRCh38
rs755554402 Chr.5: 137939623-137939623 on Build GRCh38
rs994809870 Chr.5: 137939757-137939757 on Build GRCh38
rs956898847 Chr.5: 137939849-137939849 on Build GRCh38
rs531280171 Chr.5: 137939729-137939729 on Build GRCh38
rs145927578 Chr.5: 137939527-137939527 on Build GRCh38
rs915442062 Chr.5: 137939561-137939561 on Build GRCh38
rs56124297 Chr.5: 137939701-137939701 on Build GRCh38
rs900271878 Chr.5: 137939605-137939605 on Build GRCh38
rs551828739 Chr.5: 137939595-137939595 on Build GRCh38
rs761932584 Chr.5: 137939839-137939839 on Build GRCh38
rs1005973683 Chr.5: 137939802-137939802 on Build GRCh38
rs567853011 Chr.5: 137939877-137939877 on Build GRCh38
rs951883982 Chr.5: 137939801-137939801 on Build GRCh38
rs191748428 Chr.5: 137939876-137939876 on Build GRCh38
rs187412806 Chr.5: 137939726-137939726 on Build GRCh38
rs139187964 Chr.5: 137939552-137939552 on Build GRCh38
rs925461629 Chr.5: 137939663-137939663 on Build GRCh38
rs1141538 Chr.5: 137939510-137939510 on Build GRCh38
rs555642739 Chr.5: 137939540-137939540 on Build GRCh38
rs1023565552 Chr.5: 137939758-137939758 on Build GRCh38
rs143662048 Chr.5: 137939518-137939518 on Build GRCh38
rs1042504673 Chr.5: 137939647-137939647 on Build GRCh38
rs890076936 Chr.5: 137939714-137939714 on Build GRCh38
rs945177289 Chr.5: 137939604-137939604 on Build GRCh38
rs546678274 Chr.5: 137939819-137939819 on Build GRCh38
rs917569108 Chr.5: 137939598-137939598 on Build GRCh38
rs926501408 Chr.5: 137939542-137939542 on Build GRCh38
rs372478250 Chr.5: 137939806-137939806 on Build GRCh38
rs994276885 Chr.5: 137939760-137939760 on Build GRCh38
rs899041552 Chr.5: 137939750-137939750 on Build GRCh38
rs866374269 Chr.5: 137939702-137939702 on Build GRCh38
rs544347055 Chr.5: 137939559-137939559 on Build GRCh38
rs564114888 Chr.5: 137939792-137939792 on Build GRCh38
rs1041075917 Chr.5: 137939735-137939735 on Build GRCh38
rs545898880 Chr.5: 137939741-137939741 on Build GRCh38
rs947081384 Chr.5: 137939567-137939567 on Build GRCh38
rs931701394 Chr.5: 137939622-137939622 on Build GRCh38
rs959304605 Chr.5: 137939566-137939566 on Build GRCh38
rs7711607 Chr.5: 137939569-137939569 on Build GRCh38

View Full Product Details