Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154819541-154820042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAXDC2
Entrez Gene ID
10826
Gene Name
fatty acid hydroxylase domain containing 2
Gene Aliases
C5orf4
UniGene
Hs.519694
Gene Chromosome Location
Chr.5: 154817018-154850685 on Build GRCh38
Transcripts
NM_032385.4 XM_005268358.2 XM_006714753.2 XM_017008965.1 XM_017008966.1

SNP Detail

SNP ID Chromosome Location
rs910830987 Chr.5: 154819892-154819892 on Build GRCh38
rs958911437 Chr.5: 154819783-154819783 on Build GRCh38
rs1020477300 Chr.5: 154819746-154819746 on Build GRCh38
rs965424332 Chr.5: 154819722-154819722 on Build GRCh38
rs926144593 Chr.5: 154819780-154819780 on Build GRCh38
rs142689940 Chr.5: 154819771-154819771 on Build GRCh38
rs919363036 Chr.5: 154819923-154819923 on Build GRCh38
rs929042510 Chr.5: 154819946-154819947 on Build GRCh38
rs991711649 Chr.5: 154819785-154819785 on Build GRCh38
rs779785397 Chr.5: 154819748-154819749 on Build GRCh38
rs751762882 Chr.5: 154819747-154819748 on Build GRCh38
rs942237382 Chr.5: 154819893-154819893 on Build GRCh38
rs950130914 Chr.5: 154819660-154819660 on Build GRCh38
rs895546646 Chr.5: 154819962-154819962 on Build GRCh38
rs976526945 Chr.5: 154819742-154819742 on Build GRCh38
rs889361080 Chr.5: 154819657-154819657 on Build GRCh38
rs888282322 Chr.5: 154819975-154819975 on Build GRCh38
rs897597264 Chr.5: 154819684-154819684 on Build GRCh38
rs188470842 Chr.5: 154819773-154819773 on Build GRCh38
rs750878798 Chr.5: 154819746-154819747 on Build GRCh38
rs551008992 Chr.5: 154819997-154819997 on Build GRCh38
rs879792493 Chr.5: 154819647-154819647 on Build GRCh38
rs1050741439 Chr.5: 154819654-154819654 on Build GRCh38
rs923433089 Chr.5: 154819639-154819639 on Build GRCh38
rs1048234603 Chr.5: 154819965-154819965 on Build GRCh38
rs138007742 Chr.5: 154819685-154819685 on Build GRCh38
rs146036621 Chr.5: 154819935-154819935 on Build GRCh38
rs954738780 Chr.5: 154819925-154819925 on Build GRCh38
rs923349897 Chr.5: 154819805-154819805 on Build GRCh38
rs1006777599 Chr.5: 154820021-154820021 on Build GRCh38
rs1026888602 Chr.5: 154819686-154819686 on Build GRCh38
rs191325699 Chr.5: 154819572-154819572 on Build GRCh38
rs77632402 Chr.5: 154819579-154819579 on Build GRCh38
rs950131353 Chr.5: 154819842-154819842 on Build GRCh38
rs1006830073 Chr.5: 154819691-154819691 on Build GRCh38
rs1000845013 Chr.5: 154819766-154819766 on Build GRCh38
rs895825857 Chr.5: 154819609-154819609 on Build GRCh38
rs933573702 Chr.5: 154819646-154819649 on Build GRCh38
rs896983468 Chr.5: 154819662-154819662 on Build GRCh38
rs1036428231 Chr.5: 154819847-154819847 on Build GRCh38
rs1022915223 Chr.5: 154819739-154819739 on Build GRCh38
rs375780063 Chr.5: 154819994-154819994 on Build GRCh38
rs571566535 Chr.5: 154819933-154819933 on Build GRCh38
rs561820223 Chr.5: 154819606-154819606 on Build GRCh38
rs1044320 Chr.5: 154819698-154819698 on Build GRCh38
rs772551378 Chr.5: 154819924-154819924 on Build GRCh38
rs1036889752 Chr.5: 154819661-154819661 on Build GRCh38
rs1055700984 Chr.5: 154819589-154819589 on Build GRCh38
rs373349264 Chr.5: 154819951-154819952 on Build GRCh38
rs183140243 Chr.5: 154819703-154819703 on Build GRCh38
rs988206586 Chr.5: 154819777-154819777 on Build GRCh38
rs1056921096 Chr.5: 154819961-154819961 on Build GRCh38
rs888333405 Chr.5: 154819688-154819688 on Build GRCh38
rs39614 Chr.5: 154819808-154819808 on Build GRCh38

View Full Product Details