Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154874899-154875419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT8
Entrez Gene ID
9337
Gene Name
CCR4-NOT transcription complex subunit 8
Gene Aliases
CAF1, CALIF, Caf1b, POP2, hCAF1
UniGene
Hs.26703
Gene Chromosome Location
Chr.5: 154857952-154876792 on Build GRCh38
Transcripts
NM_001301073.1 NM_001301074.1 NM_001301075.1 NM_001301077.1 NM_001301080.1 NM_001301082.1 NM_001301083.1 NM_001301086.1 NM_004779.5 XM_005268527.2 XM_011537706.1 XM_011537708.2 XM_017010048.1 XM_017010049.1 XM_017010050.1 XM_017010051.1 XM_017010052.1 XM_017010053.1 XM_017010054.1 XM_017010055.1 XM_017010056.1 XM_017010057.1 XM_017010058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5996402 c.772C>T p.R258W Chr.5: 154875332-154875332 on Build GRCh38
3715028 c.832C>T p.Q278* Chr.5: 154875392-154875392 on Build GRCh38
6418013 c.802C>A p.Q268K Chr.5: 154875362-154875362 on Build GRCh38
3612907 c.760A>T p.K254* Chr.5: 154875320-154875320 on Build GRCh38
449169 c.800C>T p.A267V Chr.5: 154875360-154875360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746402158 Chr.5: 154875227-154875227 on Build GRCh38
rs762369247 Chr.5: 154875291-154875321 on Build GRCh38
rs969835440 Chr.5: 154875001-154875001 on Build GRCh38
rs960834550 Chr.5: 154875054-154875054 on Build GRCh38
rs947370577 Chr.5: 154874960-154874960 on Build GRCh38
rs892701829 Chr.5: 154874989-154874989 on Build GRCh38
rs557001678 Chr.5: 154874947-154874947 on Build GRCh38
rs781347799 Chr.5: 154875137-154875137 on Build GRCh38
rs1015345065 Chr.5: 154875101-154875101 on Build GRCh38
rs559373413 Chr.5: 154875142-154875142 on Build GRCh38
rs1028576706 Chr.5: 154875050-154875050 on Build GRCh38
rs775284433 Chr.5: 154875367-154875367 on Build GRCh38
rs973351112 Chr.5: 154875085-154875085 on Build GRCh38
rs763773990 Chr.5: 154875387-154875387 on Build GRCh38
rs780483114 Chr.5: 154875246-154875246 on Build GRCh38
rs746699293 Chr.5: 154875163-154875163 on Build GRCh38
rs1052571201 Chr.5: 154874980-154874980 on Build GRCh38
rs151108307 Chr.5: 154875065-154875065 on Build GRCh38
rs944496357 Chr.5: 154875175-154875175 on Build GRCh38
rs1050110621 Chr.5: 154875117-154875117 on Build GRCh38
rs961392731 Chr.5: 154875118-154875118 on Build GRCh38
rs760574723 Chr.5: 154875380-154875380 on Build GRCh38
rs1014757637 Chr.5: 154875383-154875383 on Build GRCh38
rs1005596976 Chr.5: 154875093-154875093 on Build GRCh38
rs1041583952 Chr.5: 154875005-154875006 on Build GRCh38
rs776973664 Chr.5: 154875317-154875317 on Build GRCh38
rs1043179885 Chr.5: 154874965-154874965 on Build GRCh38
rs757638675 Chr.5: 154875089-154875089 on Build GRCh38
rs1011143895 Chr.5: 154874992-154874992 on Build GRCh38
rs772590860 Chr.5: 154875240-154875240 on Build GRCh38
rs770097487 Chr.5: 154875321-154875321 on Build GRCh38
rs1022247334 Chr.5: 154874998-154874998 on Build GRCh38
rs182413256 Chr.5: 154874941-154874941 on Build GRCh38
rs199732823 Chr.5: 154875326-154875326 on Build GRCh38
rs545223549 Chr.5: 154875185-154875185 on Build GRCh38
rs748771303 Chr.5: 154875263-154875263 on Build GRCh38
rs903229736 Chr.5: 154874966-154874966 on Build GRCh38
rs748283320 Chr.5: 154875318-154875318 on Build GRCh38
rs992635542 Chr.5: 154875060-154875060 on Build GRCh38
rs931740957 Chr.5: 154874948-154874948 on Build GRCh38
rs374156365 Chr.5: 154875343-154875343 on Build GRCh38
rs768900977 Chr.5: 154875270-154875270 on Build GRCh38
rs1035479297 Chr.5: 154875028-154875028 on Build GRCh38
rs751929260 Chr.5: 154875013-154875013 on Build GRCh38
rs79250798 Chr.5: 154875393-154875393 on Build GRCh38
rs1043918289 Chr.5: 154875213-154875213 on Build GRCh38
rs17116534 Chr.5: 154875258-154875258 on Build GRCh38
rs1050638778 Chr.5: 154874951-154874951 on Build GRCh38
rs564924597 Chr.5: 154875196-154875196 on Build GRCh38
rs558498005 Chr.5: 154874979-154874979 on Build GRCh38
rs890220764 Chr.5: 154874973-154874973 on Build GRCh38
rs34488733 Chr.5: 154875358-154875358 on Build GRCh38
rs997125661 Chr.5: 154875038-154875038 on Build GRCh38
rs911608697 Chr.5: 154875121-154875121 on Build GRCh38
rs1011114125 Chr.5: 154875212-154875212 on Build GRCh38
rs528153396 Chr.5: 154875164-154875164 on Build GRCh38
rs72799503 Chr.5: 154875237-154875237 on Build GRCh38
rs931600584 Chr.5: 154875104-154875104 on Build GRCh38
rs145445405 Chr.5: 154875290-154875290 on Build GRCh38
rs542763519 Chr.5: 154875077-154875077 on Build GRCh38

View Full Product Details