Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180569029-180569539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6
Entrez Gene ID
57472
Gene Name
CCR4-NOT transcription complex subunit 6
Gene Aliases
CCR4, Ccr4a
UniGene
Hs.608380
Gene Chromosome Location
Chr.5: 180494373-180578405 on Build GRCh38
Transcripts
NM_001303241.1 XM_005265953.1 XM_011534605.2 XM_011534606.1 XM_011534607.1 XM_017009670.1 XM_017009671.1 XM_017009672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3827872 c.1191T>C p.S397S Chr.5: 180569273-180569273 on Build GRCh38
6267859 c.1111G>T p.V371L Chr.5: 180569193-180569193 on Build GRCh38
3827871 c.1172C>T p.S391F Chr.5: 180569254-180569254 on Build GRCh38
137446 c.1217C>T p.P406L Chr.5: 180569299-180569299 on Build GRCh38
3827870 c.1066C>G p.L356V Chr.5: 180569148-180569148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767306600 Chr.5: 180569099-180569099 on Build GRCh38
rs371184209 Chr.5: 180569142-180569142 on Build GRCh38
rs773360090 Chr.5: 180569080-180569080 on Build GRCh38
rs762519194 Chr.5: 180569383-180569383 on Build GRCh38
rs778107557 Chr.5: 180569138-180569138 on Build GRCh38
rs187963423 Chr.5: 180569366-180569366 on Build GRCh38
rs55999117 Chr.5: 180569381-180569381 on Build GRCh38
rs533238023 Chr.5: 180569137-180569137 on Build GRCh38
rs748666197 Chr.5: 180569078-180569078 on Build GRCh38
rs775686360 Chr.5: 180569353-180569353 on Build GRCh38
rs973730126 Chr.5: 180569124-180569124 on Build GRCh38
rs1042503572 Chr.5: 180569204-180569204 on Build GRCh38
rs765040951 Chr.5: 180569122-180569122 on Build GRCh38
rs763858007 Chr.5: 180569477-180569477 on Build GRCh38
rs190601258 Chr.5: 180569390-180569390 on Build GRCh38
rs564142808 Chr.5: 180569052-180569052 on Build GRCh38
rs375570858 Chr.5: 180569209-180569209 on Build GRCh38
rs774351366 Chr.5: 180569162-180569162 on Build GRCh38
rs752194167 Chr.5: 180569357-180569358 on Build GRCh38
rs760686713 Chr.5: 180569104-180569104 on Build GRCh38
rs769127884 Chr.5: 180569068-180569068 on Build GRCh38
rs202095631 Chr.5: 180569171-180569171 on Build GRCh38
rs759071538 Chr.5: 180569257-180569257 on Build GRCh38
rs781711611 Chr.5: 180569129-180569129 on Build GRCh38
rs763915557 Chr.5: 180569107-180569107 on Build GRCh38
rs778987910 Chr.5: 180569309-180569309 on Build GRCh38
rs982572213 Chr.5: 180569103-180569103 on Build GRCh38
rs757724982 Chr.5: 180569280-180569280 on Build GRCh38
rs920931889 Chr.5: 180569508-180569508 on Build GRCh38
rs570052121 Chr.5: 180569238-180569238 on Build GRCh38
rs777627856 Chr.5: 180569237-180569237 on Build GRCh38
rs910806809 Chr.5: 180569391-180569391 on Build GRCh38
rs746124196 Chr.5: 180569168-180569168 on Build GRCh38
rs148283671 Chr.5: 180569513-180569513 on Build GRCh38
rs897926612 Chr.5: 180569062-180569062 on Build GRCh38
rs761686926 Chr.5: 180569373-180569373 on Build GRCh38
rs776419781 Chr.5: 180569367-180569367 on Build GRCh38
rs753141951 Chr.5: 180569256-180569256 on Build GRCh38
rs745926565 Chr.5: 180569311-180569311 on Build GRCh38
rs1012738610 Chr.5: 180569055-180569055 on Build GRCh38
rs6877400 Chr.5: 180569111-180569111 on Build GRCh38
rs973784055 Chr.5: 180569507-180569507 on Build GRCh38
rs747444018 Chr.5: 180569067-180569067 on Build GRCh38
rs764158508 Chr.5: 180569211-180569211 on Build GRCh38
rs750111420 Chr.5: 180569125-180569125 on Build GRCh38
rs764854349 Chr.5: 180569106-180569106 on Build GRCh38
rs752449846 Chr.5: 180569102-180569102 on Build GRCh38
rs138378919 Chr.5: 180569272-180569272 on Build GRCh38
rs372697922 Chr.5: 180569059-180569059 on Build GRCh38
rs749446928 Chr.5: 180569159-180569159 on Build GRCh38
rs150124040 Chr.5: 180569127-180569127 on Build GRCh38
rs971269445 Chr.5: 180569423-180569423 on Build GRCh38
rs962768237 Chr.5: 180569497-180569497 on Build GRCh38
rs976370950 Chr.5: 180569350-180569350 on Build GRCh38
rs749766287 Chr.5: 180569278-180569278 on Build GRCh38
rs377314143 Chr.5: 180569131-180569131 on Build GRCh38
rs906428251 Chr.5: 180569105-180569105 on Build GRCh38
rs746878182 Chr.5: 180569356-180569356 on Build GRCh38
rs771255691 Chr.5: 180569088-180569088 on Build GRCh38
rs865882494 Chr.5: 180569387-180569387 on Build GRCh38
rs376346658 Chr.5: 180569222-180569222 on Build GRCh38
rs760356809 Chr.5: 180569186-180569186 on Build GRCh38
rs183280345 Chr.5: 180569454-180569454 on Build GRCh38
rs774547243 Chr.5: 180569092-180569092 on Build GRCh38
rs761749899 Chr.5: 180569225-180569225 on Build GRCh38
rs756716934 Chr.5: 180569114-180569114 on Build GRCh38
rs781640968 Chr.5: 180569069-180569069 on Build GRCh38
rs1051306968 Chr.5: 180569153-180569153 on Build GRCh38
rs942168147 Chr.5: 180569414-180569420 on Build GRCh38
rs762660575 Chr.5: 180569372-180569372 on Build GRCh38
rs772322116 Chr.5: 180569341-180569341 on Build GRCh38
rs764842807 Chr.5: 180569234-180569234 on Build GRCh38
rs762947510 Chr.5: 180569426-180569426 on Build GRCh38
rs770157776 Chr.5: 180569079-180569079 on Build GRCh38
rs775316079 Chr.5: 180569180-180569180 on Build GRCh38
rs762915501 Chr.5: 180569084-180569084 on Build GRCh38
rs373260577 Chr.5: 180569093-180569093 on Build GRCh38
rs748410104 Chr.5: 180569132-180569132 on Build GRCh38

View Full Product Details