Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180601488-180601987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009268.1

SNP Detail

SNP ID Chromosome Location
rs906653735 Chr.5: 180601801-180601801 on Build GRCh38
rs948143714 Chr.5: 180601649-180601649 on Build GRCh38
rs80139920 Chr.5: 180601727-180601727 on Build GRCh38
rs1036971726 Chr.5: 180601818-180601818 on Build GRCh38
rs765510643 Chr.5: 180601903-180601903 on Build GRCh38
rs915764657 Chr.5: 180601776-180601776 on Build GRCh38
rs557423997 Chr.5: 180601833-180601833 on Build GRCh38
rs56268625 Chr.5: 180601677-180601677 on Build GRCh38
rs1007683402 Chr.5: 180601841-180601841 on Build GRCh38
rs928533293 Chr.5: 180601761-180601761 on Build GRCh38
rs939532402 Chr.5: 180601806-180601806 on Build GRCh38
rs911749993 Chr.5: 180601565-180601565 on Build GRCh38
rs72816988 Chr.5: 180601509-180601509 on Build GRCh38
rs985912465 Chr.5: 180601557-180601557 on Build GRCh38
rs994453083 Chr.5: 180601685-180601685 on Build GRCh38
rs1031945911 Chr.5: 180601871-180601871 on Build GRCh38
rs574191911 Chr.5: 180601870-180601870 on Build GRCh38
rs919443418 Chr.5: 180601958-180601958 on Build GRCh38
rs968316575 Chr.5: 180601530-180601530 on Build GRCh38
rs989462861 Chr.5: 180601773-180601773 on Build GRCh38
rs893882388 Chr.5: 180601639-180601639 on Build GRCh38
rs995070641 Chr.5: 180601826-180601826 on Build GRCh38
rs557704134 Chr.5: 180601767-180601767 on Build GRCh38
rs888791189 Chr.5: 180601837-180601837 on Build GRCh38
rs911285724 Chr.5: 180601601-180601601 on Build GRCh38
rs915210773 Chr.5: 180601879-180601879 on Build GRCh38
rs865923572 Chr.5: 180601850-180601850 on Build GRCh38
rs986979864 Chr.5: 180601595-180601595 on Build GRCh38
rs768149177 Chr.5: 180601593-180601593 on Build GRCh38
rs867831718 Chr.5: 180601877-180601877 on Build GRCh38
rs891369738 Chr.5: 180601821-180601821 on Build GRCh38
rs541004213 Chr.5: 180601526-180601526 on Build GRCh38
rs915834303 Chr.5: 180601945-180601945 on Build GRCh38
rs432475 Chr.5: 180601589-180601589 on Build GRCh38
rs935319494 Chr.5: 180601622-180601622 on Build GRCh38
rs929905953 Chr.5: 180601635-180601635 on Build GRCh38
rs959941332 Chr.5: 180601939-180601939 on Build GRCh38
rs760338631 Chr.5: 180601744-180601744 on Build GRCh38
rs995695419 Chr.5: 180601887-180601887 on Build GRCh38
rs991854616 Chr.5: 180601943-180601943 on Build GRCh38
rs1008380623 Chr.5: 180601722-180601722 on Build GRCh38
rs537109594 Chr.5: 180601797-180601797 on Build GRCh38
rs369176626 Chr.5: 180601657-180601657 on Build GRCh38
rs898067403 Chr.5: 180601819-180601819 on Build GRCh38
rs57918841 Chr.5: 180601830-180601830 on Build GRCh38
rs915264808 Chr.5: 180601777-180601777 on Build GRCh38
rs548957957 Chr.5: 180601633-180601633 on Build GRCh38
rs141679676 Chr.5: 180601733-180601733 on Build GRCh38
rs1042722327 Chr.5: 180601688-180601690 on Build GRCh38
rs1031172596 Chr.5: 180601567-180601567 on Build GRCh38
rs759006343 Chr.5: 180601938-180601938 on Build GRCh38
rs71613474 Chr.5: 180601824-180601824 on Build GRCh38
rs1036511584 Chr.5: 180601950-180601950 on Build GRCh38
rs116376016 Chr.5: 180601792-180601792 on Build GRCh38
rs894207979 Chr.5: 180601669-180601669 on Build GRCh38
rs307822 Chr.5: 180601717-180601717 on Build GRCh38
rs1027303647 Chr.5: 180601702-180601702 on Build GRCh38
rs770342782 Chr.5: 180601715-180601715 on Build GRCh38
rs1018885801 Chr.5: 180601723-180601723 on Build GRCh38
rs879541932 Chr.5: 180601621-180601621 on Build GRCh38
rs387288 Chr.5: 180601577-180601577 on Build GRCh38
rs1015179607 Chr.5: 180601659-180601659 on Build GRCh38
rs977269917 Chr.5: 180601861-180601861 on Build GRCh38
rs1030361470 Chr.5: 180601704-180601713 on Build GRCh38
rs533021723 Chr.5: 180601656-180601656 on Build GRCh38
rs781337350 Chr.5: 180601635-180601637 on Build GRCh38
rs2279622 Chr.5: 180601743-180601743 on Build GRCh38
rs1027457995 Chr.5: 180601894-180601894 on Build GRCh38
rs939458512 Chr.5: 180601949-180601949 on Build GRCh38
rs1021734629 Chr.5: 180601527-180601527 on Build GRCh38
rs969378908 Chr.5: 180601916-180601916 on Build GRCh38
rs944594789 Chr.5: 180601574-180601574 on Build GRCh38
rs34937445 Chr.5: 180601612-180601613 on Build GRCh38
rs990080316 Chr.5: 180601876-180601876 on Build GRCh38
rs573248368 Chr.5: 180601826-180601826 on Build GRCh38
rs977699891 Chr.5: 180601741-180601741 on Build GRCh38
rs567761156 Chr.5: 180601673-180601673 on Build GRCh38
rs560078393 Chr.5: 180601508-180601508 on Build GRCh38
rs966223416 Chr.5: 180601853-180601853 on Build GRCh38
rs564190217 Chr.5: 180601586-180601586 on Build GRCh38
rs957252000 Chr.5: 180601875-180601875 on Build GRCh38
rs1045161974 Chr.5: 180601793-180601793 on Build GRCh38
rs981219038 Chr.5: 180601926-180601926 on Build GRCh38
rs1045619446 Chr.5: 180601653-180601653 on Build GRCh38
rs565625241 Chr.5: 180601734-180601734 on Build GRCh38
rs1053821414 Chr.5: 180601637-180601637 on Build GRCh38
rs386930 Chr.5: 180601699-180601699 on Build GRCh38
rs902887865 Chr.5: 180601696-180601696 on Build GRCh38
rs999813369 Chr.5: 180601550-180601551 on Build GRCh38

View Full Product Details