Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181059529-181060036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_017009113.1 XM_017009114.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3854441 c.1343G>A p.R448Q Chr.5: 181059594-181059594 on Build GRCh38
1634176 c.1343G>T p.R448L Chr.5: 181059597-181059597 on Build GRCh38
4893258 c.1527G>A p.G509G Chr.5: 181059781-181059781 on Build GRCh38
1620183 c.1286A>T p.E429V Chr.5: 181059540-181059540 on Build GRCh38
7204883 c.1460G>A p.R487K Chr.5: 181059714-181059714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773154157 Chr.5: 181059890-181059891 on Build GRCh38
rs752376469 Chr.5: 181059595-181059595 on Build GRCh38
rs749970809 Chr.5: 181059756-181059756 on Build GRCh38
rs201962783 Chr.5: 181059581-181059581 on Build GRCh38
rs773217170 Chr.5: 181059627-181059627 on Build GRCh38
rs10068763 Chr.5: 181059785-181059785 on Build GRCh38
rs772162101 Chr.5: 181059722-181059722 on Build GRCh38
rs773882190 Chr.5: 181059895-181059895 on Build GRCh38
rs550995366 Chr.5: 181059793-181059793 on Build GRCh38
rs149109139 Chr.5: 181059578-181059578 on Build GRCh38
rs1039652943 Chr.5: 181059973-181059973 on Build GRCh38
rs369501156 Chr.5: 181059778-181059778 on Build GRCh38
rs753334630 Chr.5: 181059679-181059679 on Build GRCh38
rs556965027 Chr.5: 181059927-181059927 on Build GRCh38
rs540764579 Chr.5: 181059570-181059570 on Build GRCh38
rs779204535 Chr.5: 181059842-181059842 on Build GRCh38
rs959116851 Chr.5: 181059828-181059828 on Build GRCh38
rs746900251 Chr.5: 181059849-181059849 on Build GRCh38
rs748525887 Chr.5: 181059697-181059697 on Build GRCh38
rs143209155 Chr.5: 181059596-181059596 on Build GRCh38
rs764049923 Chr.5: 181059728-181059728 on Build GRCh38
rs772491013 Chr.5: 181059558-181059558 on Build GRCh38
rs549176677 Chr.5: 181059639-181059639 on Build GRCh38
rs76350168 Chr.5: 181059811-181059811 on Build GRCh38
rs762827825 Chr.5: 181059583-181059583 on Build GRCh38
rs777965406 Chr.5: 181059712-181059712 on Build GRCh38
rs931332514 Chr.5: 181059981-181059981 on Build GRCh38
rs747979807 Chr.5: 181059874-181059874 on Build GRCh38
rs898541932 Chr.5: 181059749-181059749 on Build GRCh38
rs774531259 Chr.5: 181059719-181059719 on Build GRCh38
rs745753431 Chr.5: 181059720-181059720 on Build GRCh38
rs201001706 Chr.5: 181059797-181059797 on Build GRCh38
rs745978107 Chr.5: 181059779-181059779 on Build GRCh38
rs774144406 Chr.5: 181059651-181059651 on Build GRCh38
rs778161879 Chr.5: 181059771-181059771 on Build GRCh38
rs781572183 Chr.5: 181059619-181059619 on Build GRCh38
rs769542502 Chr.5: 181059875-181059875 on Build GRCh38
rs200438302 Chr.5: 181059836-181059836 on Build GRCh38
rs766239405 Chr.5: 181059584-181059584 on Build GRCh38
rs995452979 Chr.5: 181059767-181059767 on Build GRCh38
rs767185436 Chr.5: 181059656-181059656 on Build GRCh38
rs141905608 Chr.5: 181059765-181059765 on Build GRCh38
rs768014357 Chr.5: 181059762-181059762 on Build GRCh38
rs761211952 Chr.5: 181059830-181059830 on Build GRCh38
rs756491974 Chr.5: 181059571-181059571 on Build GRCh38
rs764608600 Chr.5: 181059745-181059745 on Build GRCh38
rs777382242 Chr.5: 181059597-181059597 on Build GRCh38
rs762444530 Chr.5: 181059807-181059807 on Build GRCh38
rs369136500 Chr.5: 181059553-181059553 on Build GRCh38
rs11541510 Chr.5: 181059991-181059991 on Build GRCh38
rs371740103 Chr.5: 181059562-181059562 on Build GRCh38
rs747607665 Chr.5: 181059615-181059615 on Build GRCh38
rs199990647 Chr.5: 181059658-181059658 on Build GRCh38
rs761122015 Chr.5: 181059896-181059896 on Build GRCh38
rs905663284 Chr.5: 181059706-181059706 on Build GRCh38
rs567861680 Chr.5: 181059794-181059794 on Build GRCh38
rs758884841 Chr.5: 181059605-181059605 on Build GRCh38
rs776473700 Chr.5: 181059892-181059893 on Build GRCh38
rs753181443 Chr.5: 181059822-181059822 on Build GRCh38
rs529232979 Chr.5: 181059636-181059636 on Build GRCh38
rs775371091 Chr.5: 181059723-181059723 on Build GRCh38
rs1031567209 Chr.5: 181059863-181059885 on Build GRCh38
rs887198756 Chr.5: 181059994-181059994 on Build GRCh38
rs1052776160 Chr.5: 181059667-181059667 on Build GRCh38
rs752466398 Chr.5: 181059552-181059552 on Build GRCh38
rs1002723063 Chr.5: 181059724-181059724 on Build GRCh38
rs1017820097 Chr.5: 181059810-181059810 on Build GRCh38
rs775460934 Chr.5: 181059657-181059657 on Build GRCh38
rs755536857 Chr.5: 181059695-181059695 on Build GRCh38
rs780715061 Chr.5: 181059551-181059551 on Build GRCh38
rs796629441 Chr.5: 181059734-181059734 on Build GRCh38
rs762235917 Chr.5: 181059899-181059899 on Build GRCh38
rs746350412 Chr.5: 181059555-181059555 on Build GRCh38
rs780432865 Chr.5: 181059610-181059610 on Build GRCh38
rs138779385 Chr.5: 181059655-181059655 on Build GRCh38
rs914204583 Chr.5: 181059931-181059931 on Build GRCh38
rs756706547 Chr.5: 181059683-181059683 on Build GRCh38
rs33927456 Chr.5: 181059891-181059892 on Build GRCh38
rs1036370599 Chr.5: 181059568-181059568 on Build GRCh38
rs10054109 Chr.5: 181059841-181059841 on Build GRCh38
rs543659011 Chr.5: 181059587-181059587 on Build GRCh38
rs748329786 Chr.5: 181059621-181059621 on Build GRCh38
rs771781812 Chr.5: 181059821-181059823 on Build GRCh38
rs750680841 Chr.5: 181059843-181059843 on Build GRCh38
rs557438953 Chr.5: 181059897-181059897 on Build GRCh38
rs756260354 Chr.5: 181059702-181059702 on Build GRCh38
rs754252051 Chr.5: 181059837-181059837 on Build GRCh38
rs769499166 Chr.5: 181059802-181059802 on Build GRCh38
rs770024119 Chr.5: 181059622-181059622 on Build GRCh38
rs979201957 Chr.5: 181059614-181059614 on Build GRCh38
rs768876378 Chr.5: 181059618-181059618 on Build GRCh38
rs376115758 Chr.5: 181059772-181059772 on Build GRCh38
rs577706388 Chr.5: 181059898-181059898 on Build GRCh38
rs947046105 Chr.5: 181059984-181059984 on Build GRCh38
rs749584088 Chr.5: 181059716-181059716 on Build GRCh38
rs772071794 Chr.5: 181059780-181059780 on Build GRCh38
rs923064714 Chr.5: 181059664-181059664 on Build GRCh38
rs751976307 Chr.5: 181059691-181059691 on Build GRCh38
rs747351098 Chr.5: 181059564-181059564 on Build GRCh38
rs1050926485 Chr.5: 181059673-181059673 on Build GRCh38
rs767405462 Chr.5: 181059594-181059594 on Build GRCh38
rs764848070 Chr.5: 181059689-181059689 on Build GRCh38
rs761563865 Chr.5: 181059743-181059743 on Build GRCh38
rs762644204 Chr.5: 181059888-181059888 on Build GRCh38
rs147052698 Chr.5: 181059727-181059727 on Build GRCh38
rs892832442 Chr.5: 181059690-181059690 on Build GRCh38
rs756458616 Chr.5: 181059766-181059766 on Build GRCh38
rs759248008 Chr.5: 181059591-181059591 on Build GRCh38
rs543030918 Chr.5: 181059926-181059926 on Build GRCh38
rs972102293 Chr.5: 181059577-181059577 on Build GRCh38
rs768097572 Chr.5: 181059637-181059639 on Build GRCh38
rs912433442 Chr.5: 181059919-181059919 on Build GRCh38
rs771110908 Chr.5: 181059643-181059643 on Build GRCh38
rs202184383 Chr.5: 181059833-181059833 on Build GRCh38
rs138408414 Chr.5: 181059730-181059730 on Build GRCh38
rs778469748 Chr.5: 181059664-181059669 on Build GRCh38
rs898388269 Chr.5: 181059974-181059974 on Build GRCh38
rs755381652 Chr.5: 181059757-181059757 on Build GRCh38
rs770398660 Chr.5: 181059891-181059891 on Build GRCh38
rs772905362 Chr.5: 181059886-181059886 on Build GRCh38
rs369125172 Chr.5: 181059696-181059696 on Build GRCh38
rs142809941 Chr.5: 181059718-181059718 on Build GRCh38
rs934293153 Chr.5: 181059665-181059665 on Build GRCh38
rs748292444 Chr.5: 181059986-181059986 on Build GRCh38
rs753424662 Chr.5: 181059604-181059604 on Build GRCh38
rs923674207 Chr.5: 181059645-181059645 on Build GRCh38
rs554626414 Chr.5: 181059845-181059845 on Build GRCh38
rs763855544 Chr.5: 181059661-181059661 on Build GRCh38
rs747820917 Chr.5: 181059720-181059720 on Build GRCh38
rs370306645 Chr.5: 181059872-181059872 on Build GRCh38
rs768731833 Chr.5: 181059871-181059871 on Build GRCh38

View Full Product Details