Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 1610504-1611011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXC1
Entrez Gene ID
2296
Gene Name
forkhead box C1
Gene Aliases
ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3
UniGene
Hs.348883
Gene Chromosome Location
Chr.6: 1610446-1613897 on Build GRCh38
Transcripts
NM_001453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6172980 c.534C>G p.D178E Chr.6: 1610979-1610979 on Build GRCh38
4610176 c.404G>A p.C135Y Chr.6: 1610849-1610849 on Build GRCh38
79870 c.441C>T p.G147G Chr.6: 1610886-1610886 on Build GRCh38
5869716 c.470A>G p.D157G Chr.6: 1610915-1610915 on Build GRCh38
4160349 c.327G>A p.M109I Chr.6: 1610772-1610772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748064350 Chr.6: 1610772-1610772 on Build GRCh38
rs780155404 Chr.6: 1610641-1610641 on Build GRCh38
rs141798688 Chr.6: 1610724-1610724 on Build GRCh38
rs201891353 Chr.6: 1610760-1610760 on Build GRCh38
rs1032706802 Chr.6: 1610614-1610614 on Build GRCh38
rs758053417 Chr.6: 1610615-1610615 on Build GRCh38
rs35168188 Chr.6: 1610649-1610649 on Build GRCh38
rs368260972 Chr.6: 1610637-1610637 on Build GRCh38
rs765987336 Chr.6: 1610674-1610674 on Build GRCh38
rs483352810 Chr.6: 1610822-1610822 on Build GRCh38
rs767147260 Chr.6: 1610721-1610721 on Build GRCh38
rs906356509 Chr.6: 1610792-1610792 on Build GRCh38
rs121909338 Chr.6: 1610833-1610833 on Build GRCh38
rs146275363 Chr.6: 1610775-1610775 on Build GRCh38
rs376405759 Chr.6: 1610691-1610691 on Build GRCh38
rs756248798 Chr.6: 1610858-1610858 on Build GRCh38
rs751162614 Chr.6: 1610627-1610627 on Build GRCh38
rs767127192 Chr.6: 1610805-1610805 on Build GRCh38
rs888722820 Chr.6: 1610679-1610679 on Build GRCh38
rs957987530 Chr.6: 1610864-1610864 on Build GRCh38
rs104893951 Chr.6: 1610780-1610780 on Build GRCh38
rs760058458 Chr.6: 1610904-1610904 on Build GRCh38
rs200100677 Chr.6: 1610913-1610913 on Build GRCh38
rs562467758 Chr.6: 1610844-1610844 on Build GRCh38
rs104893958 Chr.6: 1610823-1610823 on Build GRCh38
rs1050413147 Chr.6: 1610668-1610668 on Build GRCh38
rs758825480 Chr.6: 1610757-1610757 on Build GRCh38
rs746088193 Chr.6: 1610560-1610560 on Build GRCh38
rs1057519473 Chr.6: 1610713-1610713 on Build GRCh38
rs752984586 Chr.6: 1610981-1610981 on Build GRCh38
rs780403354 Chr.6: 1610827-1610827 on Build GRCh38
rs772118724 Chr.6: 1610566-1610566 on Build GRCh38
rs150084174 Chr.6: 1610640-1610640 on Build GRCh38
rs767686358 Chr.6: 1610980-1610980 on Build GRCh38
rs771450858 Chr.6: 1610543-1610543 on Build GRCh38
rs575996895 Chr.6: 1610767-1610767 on Build GRCh38
rs1037208972 Chr.6: 1610750-1610750 on Build GRCh38
rs775495187 Chr.6: 1610569-1610569 on Build GRCh38
rs372857241 Chr.6: 1610586-1610586 on Build GRCh38
rs369346224 Chr.6: 1610919-1610919 on Build GRCh38
rs1057519477 Chr.6: 1610932-1610932 on Build GRCh38
rs201072263 Chr.6: 1610766-1610766 on Build GRCh38
rs1037043441 Chr.6: 1610731-1610731 on Build GRCh38
rs749585793 Chr.6: 1610664-1610675 on Build GRCh38
rs759065232 Chr.6: 1610715-1610715 on Build GRCh38
rs1000269656 Chr.6: 1610804-1610804 on Build GRCh38
rs759264099 Chr.6: 1610801-1610801 on Build GRCh38
rs374448293 Chr.6: 1610604-1610604 on Build GRCh38
rs104893953 Chr.6: 1610690-1610690 on Build GRCh38
rs766717877 Chr.6: 1610964-1610964 on Build GRCh38
rs756088237 Chr.6: 1610982-1610982 on Build GRCh38
rs752309038 Chr.6: 1610808-1610808 on Build GRCh38
rs886044143 Chr.6: 1610735-1610736 on Build GRCh38
rs751286982 Chr.6: 1610697-1610697 on Build GRCh38
rs1057519472 Chr.6: 1610561-1610568 on Build GRCh38
rs762354349 Chr.6: 1610814-1610814 on Build GRCh38
rs778382469 Chr.6: 1610630-1610630 on Build GRCh38
rs148739656 Chr.6: 1610947-1610947 on Build GRCh38
rs548226336 Chr.6: 1610595-1610595 on Build GRCh38
rs200884695 Chr.6: 1610922-1610922 on Build GRCh38
rs772938181 Chr.6: 1610664-1610664 on Build GRCh38
rs547414473 Chr.6: 1610778-1610778 on Build GRCh38
rs1057519476 Chr.6: 1610902-1610902 on Build GRCh38
rs755160620 Chr.6: 1610979-1610979 on Build GRCh38
rs562544227 Chr.6: 1610821-1610821 on Build GRCh38
rs886041355 Chr.6: 1610901-1610901 on Build GRCh38
rs188085399 Chr.6: 1610616-1610616 on Build GRCh38
rs104893957 Chr.6: 1610837-1610837 on Build GRCh38
rs781318332 Chr.6: 1610769-1610769 on Build GRCh38
rs1057519471 Chr.6: 1610545-1610554 on Build GRCh38
rs369078747 Chr.6: 1610728-1610728 on Build GRCh38
rs35974174 Chr.6: 1610652-1610652 on Build GRCh38
rs142581779 Chr.6: 1610868-1610868 on Build GRCh38
rs774086427 Chr.6: 1610784-1610784 on Build GRCh38
rs200040370 Chr.6: 1610661-1610661 on Build GRCh38
rs751776784 Chr.6: 1610832-1610832 on Build GRCh38
rs375581157 Chr.6: 1610634-1610634 on Build GRCh38
rs9502980 Chr.6: 1610854-1610854 on Build GRCh38
rs750797555 Chr.6: 1610751-1610751 on Build GRCh38
rs104893954 Chr.6: 1610706-1610706 on Build GRCh38
rs1009903619 Chr.6: 1610847-1610847 on Build GRCh38
rs888646491 Chr.6: 1610548-1610548 on Build GRCh38
rs779318918 Chr.6: 1610744-1610744 on Build GRCh38
rs780038931 Chr.6: 1610648-1610649 on Build GRCh38
rs376255999 Chr.6: 1610886-1610886 on Build GRCh38
rs2230096 Chr.6: 1610850-1610850 on Build GRCh38
rs756196843 Chr.6: 1610526-1610534 on Build GRCh38
rs756657100 Chr.6: 1610629-1610629 on Build GRCh38
rs886039568 Chr.6: 1610701-1610701 on Build GRCh38
rs1022630517 Chr.6: 1610603-1610603 on Build GRCh38
rs538126253 Chr.6: 1610559-1610559 on Build GRCh38
rs757767235 Chr.6: 1610742-1610742 on Build GRCh38
rs775317870 Chr.6: 1610897-1610897 on Build GRCh38
rs867754327 Chr.6: 1610666-1610666 on Build GRCh38
rs527271139 Chr.6: 1610694-1610694 on Build GRCh38
rs1057519475 Chr.6: 1610761-1610761 on Build GRCh38
rs144066933 Chr.6: 1610883-1610883 on Build GRCh38
rs761549150 Chr.6: 1610599-1610599 on Build GRCh38
rs112885689 Chr.6: 1610589-1610589 on Build GRCh38
rs773527470 Chr.6: 1610946-1610946 on Build GRCh38
rs762644463 Chr.6: 1610665-1610665 on Build GRCh38
rs757318312 Chr.6: 1610990-1610990 on Build GRCh38
rs749708674 Chr.6: 1610539-1610539 on Build GRCh38
rs545478008 Chr.6: 1610523-1610523 on Build GRCh38
rs917382067 Chr.6: 1610770-1610770 on Build GRCh38
rs768741176 Chr.6: 1610875-1610877 on Build GRCh38
rs533710669 Chr.6: 1610643-1610643 on Build GRCh38
rs777521814 Chr.6: 1610863-1610863 on Build GRCh38
rs121909339 Chr.6: 1610803-1610803 on Build GRCh38
rs749833156 Chr.6: 1610631-1610631 on Build GRCh38
rs922369652 Chr.6: 1610976-1610976 on Build GRCh38
rs727503932 Chr.6: 1610708-1610708 on Build GRCh38
rs760588139 Chr.6: 1610580-1610580 on Build GRCh38
rs751970827 Chr.6: 1610977-1610977 on Build GRCh38
rs878862284 Chr.6: 1610536-1610536 on Build GRCh38
rs765626020 Chr.6: 1610820-1610820 on Build GRCh38
rs1057519474 Chr.6: 1610714-1610714 on Build GRCh38
rs142371761 Chr.6: 1610989-1610989 on Build GRCh38
rs749259978 Chr.6: 1610867-1610867 on Build GRCh38

View Full Product Details