Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10990226-10990726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL2
Entrez Gene ID
54898
Gene Name
ELOVL fatty acid elongase 2
Gene Aliases
SSC2
UniGene
Hs.656436
Gene Chromosome Location
Chr.6: 10980759-11044391 on Build GRCh38
Transcripts
NM_017770.3 XM_011514716.2 XM_011514717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3941458 c.554C>T p.S185F Chr.6: 10990394-10990394 on Build GRCh38
7153620 c.506G>T p.S169I Chr.6: 10990442-10990442 on Build GRCh38
4160116 c.586C>A p.H196N Chr.6: 10990362-10990362 on Build GRCh38
1072054 c.555C>T p.S185S Chr.6: 10990393-10990393 on Build GRCh38
5560784 c.526A>C p.N176H Chr.6: 10990422-10990422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745607092 Chr.6: 10990388-10990388 on Build GRCh38
rs548506008 Chr.6: 10990631-10990631 on Build GRCh38
rs764110484 Chr.6: 10990414-10990414 on Build GRCh38
rs1027895778 Chr.6: 10990478-10990478 on Build GRCh38
rs750684897 Chr.6: 10990378-10990379 on Build GRCh38
rs372558428 Chr.6: 10990667-10990668 on Build GRCh38
rs994985736 Chr.6: 10990675-10990675 on Build GRCh38
rs890195573 Chr.6: 10990420-10990420 on Build GRCh38
rs79871644 Chr.6: 10990675-10990676 on Build GRCh38
rs960027356 Chr.6: 10990655-10990656 on Build GRCh38
rs879098952 Chr.6: 10990252-10990252 on Build GRCh38
rs12211241 Chr.6: 10990609-10990609 on Build GRCh38
rs537696788 Chr.6: 10990671-10990671 on Build GRCh38
rs911196 Chr.6: 10990518-10990518 on Build GRCh38
rs1022983749 Chr.6: 10990677-10990677 on Build GRCh38
rs79799512 Chr.6: 10990492-10990492 on Build GRCh38
rs761984680 Chr.6: 10990418-10990418 on Build GRCh38
rs928895168 Chr.6: 10990622-10990622 on Build GRCh38
rs5874300 Chr.6: 10990673-10990674 on Build GRCh38
rs916037501 Chr.6: 10990378-10990378 on Build GRCh38
rs762790029 Chr.6: 10990405-10990405 on Build GRCh38
rs963763301 Chr.6: 10990685-10990686 on Build GRCh38
rs775495586 Chr.6: 10990401-10990401 on Build GRCh38
rs761814847 Chr.6: 10990499-10990501 on Build GRCh38
rs750623282 Chr.6: 10990446-10990446 on Build GRCh38
rs944428042 Chr.6: 10990669-10990669 on Build GRCh38
rs141945775 Chr.6: 10990509-10990509 on Build GRCh38
rs980881304 Chr.6: 10990258-10990258 on Build GRCh38
rs768804660 Chr.6: 10990278-10990279 on Build GRCh38
rs765222989 Chr.6: 10990276-10990276 on Build GRCh38
rs57121885 Chr.6: 10990677-10990678 on Build GRCh38
rs528835867 Chr.6: 10990606-10990606 on Build GRCh38
rs904228539 Chr.6: 10990337-10990337 on Build GRCh38
rs774514880 Chr.6: 10990634-10990634 on Build GRCh38
rs1012121313 Chr.6: 10990561-10990561 on Build GRCh38
rs143571302 Chr.6: 10990618-10990624 on Build GRCh38
rs553054329 Chr.6: 10990343-10990343 on Build GRCh38
rs375474344 Chr.6: 10990472-10990472 on Build GRCh38
rs3066153 Chr.6: 10990674-10990675 on Build GRCh38
rs879025257 Chr.6: 10990539-10990539 on Build GRCh38
rs1009072756 Chr.6: 10990423-10990423 on Build GRCh38
rs774292534 Chr.6: 10990469-10990470 on Build GRCh38
rs996456126 Chr.6: 10990667-10990667 on Build GRCh38
rs957844135 Chr.6: 10990555-10990555 on Build GRCh38
rs577263940 Chr.6: 10990270-10990270 on Build GRCh38
rs890409179 Chr.6: 10990674-10990674 on Build GRCh38
rs774613363 Chr.6: 10990282-10990286 on Build GRCh38
rs186121764 Chr.6: 10990367-10990367 on Build GRCh38
rs4602718 Chr.6: 10990309-10990309 on Build GRCh38
rs2281591 Chr.6: 10990260-10990260 on Build GRCh38
rs369426512 Chr.6: 10990620-10990626 on Build GRCh38
rs146315963 Chr.6: 10990521-10990521 on Build GRCh38
rs1015466321 Chr.6: 10990443-10990443 on Build GRCh38
rs1042717636 Chr.6: 10990336-10990336 on Build GRCh38
rs761274490 Chr.6: 10990393-10990393 on Build GRCh38
rs925871293 Chr.6: 10990605-10990605 on Build GRCh38
rs546590231 Chr.6: 10990528-10990528 on Build GRCh38
rs752321508 Chr.6: 10990493-10990495 on Build GRCh38
rs770419316 Chr.6: 10990368-10990368 on Build GRCh38
rs776451613 Chr.6: 10990372-10990372 on Build GRCh38
rs898095678 Chr.6: 10990466-10990466 on Build GRCh38
rs973620552 Chr.6: 10990672-10990673 on Build GRCh38
rs12209812 Chr.6: 10990495-10990495 on Build GRCh38
rs375561773 Chr.6: 10990646-10990646 on Build GRCh38
rs375263595 Chr.6: 10990308-10990308 on Build GRCh38
rs931684624 Chr.6: 10990617-10990617 on Build GRCh38
rs941851397 Chr.6: 10990445-10990445 on Build GRCh38
rs767678458 Chr.6: 10990444-10990444 on Build GRCh38
rs79554116 Chr.6: 10990391-10990391 on Build GRCh38
rs995057592 Chr.6: 10990470-10990470 on Build GRCh38
rs371798619 Chr.6: 10990458-10990458 on Build GRCh38
rs200429161 Chr.6: 10990320-10990320 on Build GRCh38
rs759405393 Chr.6: 10990272-10990272 on Build GRCh38
rs756370494 Chr.6: 10990473-10990473 on Build GRCh38
rs371668715 Chr.6: 10990287-10990287 on Build GRCh38
rs369537079 Chr.6: 10990364-10990364 on Build GRCh38
rs776510629 Chr.6: 10990271-10990271 on Build GRCh38
rs746594700 Chr.6: 10990358-10990358 on Build GRCh38
rs773199508 Chr.6: 10990564-10990564 on Build GRCh38
rs755573923 Chr.6: 10990350-10990350 on Build GRCh38
rs890104495 Chr.6: 10990498-10990498 on Build GRCh38
rs779673214 Chr.6: 10990286-10990286 on Build GRCh38
rs1007275276 Chr.6: 10990258-10990259 on Build GRCh38
rs78128406 Chr.6: 10990488-10990488 on Build GRCh38
rs190974394 Chr.6: 10990576-10990576 on Build GRCh38
rs570004355 Chr.6: 10990655-10990655 on Build GRCh38
rs767953842 Chr.6: 10990349-10990350 on Build GRCh38
rs367728257 Chr.6: 10990514-10990514 on Build GRCh38
rs763020685 Chr.6: 10990275-10990280 on Build GRCh38
rs774232616 Chr.6: 10990415-10990415 on Build GRCh38
rs201552624 Chr.6: 10990673-10990673 on Build GRCh38
rs762994521 Chr.6: 10990283-10990283 on Build GRCh38
rs558633380 Chr.6: 10990301-10990303 on Build GRCh38
rs931633714 Chr.6: 10990355-10990355 on Build GRCh38
rs913028666 Chr.6: 10990532-10990532 on Build GRCh38
rs766723872 Chr.6: 10990484-10990484 on Build GRCh38

View Full Product Details