Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16299854-16300364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
UniGene
Hs.434961
Gene Chromosome Location
Chr.6: 16299112-16761490 on Build GRCh38
Transcripts
NM_000332.3 NM_001128164.1

SNP Detail

SNP ID Chromosome Location
rs985307774 Chr.6: 16300335-16300335 on Build GRCh38
rs78566245 Chr.6: 16300276-16300276 on Build GRCh38
rs939167797 Chr.6: 16299896-16299897 on Build GRCh38
rs960583416 Chr.6: 16300087-16300087 on Build GRCh38
rs542603251 Chr.6: 16300091-16300091 on Build GRCh38
rs562493912 Chr.6: 16300159-16300159 on Build GRCh38
rs545532728 Chr.6: 16299934-16299934 on Build GRCh38
rs567145819 Chr.6: 16300317-16300317 on Build GRCh38
rs905921465 Chr.6: 16300309-16300309 on Build GRCh38
rs919646912 Chr.6: 16300207-16300207 on Build GRCh38
rs940942051 Chr.6: 16300007-16300007 on Build GRCh38
rs148689787 Chr.6: 16300213-16300214 on Build GRCh38
rs796707861 Chr.6: 16300188-16300189 on Build GRCh38
rs1024210367 Chr.6: 16300248-16300248 on Build GRCh38
rs753626269 Chr.6: 16300275-16300275 on Build GRCh38
rs1024241401 Chr.6: 16299906-16299906 on Build GRCh38
rs992017833 Chr.6: 16300096-16300096 on Build GRCh38
rs996493490 Chr.6: 16299958-16299958 on Build GRCh38
rs898150055 Chr.6: 16299943-16299943 on Build GRCh38
rs908374560 Chr.6: 16300148-16300151 on Build GRCh38
rs1053255104 Chr.6: 16300260-16300260 on Build GRCh38
rs766310031 Chr.6: 16300203-16300203 on Build GRCh38
rs1054268354 Chr.6: 16300150-16300150 on Build GRCh38
rs3134582 Chr.6: 16300076-16300076 on Build GRCh38
rs185079700 Chr.6: 16299895-16299895 on Build GRCh38
rs879531992 Chr.6: 16300182-16300182 on Build GRCh38
rs546889888 Chr.6: 16300279-16300279 on Build GRCh38
rs1021661607 Chr.6: 16300027-16300027 on Build GRCh38
rs760676186 Chr.6: 16300200-16300200 on Build GRCh38
rs370820656 Chr.6: 16300090-16300090 on Build GRCh38
rs981088233 Chr.6: 16299890-16299893 on Build GRCh38
rs577389367 Chr.6: 16299995-16299995 on Build GRCh38
rs557829722 Chr.6: 16300005-16300005 on Build GRCh38
rs58745226 Chr.6: 16300220-16300221 on Build GRCh38
rs1000372635 Chr.6: 16300095-16300095 on Build GRCh38
rs772700864 Chr.6: 16300052-16300052 on Build GRCh38
rs927364361 Chr.6: 16299960-16299960 on Build GRCh38
rs370389587 Chr.6: 16300234-16300234 on Build GRCh38
rs551459254 Chr.6: 16300256-16300256 on Build GRCh38
rs181034063 Chr.6: 16300062-16300062 on Build GRCh38
rs932328057 Chr.6: 16300228-16300228 on Build GRCh38
rs540056477 Chr.6: 16300038-16300038 on Build GRCh38
rs774011396 Chr.6: 16299897-16299897 on Build GRCh38
rs189092515 Chr.6: 16300012-16300012 on Build GRCh38
rs1010105336 Chr.6: 16300183-16300183 on Build GRCh38
rs531416833 Chr.6: 16300189-16300189 on Build GRCh38
rs891956955 Chr.6: 16300272-16300272 on Build GRCh38
rs761320134 Chr.6: 16299956-16299956 on Build GRCh38
rs766958927 Chr.6: 16299957-16299957 on Build GRCh38
rs145174144 Chr.6: 16300041-16300041 on Build GRCh38
rs891407727 Chr.6: 16300236-16300236 on Build GRCh38
rs774486628 Chr.6: 16300205-16300206 on Build GRCh38
rs891354303 Chr.6: 16299982-16299982 on Build GRCh38
rs750745177 Chr.6: 16300073-16300073 on Build GRCh38
rs564998177 Chr.6: 16300257-16300257 on Build GRCh38
rs971409900 Chr.6: 16299912-16299912 on Build GRCh38
rs930723532 Chr.6: 16299962-16299962 on Build GRCh38
rs944275777 Chr.6: 16300249-16300249 on Build GRCh38
rs762922908 Chr.6: 16299929-16299930 on Build GRCh38
rs952133558 Chr.6: 16300321-16300321 on Build GRCh38
rs891645840 Chr.6: 16300172-16300172 on Build GRCh38

View Full Product Details