Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98905185-98905715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL4
Entrez Gene ID
26235
Gene Name
F-box and leucine rich repeat protein 4
Gene Aliases
FBL4, FBL5, MTDPS13
UniGene
Hs.536850
Gene Chromosome Location
Chr.6: 98868535-98948006 on Build GRCh38
Transcripts
NM_001278716.1 NM_012160.4 NR_103836.1 NR_103837.1 XM_005266930.2 XM_011535748.2 XM_017010726.1 XM_017010727.1 XM_017010728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950544 c.929_933delTGAGC p.L310fs*5 Chr.6: 98905596-98905600 on Build GRCh38
4457503 c.1048C>T p.L350F Chr.6: 98905481-98905481 on Build GRCh38
1737347 c.998A>G p.D333G Chr.6: 98905531-98905531 on Build GRCh38
4430322 c.938A>T p.H313L Chr.6: 98905591-98905591 on Build GRCh38
3630715 c.1085C>T p.S362F Chr.6: 98905444-98905444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780707718 Chr.6: 98905504-98905504 on Build GRCh38
rs759851318 Chr.6: 98905541-98905541 on Build GRCh38
rs757780723 Chr.6: 98905610-98905610 on Build GRCh38
rs776516268 Chr.6: 98905638-98905638 on Build GRCh38
rs368029297 Chr.6: 98905446-98905446 on Build GRCh38
rs115004708 Chr.6: 98905606-98905606 on Build GRCh38
rs182076681 Chr.6: 98905413-98905413 on Build GRCh38
rs759753713 Chr.6: 98905442-98905442 on Build GRCh38
rs534670512 Chr.6: 98905585-98905585 on Build GRCh38
rs373555765 Chr.6: 98905491-98905491 on Build GRCh38
rs368965675 Chr.6: 98905671-98905671 on Build GRCh38
rs1028748836 Chr.6: 98905469-98905469 on Build GRCh38
rs953154129 Chr.6: 98905300-98905300 on Build GRCh38
rs931878277 Chr.6: 98905308-98905308 on Build GRCh38
rs750183150 Chr.6: 98905394-98905394 on Build GRCh38
rs372544390 Chr.6: 98905456-98905456 on Build GRCh38
rs751310167 Chr.6: 98905580-98905580 on Build GRCh38
rs766177758 Chr.6: 98905576-98905576 on Build GRCh38
rs747657121 Chr.6: 98905509-98905509 on Build GRCh38
rs539260316 Chr.6: 98905658-98905658 on Build GRCh38
rs997149850 Chr.6: 98905239-98905239 on Build GRCh38
rs765148228 Chr.6: 98905568-98905568 on Build GRCh38
rs747126681 Chr.6: 98905628-98905628 on Build GRCh38
rs772370168 Chr.6: 98905547-98905547 on Build GRCh38
rs762902348 Chr.6: 98905650-98905650 on Build GRCh38
rs762333324 Chr.6: 98905507-98905508 on Build GRCh38
rs984719896 Chr.6: 98905507-98905507 on Build GRCh38
rs1045218869 Chr.6: 98905392-98905392 on Build GRCh38
rs749668327 Chr.6: 98905538-98905538 on Build GRCh38
rs774697942 Chr.6: 98905426-98905426 on Build GRCh38
rs771325864 Chr.6: 98905419-98905419 on Build GRCh38
rs908867366 Chr.6: 98905323-98905323 on Build GRCh38
rs745762480 Chr.6: 98905389-98905389 on Build GRCh38
rs762795927 Chr.6: 98905395-98905396 on Build GRCh38
rs574016248 Chr.6: 98905364-98905364 on Build GRCh38
rs779748858 Chr.6: 98905473-98905473 on Build GRCh38
rs372150490 Chr.6: 98905592-98905592 on Build GRCh38
rs775198265 Chr.6: 98905396-98905397 on Build GRCh38
rs754570800 Chr.6: 98905502-98905502 on Build GRCh38
rs999441893 Chr.6: 98905643-98905643 on Build GRCh38
rs1054028380 Chr.6: 98905649-98905649 on Build GRCh38
rs571805954 Chr.6: 98905616-98905616 on Build GRCh38
rs1019681477 Chr.6: 98905244-98905244 on Build GRCh38
rs368886622 Chr.6: 98905540-98905540 on Build GRCh38
rs977577937 Chr.6: 98905383-98905383 on Build GRCh38
rs767129948 Chr.6: 98905676-98905676 on Build GRCh38
rs984621399 Chr.6: 98905448-98905448 on Build GRCh38
rs897725913 Chr.6: 98905236-98905236 on Build GRCh38
rs772461568 Chr.6: 98905625-98905625 on Build GRCh38
rs904248709 Chr.6: 98905631-98905631 on Build GRCh38
rs142814111 Chr.6: 98905593-98905593 on Build GRCh38
rs199540184 Chr.6: 98905460-98905460 on Build GRCh38
rs200701723 Chr.6: 98905388-98905389 on Build GRCh38
rs764180868 Chr.6: 98905389-98905389 on Build GRCh38
rs1040983217 Chr.6: 98905673-98905673 on Build GRCh38
rs72627757 Chr.6: 98905286-98905286 on Build GRCh38
rs996878291 Chr.6: 98905689-98905689 on Build GRCh38
rs985121357 Chr.6: 98905319-98905319 on Build GRCh38
rs544603895 Chr.6: 98905259-98905259 on Build GRCh38
rs367559138 Chr.6: 98905390-98905390 on Build GRCh38
rs892683064 Chr.6: 98905671-98905671 on Build GRCh38
rs377101732 Chr.6: 98905396-98905396 on Build GRCh38
rs376597046 Chr.6: 98905657-98905657 on Build GRCh38
rs766267288 Chr.6: 98905655-98905655 on Build GRCh38
rs541354990 Chr.6: 98905371-98905371 on Build GRCh38
rs748620607 Chr.6: 98905398-98905398 on Build GRCh38
rs778318524 Chr.6: 98905520-98905520 on Build GRCh38
rs933217926 Chr.6: 98905539-98905539 on Build GRCh38
rs758028668 Chr.6: 98905395-98905395 on Build GRCh38
rs201106681 Chr.6: 98905370-98905370 on Build GRCh38
rs374114762 Chr.6: 98905414-98905414 on Build GRCh38
rs762811521 Chr.6: 98905575-98905575 on Build GRCh38
rs768785974 Chr.6: 98905636-98905636 on Build GRCh38
rs1021827724 Chr.6: 98905346-98905346 on Build GRCh38
rs1007717555 Chr.6: 98905335-98905335 on Build GRCh38
rs778207313 Chr.6: 98905596-98905596 on Build GRCh38
rs778583873 Chr.6: 98905393-98905393 on Build GRCh38
rs147429704 Chr.6: 98905224-98905224 on Build GRCh38
rs761676733 Chr.6: 98905458-98905458 on Build GRCh38
rs150436385 Chr.6: 98905459-98905459 on Build GRCh38
rs965839472 Chr.6: 98905257-98905258 on Build GRCh38
rs758201051 Chr.6: 98905462-98905462 on Build GRCh38
rs6935315 Chr.6: 98905396-98905396 on Build GRCh38
rs769085791 Chr.6: 98905685-98905685 on Build GRCh38
rs754658764 Chr.6: 98905583-98905583 on Build GRCh38
rs189497833 Chr.6: 98905397-98905397 on Build GRCh38
rs775768793 Chr.6: 98905626-98905626 on Build GRCh38
rs186727069 Chr.6: 98905492-98905492 on Build GRCh38
rs115763595 Chr.6: 98905558-98905558 on Build GRCh38
rs770241351 Chr.6: 98905518-98905518 on Build GRCh38
rs1006020350 Chr.6: 98905217-98905217 on Build GRCh38
rs746193063 Chr.6: 98905623-98905623 on Build GRCh38
rs559360649 Chr.6: 98905391-98905391 on Build GRCh38
rs61744041 Chr.6: 98905551-98905551 on Build GRCh38

View Full Product Details