Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99371327-99371829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ3
Entrez Gene ID
51805
Gene Name
coenzyme Q3, methyltransferase
Gene Aliases
DHHBMT, DHHBMTASE, UG0215E05, bA9819.1
UniGene
Hs.713623
Gene Chromosome Location
Chr.6: 99369401-99394223 on Build GRCh38
Transcripts
NM_017421.3 XM_005267018.4 XM_006715500.3 XM_011535898.2 XM_011535899.1 XM_017010933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3876282 c.850T>G p.F284V Chr.6: 99371467-99371467 on Build GRCh38
5905660 c.734G>A p.G245D Chr.6: 99371583-99371583 on Build GRCh38
3080007 c.828delA p.G277fs*14 Chr.6: 99371489-99371489 on Build GRCh38
6352178 c.814A>G p.S272G Chr.6: 99371503-99371503 on Build GRCh38
3922278 c.776C>T p.S259F Chr.6: 99371541-99371541 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369408410 Chr.6: 99371585-99371585 on Build GRCh38
rs774061697 Chr.6: 99371615-99371615 on Build GRCh38
rs771451133 Chr.6: 99371594-99371594 on Build GRCh38
rs6925344 Chr.6: 99371503-99371503 on Build GRCh38
rs768030851 Chr.6: 99371456-99371456 on Build GRCh38
rs769495529 Chr.6: 99371588-99371588 on Build GRCh38
rs780179963 Chr.6: 99371387-99371387 on Build GRCh38
rs757464775 Chr.6: 99371555-99371555 on Build GRCh38
rs970940215 Chr.6: 99371492-99371492 on Build GRCh38
rs554950031 Chr.6: 99371553-99371553 on Build GRCh38
rs778904327 Chr.6: 99371560-99371560 on Build GRCh38
rs567548627 Chr.6: 99371714-99371714 on Build GRCh38
rs777856811 Chr.6: 99371464-99371464 on Build GRCh38
rs201937068 Chr.6: 99371518-99371518 on Build GRCh38
rs146934336 Chr.6: 99371430-99371430 on Build GRCh38
rs546963201 Chr.6: 99371542-99371542 on Build GRCh38
rs755498626 Chr.6: 99371454-99371454 on Build GRCh38
rs769564359 Chr.6: 99371504-99371504 on Build GRCh38
rs753177641 Chr.6: 99371546-99371546 on Build GRCh38
rs1054322690 Chr.6: 99371770-99371770 on Build GRCh38
rs532542171 Chr.6: 99371576-99371576 on Build GRCh38
rs757301529 Chr.6: 99371477-99371477 on Build GRCh38
rs768550326 Chr.6: 99371497-99371497 on Build GRCh38
rs921721304 Chr.6: 99371740-99371740 on Build GRCh38
rs4574651 Chr.6: 99371680-99371680 on Build GRCh38
rs779005475 Chr.6: 99371487-99371487 on Build GRCh38
rs756441765 Chr.6: 99371548-99371548 on Build GRCh38
rs75377761 Chr.6: 99371606-99371606 on Build GRCh38
rs762616170 Chr.6: 99371512-99371512 on Build GRCh38
rs760553286 Chr.6: 99371378-99371378 on Build GRCh38
rs778117306 Chr.6: 99371479-99371479 on Build GRCh38
rs926347558 Chr.6: 99371509-99371509 on Build GRCh38
rs888520712 Chr.6: 99371743-99371743 on Build GRCh38
rs753085923 Chr.6: 99371458-99371458 on Build GRCh38
rs900349074 Chr.6: 99371379-99371379 on Build GRCh38
rs773964144 Chr.6: 99371536-99371536 on Build GRCh38
rs1000078027 Chr.6: 99371394-99371394 on Build GRCh38
rs1049952341 Chr.6: 99371715-99371715 on Build GRCh38
rs374493939 Chr.6: 99371691-99371691 on Build GRCh38
rs563045377 Chr.6: 99371412-99371412 on Build GRCh38
rs200030843 Chr.6: 99371452-99371452 on Build GRCh38
rs373391087 Chr.6: 99371369-99371369 on Build GRCh38
rs534648150 Chr.6: 99371791-99371791 on Build GRCh38
rs376914158 Chr.6: 99371540-99371540 on Build GRCh38
rs911566175 Chr.6: 99371647-99371647 on Build GRCh38
rs891706318 Chr.6: 99371796-99371796 on Build GRCh38
rs544735651 Chr.6: 99371373-99371373 on Build GRCh38
rs373223154 Chr.6: 99371488-99371488 on Build GRCh38
rs760166226 Chr.6: 99371537-99371537 on Build GRCh38
rs772843589 Chr.6: 99371415-99371415 on Build GRCh38
rs764776152 Chr.6: 99371414-99371414 on Build GRCh38
rs922894764 Chr.6: 99371695-99371695 on Build GRCh38
rs776370725 Chr.6: 99371411-99371411 on Build GRCh38
rs760134484 Chr.6: 99371653-99371653 on Build GRCh38
rs756460754 Chr.6: 99371460-99371460 on Build GRCh38
rs983119847 Chr.6: 99371640-99371640 on Build GRCh38
rs575588530 Chr.6: 99371677-99371677 on Build GRCh38
rs750021431 Chr.6: 99371405-99371406 on Build GRCh38
rs749463341 Chr.6: 99371466-99371466 on Build GRCh38
rs772752474 Chr.6: 99371507-99371507 on Build GRCh38
rs35567090 Chr.6: 99371675-99371676 on Build GRCh38
rs551068346 Chr.6: 99371584-99371584 on Build GRCh38
rs377538236 Chr.6: 99371478-99371478 on Build GRCh38
rs770593900 Chr.6: 99371613-99371613 on Build GRCh38
rs997036103 Chr.6: 99371356-99371356 on Build GRCh38
rs935816199 Chr.6: 99371751-99371751 on Build GRCh38
rs755591858 Chr.6: 99371517-99371518 on Build GRCh38
rs137966773 Chr.6: 99371517-99371517 on Build GRCh38
rs571511490 Chr.6: 99371554-99371554 on Build GRCh38
rs145229869 Chr.6: 99371440-99371440 on Build GRCh38
rs773016310 Chr.6: 99371599-99371599 on Build GRCh38
rs768461999 Chr.6: 99371382-99371382 on Build GRCh38
rs556986614 Chr.6: 99371358-99371358 on Build GRCh38

View Full Product Details