Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121112174-121112670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D32
Entrez Gene ID
221322
Gene Name
TBC1 domain family member 32
Gene Aliases
BROMI, C6orf170, C6orf171, bA301B7.2, bA57L9.1, dJ310J6.1
UniGene
Hs.121396
Gene Chromosome Location
Chr.6: 121079494-121335382 on Build GRCh38
Transcripts
NM_152730.5 NR_104452.1 XM_005266861.2 XM_011535569.1 XM_011535570.1 XM_011535571.2 XM_011535574.1 XM_011535580.2 XM_017010397.1 XM_017010398.1 XM_017010399.1 XM_017010400.1 XM_017010401.1 XM_017010404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198939 c.3262C>A p.L1088I Chr.6: 121112567-121112567 on Build GRCh38
1072753 c.3273C>A p.F1091L Chr.6: 121112556-121112556 on Build GRCh38
6567556 c.3183G>A p.G1061G Chr.6: 121112646-121112646 on Build GRCh38
6579752 c.3285G>A p.L1095L Chr.6: 121112544-121112544 on Build GRCh38
6797431 c.3178C>T p.Q1060* Chr.6: 121112651-121112651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778807503 Chr.6: 121112510-121112510 on Build GRCh38
rs770341437 Chr.6: 121112524-121112524 on Build GRCh38
rs13195570 Chr.6: 121112415-121112415 on Build GRCh38
rs753732372 Chr.6: 121112490-121112490 on Build GRCh38
rs746242313 Chr.6: 121112644-121112644 on Build GRCh38
rs943999495 Chr.6: 121112357-121112357 on Build GRCh38
rs764125434 Chr.6: 121112486-121112486 on Build GRCh38
rs371460758 Chr.6: 121112632-121112632 on Build GRCh38
rs1054333876 Chr.6: 121112645-121112645 on Build GRCh38
rs781461289 Chr.6: 121112616-121112616 on Build GRCh38
rs1012865635 Chr.6: 121112244-121112244 on Build GRCh38
rs757083944 Chr.6: 121112506-121112506 on Build GRCh38
rs189458585 Chr.6: 121112498-121112498 on Build GRCh38
rs749695630 Chr.6: 121112537-121112537 on Build GRCh38
rs373972495 Chr.6: 121112512-121112512 on Build GRCh38
rs907219142 Chr.6: 121112304-121112304 on Build GRCh38
rs960305943 Chr.6: 121112205-121112205 on Build GRCh38
rs560574202 Chr.6: 121112571-121112571 on Build GRCh38
rs916288420 Chr.6: 121112214-121112214 on Build GRCh38
rs562122480 Chr.6: 121112403-121112403 on Build GRCh38
rs919706503 Chr.6: 121112292-121112292 on Build GRCh38
rs17083183 Chr.6: 121112373-121112373 on Build GRCh38
rs199671235 Chr.6: 121112647-121112647 on Build GRCh38
rs374619042 Chr.6: 121112447-121112447 on Build GRCh38
rs985001422 Chr.6: 121112421-121112421 on Build GRCh38
rs200575069 Chr.6: 121112545-121112545 on Build GRCh38
rs777791020 Chr.6: 121112628-121112628 on Build GRCh38
rs185501564 Chr.6: 121112220-121112220 on Build GRCh38
rs766962568 Chr.6: 121112459-121112459 on Build GRCh38
rs1026582645 Chr.6: 121112322-121112322 on Build GRCh38
rs9387920 Chr.6: 121112239-121112239 on Build GRCh38
rs929698292 Chr.6: 121112302-121112302 on Build GRCh38
rs890948368 Chr.6: 121112331-121112331 on Build GRCh38
rs750165137 Chr.6: 121112610-121112610 on Build GRCh38
rs756157845 Chr.6: 121112620-121112620 on Build GRCh38
rs991732580 Chr.6: 121112210-121112210 on Build GRCh38
rs368544834 Chr.6: 121112607-121112607 on Build GRCh38
rs755170219 Chr.6: 121112515-121112515 on Build GRCh38
rs374536754 Chr.6: 121112574-121112574 on Build GRCh38
rs375947439 Chr.6: 121112639-121112639 on Build GRCh38
rs756013656 Chr.6: 121112478-121112478 on Build GRCh38
rs201382292 Chr.6: 121112550-121112550 on Build GRCh38
rs182057696 Chr.6: 121112518-121112518 on Build GRCh38
rs760237654 Chr.6: 121112572-121112572 on Build GRCh38
rs757208624 Chr.6: 121112606-121112606 on Build GRCh38
rs774731973 Chr.6: 121112561-121112561 on Build GRCh38
rs528372763 Chr.6: 121112598-121112598 on Build GRCh38
rs764164450 Chr.6: 121112596-121112596 on Build GRCh38
rs760843065 Chr.6: 121112578-121112578 on Build GRCh38
rs748226334 Chr.6: 121112617-121112617 on Build GRCh38
rs748253784 Chr.6: 121112521-121112521 on Build GRCh38
rs1054934144 Chr.6: 121112234-121112235 on Build GRCh38
rs752197688 Chr.6: 121112474-121112474 on Build GRCh38
rs758347072 Chr.6: 121112415-121112415 on Build GRCh38
rs573451448 Chr.6: 121112311-121112311 on Build GRCh38
rs999332096 Chr.6: 121112542-121112542 on Build GRCh38
rs773689662 Chr.6: 121112525-121112525 on Build GRCh38
rs548490962 Chr.6: 121112634-121112634 on Build GRCh38
rs771090638 Chr.6: 121112578-121112579 on Build GRCh38
rs199840167 Chr.6: 121112568-121112568 on Build GRCh38
rs759188700 Chr.6: 121112457-121112457 on Build GRCh38
rs575468470 Chr.6: 121112437-121112437 on Build GRCh38

View Full Product Details