Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154330305-154330811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B2
Entrez Gene ID
57198
Gene Name
ATPase phospholipid transporting 8B2
Gene Aliases
ATPID
UniGene
Hs.435700
Gene Chromosome Location
Chr.1: 154325541-154351307 on Build GRCh38
Transcripts
NM_001005855.1 NM_020452.3 XM_005245355.2 XM_005245356.3 XM_017001871.1 XM_017001872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022789 c.41G>T p.R14M Chr.1: 154330405-154330405 on Build GRCh38
6443182 c.136G>A p.E46K Chr.1: 154330401-154330401 on Build GRCh38
5547620 c.32-2_32delAGA p.? Chr.1: 154330394-154330396 on Build GRCh38
5547621 c.89-2_89delAGA p.? Chr.1: 154330394-154330396 on Build GRCh38
423987 c.33A>C p.E11D Chr.1: 154330397-154330397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949337826 Chr.1: 154330367-154330367 on Build GRCh38
rs772506272 Chr.1: 154330382-154330382 on Build GRCh38
rs761401717 Chr.1: 154330394-154330396 on Build GRCh38
rs12735225 Chr.1: 154330418-154330418 on Build GRCh38
rs370461095 Chr.1: 154330524-154330524 on Build GRCh38
rs551271217 Chr.1: 154330341-154330341 on Build GRCh38
rs949017669 Chr.1: 154330492-154330492 on Build GRCh38
rs943432892 Chr.1: 154330349-154330349 on Build GRCh38
rs549106510 Chr.1: 154330546-154330546 on Build GRCh38
rs368910102 Chr.1: 154330411-154330411 on Build GRCh38
rs780067113 Chr.1: 154330353-154330353 on Build GRCh38
rs200836965 Chr.1: 154330453-154330453 on Build GRCh38
rs115165870 Chr.1: 154330752-154330752 on Build GRCh38
rs779300153 Chr.1: 154330347-154330347 on Build GRCh38
rs748400109 Chr.1: 154330376-154330376 on Build GRCh38
rs766554991 Chr.1: 154330501-154330501 on Build GRCh38
rs200631375 Chr.1: 154330358-154330358 on Build GRCh38
rs964688854 Chr.1: 154330686-154330686 on Build GRCh38
rs762392413 Chr.1: 154330460-154330460 on Build GRCh38
rs752629526 Chr.1: 154330610-154330610 on Build GRCh38
rs978813327 Chr.1: 154330664-154330664 on Build GRCh38
rs771740360 Chr.1: 154330422-154330422 on Build GRCh38
rs926108295 Chr.1: 154330366-154330366 on Build GRCh38
rs750799061 Chr.1: 154330474-154330474 on Build GRCh38
rs981966466 Chr.1: 154330723-154330723 on Build GRCh38
rs1008788748 Chr.1: 154330649-154330649 on Build GRCh38
rs74976887 Chr.1: 154330768-154330768 on Build GRCh38
rs758618879 Chr.1: 154330351-154330351 on Build GRCh38
rs779797404 Chr.1: 154330758-154330758 on Build GRCh38
rs116322496 Chr.1: 154330673-154330673 on Build GRCh38
rs910568007 Chr.1: 154330335-154330335 on Build GRCh38
rs898938619 Chr.1: 154330473-154330473 on Build GRCh38
rs760197000 Chr.1: 154330432-154330432 on Build GRCh38
rs774164452 Chr.1: 154330412-154330412 on Build GRCh38
rs757656345 Chr.1: 154330346-154330346 on Build GRCh38
rs759176827 Chr.1: 154330414-154330414 on Build GRCh38
rs11265553 Chr.1: 154330692-154330692 on Build GRCh38
rs12077882 Chr.1: 154330417-154330417 on Build GRCh38
rs765564496 Chr.1: 154330464-154330464 on Build GRCh38
rs6691345 Chr.1: 154330350-154330350 on Build GRCh38
rs564127917 Chr.1: 154330663-154330663 on Build GRCh38
rs751620019 Chr.1: 154330764-154330764 on Build GRCh38
rs916380630 Chr.1: 154330713-154330713 on Build GRCh38
rs374528384 Chr.1: 154330386-154330386 on Build GRCh38
rs1046067052 Chr.1: 154330504-154330504 on Build GRCh38
rs754348764 Chr.1: 154330454-154330454 on Build GRCh38
rs567252138 Chr.1: 154330558-154330558 on Build GRCh38
rs367919166 Chr.1: 154330716-154330716 on Build GRCh38
rs993762689 Chr.1: 154330601-154330601 on Build GRCh38
rs747041374 Chr.1: 154330708-154330708 on Build GRCh38
rs771489863 Chr.1: 154330576-154330576 on Build GRCh38
rs373621889 Chr.1: 154330483-154330483 on Build GRCh38
rs769724099 Chr.1: 154330364-154330364 on Build GRCh38
rs769483784 Chr.1: 154330488-154330491 on Build GRCh38
rs759051331 Chr.1: 154330687-154330687 on Build GRCh38
rs11430764 Chr.1: 154330377-154330378 on Build GRCh38
rs898997644 Chr.1: 154330600-154330600 on Build GRCh38
rs149978531 Chr.1: 154330426-154330426 on Build GRCh38
rs1026217931 Chr.1: 154330603-154330603 on Build GRCh38
rs907814144 Chr.1: 154330741-154330741 on Build GRCh38
rs754998967 Chr.1: 154330767-154330767 on Build GRCh38
rs758609136 Chr.1: 154330498-154330498 on Build GRCh38
rs777643963 Chr.1: 154330380-154330380 on Build GRCh38
rs1051310318 Chr.1: 154330650-154330681 on Build GRCh38
rs1037411520 Chr.1: 154330763-154330763 on Build GRCh38
rs767378065 Chr.1: 154330598-154330598 on Build GRCh38
rs373800163 Chr.1: 154330780-154330780 on Build GRCh38
rs989411193 Chr.1: 154330715-154330715 on Build GRCh38

View Full Product Details