Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143497308-143497809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA2
Entrez Gene ID
2519
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
dJ20N2.5
UniGene
Hs.17680
Gene Chromosome Location
Chr.6: 143494812-143511883 on Build GRCh38
Transcripts
NM_032020.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448768 c.1263+2T>C p.? Chr.6: 143497387-143497387 on Build GRCh38
4875552 c.236T>G p.L79R Chr.6: 143497453-143497453 on Build GRCh38
1074055 c.1199T>G p.L400R Chr.6: 143497453-143497453 on Build GRCh38
1074054 c.1206G>A p.W402* Chr.6: 143497446-143497446 on Build GRCh38
4867773 c.243G>A p.W81* Chr.6: 143497446-143497446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890619916 Chr.6: 143497724-143497724 on Build GRCh38
rs942010041 Chr.6: 143497617-143497617 on Build GRCh38
rs752451979 Chr.6: 143497421-143497421 on Build GRCh38
rs550604414 Chr.6: 143497580-143497580 on Build GRCh38
rs1019413704 Chr.6: 143497400-143497400 on Build GRCh38
rs755819872 Chr.6: 143497424-143497424 on Build GRCh38
rs768512750 Chr.6: 143497509-143497509 on Build GRCh38
rs753227229 Chr.6: 143497443-143497443 on Build GRCh38
rs892797781 Chr.6: 143497670-143497670 on Build GRCh38
rs866162357 Chr.6: 143497380-143497380 on Build GRCh38
rs763661877 Chr.6: 143497441-143497441 on Build GRCh38
rs767482755 Chr.6: 143497523-143497523 on Build GRCh38
rs990622250 Chr.6: 143497527-143497527 on Build GRCh38
rs780383907 Chr.6: 143497464-143497464 on Build GRCh38
rs770935093 Chr.6: 143497508-143497508 on Build GRCh38
rs1049862350 Chr.6: 143497743-143497743 on Build GRCh38
rs774199575 Chr.6: 143497510-143497510 on Build GRCh38
rs767393003 Chr.6: 143497411-143497411 on Build GRCh38
rs777994840 Chr.6: 143497388-143497388 on Build GRCh38
rs535701108 Chr.6: 143497539-143497539 on Build GRCh38
rs201539810 Chr.6: 143497476-143497476 on Build GRCh38
rs748020087 Chr.6: 143497513-143497516 on Build GRCh38
rs879941392 Chr.6: 143497786-143497786 on Build GRCh38
rs576955674 Chr.6: 143497689-143497689 on Build GRCh38
rs543571091 Chr.6: 143497619-143497619 on Build GRCh38
rs753417970 Chr.6: 143497544-143497544 on Build GRCh38
rs766171853 Chr.6: 143497376-143497376 on Build GRCh38
rs919907705 Chr.6: 143497524-143497524 on Build GRCh38
rs781523105 Chr.6: 143497491-143497491 on Build GRCh38
rs773356326 Chr.6: 143497500-143497500 on Build GRCh38
rs935553416 Chr.6: 143497637-143497637 on Build GRCh38
rs114279752 Chr.6: 143497742-143497742 on Build GRCh38
rs748436332 Chr.6: 143497497-143497497 on Build GRCh38
rs369469343 Chr.6: 143497526-143497526 on Build GRCh38
rs746114736 Chr.6: 143497506-143497507 on Build GRCh38
rs774227181 Chr.6: 143497385-143497385 on Build GRCh38
rs747481878 Chr.6: 143497466-143497466 on Build GRCh38
rs954008499 Chr.6: 143497336-143497336 on Build GRCh38
rs748348067 Chr.6: 143497338-143497338 on Build GRCh38
rs141552353 Chr.6: 143497610-143497610 on Build GRCh38
rs889509326 Chr.6: 143497752-143497752 on Build GRCh38
rs111404250 Chr.6: 143497529-143497529 on Build GRCh38
rs751579324 Chr.6: 143497410-143497410 on Build GRCh38
rs762953761 Chr.6: 143497359-143497359 on Build GRCh38
rs1054070092 Chr.6: 143497658-143497658 on Build GRCh38
rs757396957 Chr.6: 143497479-143497479 on Build GRCh38
rs759368449 Chr.6: 143497404-143497404 on Build GRCh38
rs755385060 Chr.6: 143497486-143497486 on Build GRCh38
rs996403234 Chr.6: 143497659-143497659 on Build GRCh38
rs1008405882 Chr.6: 143497755-143497755 on Build GRCh38
rs778852364 Chr.6: 143497512-143497516 on Build GRCh38
rs558612535 Chr.6: 143497642-143497642 on Build GRCh38
rs758965632 Chr.6: 143497454-143497454 on Build GRCh38
rs546664734 Chr.6: 143497348-143497348 on Build GRCh38
rs961248488 Chr.6: 143497444-143497444 on Build GRCh38
rs759528083 Chr.6: 143497522-143497522 on Build GRCh38
rs553288492 Chr.6: 143497782-143497782 on Build GRCh38
rs966462144 Chr.6: 143497770-143497770 on Build GRCh38
rs916631203 Chr.6: 143497608-143497608 on Build GRCh38
rs1019381569 Chr.6: 143497778-143497778 on Build GRCh38
rs199878021 Chr.6: 143497343-143497343 on Build GRCh38
rs949508601 Chr.6: 143497609-143497609 on Build GRCh38
rs1020666348 Chr.6: 143497363-143497363 on Build GRCh38
rs781228770 Chr.6: 143497505-143497506 on Build GRCh38

View Full Product Details