Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26213302-26213803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX3
Entrez Gene ID
11335
Gene Name
chromobox 3
Gene Aliases
HECH, HP1-GAMMA, HP1Hs-gamma
UniGene
Hs.381189
Gene Chromosome Location
Chr.7: 26201211-26213607 on Build GRCh38
Transcripts
NM_007276.4 NM_016587.3 XM_005249611.3

SNP Detail

SNP ID Chromosome Location
rs893273027 Chr.7: 26213637-26213637 on Build GRCh38
rs1038261658 Chr.7: 26213529-26213529 on Build GRCh38
rs771993111 Chr.7: 26213726-26213726 on Build GRCh38
rs774467142 Chr.7: 26213569-26213569 on Build GRCh38
rs932684222 Chr.7: 26213697-26213697 on Build GRCh38
rs1055823831 Chr.7: 26213772-26213772 on Build GRCh38
rs1042475867 Chr.7: 26213756-26213756 on Build GRCh38
rs767085030 Chr.7: 26213741-26213742 on Build GRCh38
rs1042589781 Chr.7: 26213401-26213401 on Build GRCh38
rs1026570331 Chr.7: 26213486-26213486 on Build GRCh38
rs906901315 Chr.7: 26213576-26213576 on Build GRCh38
rs181300726 Chr.7: 26213365-26213365 on Build GRCh38
rs945460606 Chr.7: 26213745-26213745 on Build GRCh38
rs897083688 Chr.7: 26213530-26213530 on Build GRCh38
rs537161081 Chr.7: 26213512-26213512 on Build GRCh38
rs977104483 Chr.7: 26213671-26213671 on Build GRCh38
rs1024036095 Chr.7: 26213398-26213398 on Build GRCh38
rs538815981 Chr.7: 26213683-26213683 on Build GRCh38
rs376991633 Chr.7: 26213739-26213740 on Build GRCh38
rs754181630 Chr.7: 26213337-26213337 on Build GRCh38
rs895401599 Chr.7: 26213775-26213775 on Build GRCh38
rs915215517 Chr.7: 26213743-26213743 on Build GRCh38
rs71954388 Chr.7: 26213740-26213741 on Build GRCh38
rs906723893 Chr.7: 26213763-26213763 on Build GRCh38
rs142267744 Chr.7: 26213351-26213351 on Build GRCh38
rs923328061 Chr.7: 26213470-26213470 on Build GRCh38
rs1026622590 Chr.7: 26213778-26213778 on Build GRCh38
rs962494559 Chr.7: 26213718-26213718 on Build GRCh38
rs893896515 Chr.7: 26213384-26213384 on Build GRCh38
rs369134863 Chr.7: 26213385-26213385 on Build GRCh38
rs889651306 Chr.7: 26213341-26213341 on Build GRCh38
rs1012580571 Chr.7: 26213463-26213463 on Build GRCh38
rs140809582 Chr.7: 26213714-26213717 on Build GRCh38
rs111891412 Chr.7: 26213448-26213448 on Build GRCh38
rs1034507887 Chr.7: 26213423-26213423 on Build GRCh38
rs766588991 Chr.7: 26213328-26213331 on Build GRCh38
rs929933938 Chr.7: 26213574-26213574 on Build GRCh38
rs749155788 Chr.7: 26213461-26213461 on Build GRCh38
rs563164402 Chr.7: 26213354-26213354 on Build GRCh38
rs953385101 Chr.7: 26213707-26213710 on Build GRCh38
rs929874498 Chr.7: 26213734-26213734 on Build GRCh38
rs1001282371 Chr.7: 26213582-26213582 on Build GRCh38
rs372844864 Chr.7: 26213611-26213611 on Build GRCh38
rs994251777 Chr.7: 26213373-26213376 on Build GRCh38
rs939527349 Chr.7: 26213767-26213767 on Build GRCh38
rs921184762 Chr.7: 26213696-26213696 on Build GRCh38
rs141646491 Chr.7: 26213768-26213768 on Build GRCh38
rs945407009 Chr.7: 26213392-26213392 on Build GRCh38
rs867202440 Chr.7: 26213653-26213653 on Build GRCh38
rs13234120 Chr.7: 26213708-26213708 on Build GRCh38
rs568198570 Chr.7: 26213494-26213494 on Build GRCh38
rs374889372 Chr.7: 26213715-26213718 on Build GRCh38
rs911018000 Chr.7: 26213513-26213513 on Build GRCh38
rs1055521862 Chr.7: 26213758-26213758 on Build GRCh38
rs547853175 Chr.7: 26213451-26213451 on Build GRCh38
rs552155077 Chr.7: 26213465-26213465 on Build GRCh38
rs1034102549 Chr.7: 26213592-26213592 on Build GRCh38
rs967373176 Chr.7: 26213452-26213452 on Build GRCh38
rs968014324 Chr.7: 26213673-26213673 on Build GRCh38
rs13247806 Chr.7: 26213339-26213339 on Build GRCh38
rs28407029 Chr.7: 26213346-26213346 on Build GRCh38
rs909314954 Chr.7: 26213714-26213714 on Build GRCh38
rs571562692 Chr.7: 26213665-26213665 on Build GRCh38
rs557068940 Chr.7: 26213640-26213640 on Build GRCh38
rs551708454 Chr.7: 26213663-26213663 on Build GRCh38
rs1030288241 Chr.7: 26213707-26213708 on Build GRCh38
rs117232097 Chr.7: 26213776-26213776 on Build GRCh38
rs918456904 Chr.7: 26213731-26213731 on Build GRCh38
rs982467965 Chr.7: 26213556-26213556 on Build GRCh38
rs941206348 Chr.7: 26213516-26213516 on Build GRCh38
rs1001313421 Chr.7: 26213410-26213410 on Build GRCh38
rs373262978 Chr.7: 26213357-26213357 on Build GRCh38
rs928246031 Chr.7: 26213756-26213757 on Build GRCh38
rs115829318 Chr.7: 26213449-26213449 on Build GRCh38

View Full Product Details